Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 27368826-27369321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LGR4
Entrez Gene ID
55366
Gene Name
leucine rich repeat containing G protein-coupled receptor 4
Gene Aliases
BNMD17, GPR48
UniGene
Hs.502176
Gene Chromosome Location
Chr.11: 27365961-27472787 on Build GRCh38
Transcripts
NM_018490.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6262809 c.1815C>A p.F605L Chr.11: 27368908-27368908 on Build GRCh38
4032211 c.1715C>T p.S572L Chr.11: 27369008-27369008 on Build GRCh38
5568647 c.1848T>C p.S616S Chr.11: 27368875-27368875 on Build GRCh38
1353334 c.1759A>G p.M587V Chr.11: 27368964-27368964 on Build GRCh38
1638804 c.1580-3delT p.? Chr.11: 27369146-27369146 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758122005 Chr.11: 27368936-27368936 on Build GRCh38
rs35947785 Chr.11: 27368941-27368941 on Build GRCh38
rs774510969 Chr.11: 27368985-27368985 on Build GRCh38
rs765192487 Chr.11: 27369012-27369012 on Build GRCh38
rs746797439 Chr.11: 27369145-27369146 on Build GRCh38
rs190007582 Chr.11: 27369007-27369007 on Build GRCh38
rs200626048 Chr.11: 27369098-27369098 on Build GRCh38
rs1015047925 Chr.11: 27369073-27369073 on Build GRCh38
rs777772234 Chr.11: 27369146-27369146 on Build GRCh38
rs766082558 Chr.11: 27369165-27369165 on Build GRCh38
rs748467080 Chr.11: 27368877-27368877 on Build GRCh38
rs757925752 Chr.11: 27368856-27368856 on Build GRCh38
rs769838846 Chr.11: 27369161-27369161 on Build GRCh38
rs758259278 Chr.11: 27369144-27369145 on Build GRCh38
rs538451022 Chr.11: 27369038-27369038 on Build GRCh38
rs759199286 Chr.11: 27369179-27369179 on Build GRCh38
rs994850775 Chr.11: 27369243-27369243 on Build GRCh38
rs1014136174 Chr.11: 27369216-27369216 on Build GRCh38
rs368429795 Chr.11: 27369160-27369160 on Build GRCh38
rs376059218 Chr.11: 27369038-27369040 on Build GRCh38
rs1049711882 Chr.11: 27369266-27369266 on Build GRCh38
rs778459302 Chr.11: 27369105-27369105 on Build GRCh38
rs779418604 Chr.11: 27368864-27368864 on Build GRCh38
rs373408874 Chr.11: 27368902-27368902 on Build GRCh38
rs745335684 Chr.11: 27369161-27369162 on Build GRCh38
rs372583712 Chr.11: 27369162-27369162 on Build GRCh38
rs780741417 Chr.11: 27369153-27369154 on Build GRCh38
rs756869357 Chr.11: 27369152-27369153 on Build GRCh38
rs763888757 Chr.11: 27368908-27368908 on Build GRCh38
rs751116300 Chr.11: 27369042-27369042 on Build GRCh38
rs371950786 Chr.11: 27369120-27369120 on Build GRCh38
rs372955496 Chr.11: 27368977-27368977 on Build GRCh38
rs762583974 Chr.11: 27369163-27369163 on Build GRCh38
rs761634295 Chr.11: 27368918-27368918 on Build GRCh38
rs781480626 Chr.11: 27368973-27368973 on Build GRCh38
rs761830490 Chr.11: 27369008-27369008 on Build GRCh38
rs1046561459 Chr.11: 27369226-27369226 on Build GRCh38
rs778104650 Chr.11: 27368978-27368978 on Build GRCh38
rs759494168 Chr.11: 27368986-27368986 on Build GRCh38
rs777106298 Chr.11: 27368867-27368867 on Build GRCh38
rs745525143 Chr.11: 27368980-27368980 on Build GRCh38
rs1036281201 Chr.11: 27369218-27369218 on Build GRCh38
rs1052104629 Chr.11: 27368878-27368878 on Build GRCh38
rs772091571 Chr.11: 27369126-27369126 on Build GRCh38
rs770003664 Chr.11: 27368880-27368880 on Build GRCh38
rs545742704 Chr.11: 27369001-27369001 on Build GRCh38
rs780736159 Chr.11: 27369071-27369071 on Build GRCh38
rs1049750271 Chr.11: 27369268-27369272 on Build GRCh38
rs943781624 Chr.11: 27369211-27369211 on Build GRCh38
rs369900849 Chr.11: 27368939-27368939 on Build GRCh38
rs760715826 Chr.11: 27368907-27368907 on Build GRCh38
rs754383451 Chr.11: 27369056-27369056 on Build GRCh38
rs894623359 Chr.11: 27369285-27369285 on Build GRCh38
rs182255585 Chr.11: 27369152-27369152 on Build GRCh38
rs12291757 Chr.11: 27368929-27368929 on Build GRCh38
rs763145885 Chr.11: 27368895-27368895 on Build GRCh38
rs771215153 Chr.11: 27369109-27369109 on Build GRCh38
rs773360974 Chr.11: 27368892-27368892 on Build GRCh38
rs750215283 Chr.11: 27369030-27369030 on Build GRCh38
rs778935349 Chr.11: 27369117-27369117 on Build GRCh38
rs944705168 Chr.11: 27369215-27369215 on Build GRCh38
rs952168578 Chr.11: 27369039-27369039 on Build GRCh38
rs771062877 Chr.11: 27368896-27368896 on Build GRCh38
rs768642910 Chr.11: 27369003-27369003 on Build GRCh38
rs755535013 Chr.11: 27368945-27368945 on Build GRCh38
rs532090983 Chr.11: 27369240-27369240 on Build GRCh38
rs752717825 Chr.11: 27369100-27369101 on Build GRCh38
rs536392836 Chr.11: 27369239-27369239 on Build GRCh38
rs775679523 Chr.11: 27369158-27369158 on Build GRCh38
rs377257576 Chr.11: 27369167-27369167 on Build GRCh38
rs769343247 Chr.11: 27369171-27369171 on Build GRCh38
rs747539328 Chr.11: 27369131-27369131 on Build GRCh38
rs560180132 Chr.11: 27368865-27368865 on Build GRCh38
rs769015281 Chr.11: 27368866-27368866 on Build GRCh38
rs765241487 Chr.11: 27369034-27369036 on Build GRCh38
rs111618778 Chr.11: 27368972-27368972 on Build GRCh38
rs749550574 Chr.11: 27368979-27368979 on Build GRCh38
rs762745985 Chr.11: 27369036-27369036 on Build GRCh38
rs200476014 Chr.11: 27368859-27368859 on Build GRCh38
rs377568358 Chr.11: 27368916-27368916 on Build GRCh38

View Full Product Details