Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65866630-65867132 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFEMP2
Entrez Gene ID
30008
Gene Name
EGF containing fibulin like extracellular matrix protein 2
Gene Aliases
ARCL1B, FBLN4, MBP1, UPH1
UniGene
Hs.731454
Gene Chromosome Location
Chr.11: 65866441-65872934 on Build GRCh38
Transcripts
NM_016938.4 NR_037718.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1158535 c.1208G>A p.R403Q Chr.11: 65867042-65867042 on Build GRCh38
6651972 c.1188C>T p.S396S Chr.11: 65867062-65867062 on Build GRCh38
5970294 c.1270A>G p.M424V Chr.11: 65866980-65866980 on Build GRCh38
4680312 c.1212G>A p.P404P Chr.11: 65867038-65867038 on Build GRCh38
1676189 c.1211C>T p.P404L Chr.11: 65867039-65867039 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748635286 Chr.11: 65867002-65867002 on Build GRCh38
rs747521684 Chr.11: 65866904-65866904 on Build GRCh38
rs757637822 Chr.11: 65867042-65867042 on Build GRCh38
rs776539932 Chr.11: 65866722-65866722 on Build GRCh38
rs752636416 Chr.11: 65866711-65866711 on Build GRCh38
rs537197894 Chr.11: 65866794-65866794 on Build GRCh38
rs1056545955 Chr.11: 65866971-65866971 on Build GRCh38
rs779595425 Chr.11: 65866998-65866998 on Build GRCh38
rs754584204 Chr.11: 65866781-65866781 on Build GRCh38
rs974586100 Chr.11: 65866769-65866769 on Build GRCh38
rs11546251 Chr.11: 65866934-65866934 on Build GRCh38
rs1033270562 Chr.11: 65867091-65867091 on Build GRCh38
rs951322003 Chr.11: 65866728-65866728 on Build GRCh38
rs543697634 Chr.11: 65867098-65867098 on Build GRCh38
rs893023041 Chr.11: 65867096-65867096 on Build GRCh38
rs528615507 Chr.11: 65866675-65866675 on Build GRCh38
rs964150130 Chr.11: 65867100-65867100 on Build GRCh38
rs201135561 Chr.11: 65866753-65866753 on Build GRCh38
rs201815777 Chr.11: 65867110-65867110 on Build GRCh38
rs769212889 Chr.11: 65866827-65866827 on Build GRCh38
rs745886179 Chr.11: 65866685-65866686 on Build GRCh38
rs755250789 Chr.11: 65866996-65866996 on Build GRCh38
rs780449919 Chr.11: 65866814-65866814 on Build GRCh38
rs946630454 Chr.11: 65866805-65866805 on Build GRCh38
rs758608227 Chr.11: 65867012-65867012 on Build GRCh38
rs771139031 Chr.11: 65866760-65866760 on Build GRCh38
rs61893867 Chr.11: 65867024-65867024 on Build GRCh38
rs748261483 Chr.11: 65866845-65866845 on Build GRCh38
rs765059134 Chr.11: 65866673-65866673 on Build GRCh38
rs542945122 Chr.11: 65866654-65866654 on Build GRCh38
rs552721291 Chr.11: 65866925-65866925 on Build GRCh38
rs563662567 Chr.11: 65867107-65867107 on Build GRCh38
rs769542611 Chr.11: 65866872-65866872 on Build GRCh38
rs923861102 Chr.11: 65866843-65866843 on Build GRCh38
rs1055767498 Chr.11: 65866812-65866812 on Build GRCh38
rs146879673 Chr.11: 65867016-65867016 on Build GRCh38
rs780857347 Chr.11: 65866797-65866797 on Build GRCh38
rs986808252 Chr.11: 65866731-65866731 on Build GRCh38
rs755789962 Chr.11: 65866972-65866972 on Build GRCh38
rs548693216 Chr.11: 65866758-65866758 on Build GRCh38
rs774130978 Chr.11: 65867101-65867101 on Build GRCh38
rs3182964 Chr.11: 65867035-65867035 on Build GRCh38
rs754245606 Chr.11: 65866980-65866980 on Build GRCh38
rs920823319 Chr.11: 65866822-65866822 on Build GRCh38
rs759435892 Chr.11: 65866660-65866660 on Build GRCh38
rs779505726 Chr.11: 65866836-65866836 on Build GRCh38
rs570619088 Chr.11: 65866869-65866869 on Build GRCh38
rs772435021 Chr.11: 65866653-65866653 on Build GRCh38
rs772239460 Chr.11: 65866849-65866849 on Build GRCh38
rs756711623 Chr.11: 65866757-65866757 on Build GRCh38
rs762945459 Chr.11: 65866744-65866744 on Build GRCh38
rs374950957 Chr.11: 65867047-65867047 on Build GRCh38
rs781514127 Chr.11: 65867078-65867078 on Build GRCh38
rs768555558 Chr.11: 65866752-65866753 on Build GRCh38
rs1043468 Chr.11: 65866678-65866678 on Build GRCh38
rs886048505 Chr.11: 65866915-65866915 on Build GRCh38
rs372299119 Chr.11: 65867033-65867033 on Build GRCh38
rs775534736 Chr.11: 65866733-65866734 on Build GRCh38
rs778365796 Chr.11: 65867057-65867057 on Build GRCh38
rs535211927 Chr.11: 65866961-65866961 on Build GRCh38
rs756480457 Chr.11: 65866804-65866804 on Build GRCh38
rs568308652 Chr.11: 65866774-65866774 on Build GRCh38
rs765684268 Chr.11: 65867043-65867043 on Build GRCh38
rs775357834 Chr.11: 65866889-65866889 on Build GRCh38
rs572780561 Chr.11: 65866942-65866942 on Build GRCh38
rs751085402 Chr.11: 65866753-65866753 on Build GRCh38
rs966744584 Chr.11: 65866740-65866740 on Build GRCh38
rs766995796 Chr.11: 65866970-65866970 on Build GRCh38
rs193238913 Chr.11: 65866974-65866974 on Build GRCh38
rs773616908 Chr.11: 65866867-65866867 on Build GRCh38
rs755411303 Chr.11: 65867040-65867040 on Build GRCh38
rs761527042 Chr.11: 65866877-65866877 on Build GRCh38
rs539438848 Chr.11: 65866895-65866895 on Build GRCh38
rs2234473 Chr.11: 65867062-65867062 on Build GRCh38
rs749135888 Chr.11: 65867021-65867021 on Build GRCh38
rs746051296 Chr.11: 65867086-65867086 on Build GRCh38
rs767083801 Chr.11: 65867029-65867029 on Build GRCh38
rs187686630 Chr.11: 65866803-65866803 on Build GRCh38
rs763872698 Chr.11: 65866748-65866748 on Build GRCh38
rs34445533 Chr.11: 65866893-65866894 on Build GRCh38
rs543567156 Chr.11: 65867067-65867067 on Build GRCh38
rs761729008 Chr.11: 65867027-65867027 on Build GRCh38
rs753393039 Chr.11: 65867039-65867039 on Build GRCh38
rs193302868 Chr.11: 65867061-65867061 on Build GRCh38
rs757242078 Chr.11: 65867072-65867072 on Build GRCh38
rs772715527 Chr.11: 65867032-65867032 on Build GRCh38
rs767042101 Chr.11: 65867068-65867070 on Build GRCh38
rs747875416 Chr.11: 65867103-65867103 on Build GRCh38
rs774228818 Chr.11: 65866944-65866944 on Build GRCh38
rs780606460 Chr.11: 65866824-65866827 on Build GRCh38
rs749830598 Chr.11: 65866826-65866826 on Build GRCh38
rs769817128 Chr.11: 65866728-65866728 on Build GRCh38
rs774325189 Chr.11: 65867025-65867025 on Build GRCh38
rs768502421 Chr.11: 65867090-65867090 on Build GRCh38
rs995787355 Chr.11: 65866669-65866669 on Build GRCh38
rs776665492 Chr.11: 65867017-65867017 on Build GRCh38
rs148302546 Chr.11: 65867038-65867038 on Build GRCh38
rs930910946 Chr.11: 65866833-65866833 on Build GRCh38
rs774226032 Chr.11: 65866824-65866825 on Build GRCh38

View Full Product Details