Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66283413-66283906 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNIH2
Entrez Gene ID
254263
Gene Name
cornichon family AMPA receptor auxiliary protein 2
Gene Aliases
CNIH-2, Cnil
UniGene
Hs.437072
Gene Chromosome Location
Chr.11: 66278201-66284214 on Build GRCh38
Transcripts
NM_182553.2 NR_073078.1 NR_073079.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6257198 c.459G>T p.M153I Chr.11: 66283573-66283573 on Build GRCh38
5448242 c.468G>A p.T156T Chr.11: 66283582-66283582 on Build GRCh38
3687538 c.467C>T p.T156M Chr.11: 66283581-66283581 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs542790452 Chr.11: 66283557-66283557 on Build GRCh38
rs938969260 Chr.11: 66283446-66283446 on Build GRCh38
rs34082665 Chr.11: 66283725-66283725 on Build GRCh38
rs754671191 Chr.11: 66283526-66283526 on Build GRCh38
rs993132948 Chr.11: 66283750-66283750 on Build GRCh38
rs374309686 Chr.11: 66283726-66283726 on Build GRCh38
rs759820496 Chr.11: 66283870-66283870 on Build GRCh38
rs753649918 Chr.11: 66283638-66283638 on Build GRCh38
rs1013000701 Chr.11: 66283522-66283522 on Build GRCh38
rs534893208 Chr.11: 66283878-66283878 on Build GRCh38
rs948829504 Chr.11: 66283792-66283792 on Build GRCh38
rs764260297 Chr.11: 66283442-66283442 on Build GRCh38
rs1010485188 Chr.11: 66283455-66283455 on Build GRCh38
rs894675703 Chr.11: 66283488-66283488 on Build GRCh38
rs570157966 Chr.11: 66283756-66283756 on Build GRCh38
rs951176223 Chr.11: 66283776-66283776 on Build GRCh38
rs538794979 Chr.11: 66283786-66283786 on Build GRCh38
rs899226753 Chr.11: 66283831-66283831 on Build GRCh38
rs938922575 Chr.11: 66283749-66283749 on Build GRCh38
rs559127372 Chr.11: 66283598-66283598 on Build GRCh38
rs1020977645 Chr.11: 66283460-66283460 on Build GRCh38
rs765696656 Chr.11: 66283582-66283582 on Build GRCh38
rs779813601 Chr.11: 66283521-66283521 on Build GRCh38
rs771010222 Chr.11: 66283527-66283527 on Build GRCh38
rs957392770 Chr.11: 66283451-66283451 on Build GRCh38
rs367930521 Chr.11: 66283581-66283581 on Build GRCh38
rs919706809 Chr.11: 66283764-66283764 on Build GRCh38
rs547786843 Chr.11: 66283545-66283545 on Build GRCh38
rs142635924 Chr.11: 66283560-66283560 on Build GRCh38
rs34970605 Chr.11: 66283848-66283849 on Build GRCh38
rs776821845 Chr.11: 66283527-66283528 on Build GRCh38
rs752930600 Chr.11: 66283748-66283749 on Build GRCh38
rs1023445508 Chr.11: 66283758-66283758 on Build GRCh38
rs75417353 Chr.11: 66283630-66283630 on Build GRCh38
rs770497615 Chr.11: 66283829-66283829 on Build GRCh38
rs929401004 Chr.11: 66283843-66283843 on Build GRCh38
rs775396019 Chr.11: 66283596-66283602 on Build GRCh38
rs759506682 Chr.11: 66283527-66283530 on Build GRCh38
rs115798559 Chr.11: 66283518-66283518 on Build GRCh38
rs765229799 Chr.11: 66283544-66283545 on Build GRCh38
rs975219359 Chr.11: 66283629-66283629 on Build GRCh38
rs1010679573 Chr.11: 66283696-66283696 on Build GRCh38
rs199647150 Chr.11: 66283528-66283528 on Build GRCh38
rs536224318 Chr.11: 66283749-66283750 on Build GRCh38
rs372170161 Chr.11: 66283618-66283618 on Build GRCh38
rs1042068730 Chr.11: 66283881-66283881 on Build GRCh38
rs751735071 Chr.11: 66283445-66283445 on Build GRCh38
rs927501414 Chr.11: 66283493-66283493 on Build GRCh38
rs866931099 Chr.11: 66283590-66283590 on Build GRCh38
rs558872212 Chr.11: 66283797-66283797 on Build GRCh38
rs893194590 Chr.11: 66283877-66283877 on Build GRCh38
rs187803185 Chr.11: 66283635-66283635 on Build GRCh38
rs572354290 Chr.11: 66283859-66283859 on Build GRCh38
rs1020295885 Chr.11: 66283730-66283730 on Build GRCh38
rs776059852 Chr.11: 66283583-66283583 on Build GRCh38
rs919592566 Chr.11: 66283539-66283539 on Build GRCh38
rs369782697 Chr.11: 66283720-66283720 on Build GRCh38
rs943918733 Chr.11: 66283813-66283814 on Build GRCh38
rs150988199 Chr.11: 66283742-66283742 on Build GRCh38
rs4073583 Chr.11: 66283481-66283481 on Build GRCh38
rs375732442 Chr.11: 66283670-66283670 on Build GRCh38
rs34620898 Chr.11: 66283822-66283822 on Build GRCh38
rs368845133 Chr.11: 66283486-66283486 on Build GRCh38
rs776254344 Chr.11: 66283758-66283758 on Build GRCh38
rs199877182 Chr.11: 66283749-66283749 on Build GRCh38
rs779341306 Chr.11: 66283439-66283439 on Build GRCh38
rs373083100 Chr.11: 66283669-66283669 on Build GRCh38
rs749268222 Chr.11: 66283523-66283523 on Build GRCh38
rs559480992 Chr.11: 66283639-66283639 on Build GRCh38
rs1003137009 Chr.11: 66283746-66283746 on Build GRCh38
rs3016315 Chr.11: 66283477-66283477 on Build GRCh38
rs776948010 Chr.11: 66283579-66283579 on Build GRCh38
rs376182723 Chr.11: 66283619-66283619 on Build GRCh38
rs771761682 Chr.11: 66283533-66283533 on Build GRCh38
rs148871318 Chr.11: 66283552-66283552 on Build GRCh38

View Full Product Details