Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66422329-66422822 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NPAS4
Entrez Gene ID
266743
Gene Name
neuronal PAS domain protein 4
Gene Aliases
Le-PAS, NXF, PASD10, bHLHe79
UniGene
Hs.256036
Gene Chromosome Location
Chr.11: 66409158-66426707 on Build GRCh38
Transcripts
NM_001318804.1 NM_178864.3 XM_017017537.1 XM_017017539.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4980743 c.523G>A p.A175T Chr.11: 66422766-66422766 on Build GRCh38
6728556 c.449G>A p.R150H Chr.11: 66422692-66422692 on Build GRCh38
5046408 c.484G>A p.A162T Chr.11: 66422727-66422727 on Build GRCh38
6519268 c.360C>A p.Y120* Chr.11: 66422483-66422483 on Build GRCh38
276374 c.515G>A p.R172Q Chr.11: 66422758-66422758 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377576767 Chr.11: 66422483-66422483 on Build GRCh38
rs1028270009 Chr.11: 66422631-66422631 on Build GRCh38
rs781075841 Chr.11: 66422647-66422647 on Build GRCh38
rs748191401 Chr.11: 66422544-66422544 on Build GRCh38
rs113025811 Chr.11: 66422668-66422668 on Build GRCh38
rs765475757 Chr.11: 66422735-66422735 on Build GRCh38
rs377470432 Chr.11: 66422550-66422550 on Build GRCh38
rs996563429 Chr.11: 66422621-66422621 on Build GRCh38
rs772188887 Chr.11: 66422645-66422645 on Build GRCh38
rs779109369 Chr.11: 66422543-66422543 on Build GRCh38
rs759331205 Chr.11: 66422491-66422491 on Build GRCh38
rs373536098 Chr.11: 66422418-66422418 on Build GRCh38
rs370764299 Chr.11: 66422498-66422498 on Build GRCh38
rs761323840 Chr.11: 66422716-66422716 on Build GRCh38
rs770700942 Chr.11: 66422480-66422480 on Build GRCh38
rs756935260 Chr.11: 66422768-66422768 on Build GRCh38
rs375164036 Chr.11: 66422542-66422542 on Build GRCh38
rs749058982 Chr.11: 66422783-66422783 on Build GRCh38
rs756520070 Chr.11: 66422584-66422584 on Build GRCh38
rs780119454 Chr.11: 66422535-66422535 on Build GRCh38
rs756009378 Chr.11: 66422771-66422771 on Build GRCh38
rs749039442 Chr.11: 66422677-66422677 on Build GRCh38
rs758467501 Chr.11: 66422740-66422740 on Build GRCh38
rs763663656 Chr.11: 66422578-66422578 on Build GRCh38
rs749282742 Chr.11: 66422453-66422453 on Build GRCh38
rs547216404 Chr.11: 66422427-66422427 on Build GRCh38
rs750781616 Chr.11: 66422518-66422518 on Build GRCh38
rs780537675 Chr.11: 66422591-66422591 on Build GRCh38
rs768092555 Chr.11: 66422580-66422580 on Build GRCh38
rs200639446 Chr.11: 66422564-66422564 on Build GRCh38
rs1032809717 Chr.11: 66422708-66422708 on Build GRCh38
rs773853807 Chr.11: 66422715-66422715 on Build GRCh38
rs763725239 Chr.11: 66422507-66422507 on Build GRCh38
rs752956327 Chr.11: 66422739-66422739 on Build GRCh38
rs1054702408 Chr.11: 66422592-66422592 on Build GRCh38
rs757778610 Chr.11: 66422512-66422512 on Build GRCh38
rs149601016 Chr.11: 66422778-66422778 on Build GRCh38
rs868357121 Chr.11: 66422763-66422763 on Build GRCh38
rs185983665 Chr.11: 66422611-66422611 on Build GRCh38
rs781764783 Chr.11: 66422517-66422517 on Build GRCh38
rs916919931 Chr.11: 66422374-66422374 on Build GRCh38
rs751994670 Chr.11: 66422509-66422509 on Build GRCh38
rs762336996 Chr.11: 66422504-66422504 on Build GRCh38
rs1034278956 Chr.11: 66422386-66422386 on Build GRCh38
rs762722098 Chr.11: 66422570-66422570 on Build GRCh38
rs779008827 Chr.11: 66422634-66422634 on Build GRCh38
rs776571081 Chr.11: 66422560-66422560 on Build GRCh38
rs775294523 Chr.11: 66422676-66422676 on Build GRCh38
rs748186845 Chr.11: 66422641-66422641 on Build GRCh38
rs747102903 Chr.11: 66422756-66422756 on Build GRCh38
rs767179952 Chr.11: 66422702-66422702 on Build GRCh38
rs755330363 Chr.11: 66422733-66422733 on Build GRCh38
rs781726485 Chr.11: 66422406-66422406 on Build GRCh38
rs745871427 Chr.11: 66422649-66422649 on Build GRCh38
rs772995120 Chr.11: 66422476-66422476 on Build GRCh38
rs373552270 Chr.11: 66422425-66422425 on Build GRCh38
rs146312650 Chr.11: 66422720-66422720 on Build GRCh38
rs1026900541 Chr.11: 66422712-66422712 on Build GRCh38
rs780026330 Chr.11: 66422451-66422451 on Build GRCh38
rs777886600 Chr.11: 66422754-66422754 on Build GRCh38
rs755172181 Chr.11: 66422630-66422630 on Build GRCh38
rs749312787 Chr.11: 66422765-66422765 on Build GRCh38
rs79072452 Chr.11: 66422683-66422683 on Build GRCh38
rs756573371 Chr.11: 66422531-66422531 on Build GRCh38
rs760734772 Chr.11: 66422478-66422478 on Build GRCh38
rs140363432 Chr.11: 66422356-66422356 on Build GRCh38
rs766810347 Chr.11: 66422719-66422719 on Build GRCh38
rs893335216 Chr.11: 66422601-66422601 on Build GRCh38
rs949741660 Chr.11: 66422424-66422424 on Build GRCh38
rs753994979 Chr.11: 66422602-66422602 on Build GRCh38
rs768832716 Chr.11: 66422457-66422457 on Build GRCh38
rs958848493 Chr.11: 66422412-66422412 on Build GRCh38
rs768311502 Chr.11: 66422679-66422679 on Build GRCh38
rs569596792 Chr.11: 66422801-66422801 on Build GRCh38
rs749567499 Chr.11: 66422541-66422541 on Build GRCh38
rs368745643 Chr.11: 66422583-66422583 on Build GRCh38
rs142212942 Chr.11: 66422484-66422484 on Build GRCh38
rs773893309 Chr.11: 66422687-66422687 on Build GRCh38
rs139242618 Chr.11: 66422760-66422760 on Build GRCh38
rs267603125 Chr.11: 66422757-66422757 on Build GRCh38
rs774999808 Chr.11: 66422567-66422567 on Build GRCh38
rs374607165 Chr.11: 66422579-66422579 on Build GRCh38
rs772160928 Chr.11: 66422442-66422442 on Build GRCh38
rs764820082 Chr.11: 66422495-66422495 on Build GRCh38
rs370362797 Chr.11: 66422565-66422565 on Build GRCh38
rs775890164 Chr.11: 66422449-66422449 on Build GRCh38
rs761388893 Chr.11: 66422701-66422701 on Build GRCh38
rs757556367 Chr.11: 66422404-66422404 on Build GRCh38
rs929792476 Chr.11: 66422519-66422519 on Build GRCh38
rs768389587 Chr.11: 66422794-66422794 on Build GRCh38

View Full Product Details