Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66475513-66476010 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101928069
Entrez Gene ID
101928069
Gene Name
uncharacterized LOC101928069
Gene Aliases
-
UniGene
Hs.523816
Gene Chromosome Location
Chr.11: 66473490-66480249 on Build GRCh38
Transcripts
NR_120586.1 NM_001098510.1 NM_001243135.1 NM_001243136.1 NM_145065.2 XM_011544884.2 XM_017017465.1

Gene Symbol
PELI3
Entrez Gene ID
246330
Gene Name
pellino E3 ubiquitin protein ligase family member 3
Gene Aliases
-
UniGene
Hs.656875
Gene Chromosome Location
Chr.11: 66466327-66477337 on Build GRCh38
Transcripts
NR_120586.1 NM_001098510.1 NM_001243135.1 NM_001243136.1 NM_145065.2 XM_011544884.2 XM_017017465.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4406629 c.860T>A p.V287E Chr.11: 66475617-66475617 on Build GRCh38
1356216 c.1162C>T p.R388C Chr.11: 66475919-66475919 on Build GRCh38
5638612 c.1163G>A p.R388H Chr.11: 66475920-66475920 on Build GRCh38
1605077 c.1156C>A p.L386I Chr.11: 66475913-66475913 on Build GRCh38
4461864 c.1223C>T p.P408L Chr.11: 66475980-66475980 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1050099789 Chr.11: 66475678-66475678 on Build GRCh38
rs770067313 Chr.11: 66475645-66475645 on Build GRCh38
rs200001809 Chr.11: 66475590-66475590 on Build GRCh38
rs770127451 Chr.11: 66475724-66475724 on Build GRCh38
rs148240903 Chr.11: 66475960-66475960 on Build GRCh38
rs369045921 Chr.11: 66475740-66475740 on Build GRCh38
rs373269274 Chr.11: 66475880-66475880 on Build GRCh38
rs761939299 Chr.11: 66475762-66475762 on Build GRCh38
rs999987385 Chr.11: 66475735-66475735 on Build GRCh38
rs759475468 Chr.11: 66475846-66475846 on Build GRCh38
rs547475462 Chr.11: 66475600-66475600 on Build GRCh38
rs35712240 Chr.11: 66475747-66475747 on Build GRCh38
rs747470208 Chr.11: 66475584-66475584 on Build GRCh38
rs759580566 Chr.11: 66475534-66475534 on Build GRCh38
rs752334697 Chr.11: 66475674-66475674 on Build GRCh38
rs749650603 Chr.11: 66475890-66475890 on Build GRCh38
rs771538584 Chr.11: 66475719-66475719 on Build GRCh38
rs763100095 Chr.11: 66475614-66475614 on Build GRCh38
rs201923086 Chr.11: 66475799-66475799 on Build GRCh38
rs35467035 Chr.11: 66475930-66475931 on Build GRCh38
rs889666190 Chr.11: 66475688-66475688 on Build GRCh38
rs772082880 Chr.11: 66475843-66475843 on Build GRCh38
rs774879559 Chr.11: 66475887-66475887 on Build GRCh38
rs759404238 Chr.11: 66475790-66475790 on Build GRCh38
rs549816627 Chr.11: 66475675-66475675 on Build GRCh38
rs746212635 Chr.11: 66475722-66475722 on Build GRCh38
rs777614581 Chr.11: 66475805-66475805 on Build GRCh38
rs767017110 Chr.11: 66475878-66475878 on Build GRCh38
rs369405704 Chr.11: 66475800-66475800 on Build GRCh38
rs141386780 Chr.11: 66475975-66475975 on Build GRCh38
rs756626915 Chr.11: 66475886-66475886 on Build GRCh38
rs756980318 Chr.11: 66475877-66475877 on Build GRCh38
rs763921158 Chr.11: 66475936-66475936 on Build GRCh38
rs34989499 Chr.11: 66475616-66475616 on Build GRCh38
rs775846105 Chr.11: 66475611-66475611 on Build GRCh38
rs970277344 Chr.11: 66475884-66475884 on Build GRCh38
rs1020242772 Chr.11: 66475865-66475865 on Build GRCh38
rs923331282 Chr.11: 66475907-66475907 on Build GRCh38
rs138396296 Chr.11: 66475810-66475810 on Build GRCh38
rs778577702 Chr.11: 66475667-66475667 on Build GRCh38
rs930642996 Chr.11: 66475714-66475714 on Build GRCh38
rs952241632 Chr.11: 66475850-66475850 on Build GRCh38
rs781778065 Chr.11: 66475721-66475721 on Build GRCh38
rs558065068 Chr.11: 66475902-66475902 on Build GRCh38
rs747608891 Chr.11: 66475548-66475548 on Build GRCh38
rs141117623 Chr.11: 66475615-66475615 on Build GRCh38
rs766302098 Chr.11: 66475765-66475765 on Build GRCh38
rs765030587 Chr.11: 66475794-66475794 on Build GRCh38
rs375249910 Chr.11: 66475546-66475546 on Build GRCh38
rs775352193 Chr.11: 66475925-66475925 on Build GRCh38
rs745920534 Chr.11: 66475967-66475971 on Build GRCh38
rs772859688 Chr.11: 66475578-66475578 on Build GRCh38
rs202043323 Chr.11: 66475952-66475952 on Build GRCh38
rs372697971 Chr.11: 66475756-66475756 on Build GRCh38
rs763830890 Chr.11: 66475864-66475864 on Build GRCh38
rs774321848 Chr.11: 66475949-66475949 on Build GRCh38
rs754870704 Chr.11: 66475663-66475663 on Build GRCh38
rs145732233 Chr.11: 66475677-66475677 on Build GRCh38
rs765127805 Chr.11: 66475849-66475849 on Build GRCh38
rs762029235 Chr.11: 66475839-66475839 on Build GRCh38
rs765918697 Chr.11: 66475989-66475989 on Build GRCh38
rs774622996 Chr.11: 66475834-66475834 on Build GRCh38
rs368146253 Chr.11: 66475587-66475587 on Build GRCh38
rs892849062 Chr.11: 66475838-66475838 on Build GRCh38
rs759990043 Chr.11: 66475606-66475606 on Build GRCh38
rs770237444 Chr.11: 66475811-66475811 on Build GRCh38
rs751188795 Chr.11: 66475873-66475873 on Build GRCh38
rs767128060 Chr.11: 66475963-66475963 on Build GRCh38
rs202171966 Chr.11: 66475657-66475657 on Build GRCh38
rs1003180821 Chr.11: 66475893-66475893 on Build GRCh38
rs749668015 Chr.11: 66475823-66475823 on Build GRCh38
rs371559639 Chr.11: 66475589-66475589 on Build GRCh38
rs116062588 Chr.11: 66475903-66475903 on Build GRCh38
rs756892130 Chr.11: 66475807-66475807 on Build GRCh38
rs143139613 Chr.11: 66475927-66475927 on Build GRCh38
rs756018771 Chr.11: 66475627-66475627 on Build GRCh38
rs768910123 Chr.11: 66475744-66475744 on Build GRCh38
rs768852872 Chr.11: 66475832-66475832 on Build GRCh38
rs773185677 Chr.11: 66475844-66475844 on Build GRCh38
rs747106170 Chr.11: 66475918-66475918 on Build GRCh38
rs769753649 Chr.11: 66475919-66475919 on Build GRCh38
rs755270762 Chr.11: 66475891-66475891 on Build GRCh38
rs1040394953 Chr.11: 66475660-66475660 on Build GRCh38
rs780439573 Chr.11: 66475815-66475815 on Build GRCh38
rs771542241 Chr.11: 66475564-66475564 on Build GRCh38
rs147025276 Chr.11: 66475783-66475783 on Build GRCh38
rs962144383 Chr.11: 66475894-66475894 on Build GRCh38
rs141532058 Chr.11: 66475741-66475741 on Build GRCh38
rs200550810 Chr.11: 66475987-66475987 on Build GRCh38
rs200937664 Chr.11: 66475640-66475640 on Build GRCh38
rs749940073 Chr.11: 66475964-66475964 on Build GRCh38
rs773310999 Chr.11: 66475909-66475909 on Build GRCh38
rs752514383 Chr.11: 66475855-66475855 on Build GRCh38
rs746556803 Chr.11: 66475683-66475683 on Build GRCh38
rs751096914 Chr.11: 66475806-66475806 on Build GRCh38
rs779264293 Chr.11: 66475901-66475901 on Build GRCh38
rs762895508 Chr.11: 66475861-66475861 on Build GRCh38

View Full Product Details