Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66605622-66606127 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCS
Entrez Gene ID
9973
Gene Name
copper chaperone for superoxide dismutase
Gene Aliases
-
UniGene
Hs.502917
Gene Chromosome Location
Chr.11: 66593162-66606019 on Build GRCh38
Transcripts
NM_005125.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4999140 c.743G>A p.G248D Chr.11: 66605773-66605773 on Build GRCh38
4801272 c.709T>G p.F237V Chr.11: 66605739-66605739 on Build GRCh38
6623245 c.786G>T p.K262N Chr.11: 66605816-66605816 on Build GRCh38
3703625 c.819C>G p.H273Q Chr.11: 66605849-66605849 on Build GRCh38
6623243 c.699C>T p.S233S Chr.11: 66605729-66605729 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs992639743 Chr.11: 66605748-66605748 on Build GRCh38
rs776100556 Chr.11: 66605700-66605700 on Build GRCh38
rs137970813 Chr.11: 66605813-66605813 on Build GRCh38
rs1033983953 Chr.11: 66605932-66605932 on Build GRCh38
rs750467305 Chr.11: 66605656-66605656 on Build GRCh38
rs956673986 Chr.11: 66605913-66605913 on Build GRCh38
rs755646301 Chr.11: 66605660-66605661 on Build GRCh38
rs747225308 Chr.11: 66605922-66605922 on Build GRCh38
rs770480928 Chr.11: 66605676-66605676 on Build GRCh38
rs927017652 Chr.11: 66605651-66605651 on Build GRCh38
rs920764685 Chr.11: 66605686-66605686 on Build GRCh38
rs920101011 Chr.11: 66605773-66605773 on Build GRCh38
rs373751466 Chr.11: 66605800-66605800 on Build GRCh38
rs143494418 Chr.11: 66605805-66605805 on Build GRCh38
rs755815575 Chr.11: 66605886-66605886 on Build GRCh38
rs765194748 Chr.11: 66605742-66605742 on Build GRCh38
rs1127141 Chr.11: 66605783-66605783 on Build GRCh38
rs779503034 Chr.11: 66605681-66605681 on Build GRCh38
rs771207391 Chr.11: 66606017-66606017 on Build GRCh38
rs776242640 Chr.11: 66605677-66605677 on Build GRCh38
rs150455423 Chr.11: 66605649-66605649 on Build GRCh38
rs1040413982 Chr.11: 66605759-66605759 on Build GRCh38
rs769362989 Chr.11: 66605794-66605794 on Build GRCh38
rs763913397 Chr.11: 66605862-66605862 on Build GRCh38
rs1047957280 Chr.11: 66605752-66605752 on Build GRCh38
rs756139724 Chr.11: 66605736-66605736 on Build GRCh38
rs138370847 Chr.11: 66605713-66605713 on Build GRCh38
rs147502080 Chr.11: 66605821-66605821 on Build GRCh38
rs758314066 Chr.11: 66605762-66605762 on Build GRCh38
rs745347041 Chr.11: 66605690-66605690 on Build GRCh38
rs745413014 Chr.11: 66605793-66605793 on Build GRCh38
rs920091201 Chr.11: 66606065-66606065 on Build GRCh38
rs149284164 Chr.11: 66605769-66605769 on Build GRCh38
rs996714499 Chr.11: 66605709-66605709 on Build GRCh38
rs746729172 Chr.11: 66605673-66605673 on Build GRCh38
rs763414106 Chr.11: 66605702-66605702 on Build GRCh38
rs528045705 Chr.11: 66606067-66606067 on Build GRCh38
rs958398936 Chr.11: 66605972-66605972 on Build GRCh38
rs1021893681 Chr.11: 66605715-66605715 on Build GRCh38
rs973789740 Chr.11: 66605996-66605996 on Build GRCh38
rs143215772 Chr.11: 66605818-66605818 on Build GRCh38
rs981265886 Chr.11: 66606061-66606061 on Build GRCh38
rs760753625 Chr.11: 66605832-66605832 on Build GRCh38
rs908148014 Chr.11: 66605754-66605754 on Build GRCh38
rs977341120 Chr.11: 66605722-66605722 on Build GRCh38
rs944659445 Chr.11: 66605758-66605758 on Build GRCh38
rs969464081 Chr.11: 66606002-66606002 on Build GRCh38
rs780901578 Chr.11: 66605874-66605874 on Build GRCh38
rs931515796 Chr.11: 66606077-66606077 on Build GRCh38
rs754984299 Chr.11: 66605662-66605662 on Build GRCh38
rs370702728 Chr.11: 66605670-66605670 on Build GRCh38
rs750022471 Chr.11: 66605656-66605656 on Build GRCh38
rs558948617 Chr.11: 66605998-66605998 on Build GRCh38
rs767806191 Chr.11: 66605820-66605820 on Build GRCh38
rs1021421444 Chr.11: 66605839-66605839 on Build GRCh38
rs750568458 Chr.11: 66605731-66605731 on Build GRCh38
rs201822594 Chr.11: 66605871-66605871 on Build GRCh38
rs752568011 Chr.11: 66605746-66605746 on Build GRCh38
rs1127145 Chr.11: 66605834-66605834 on Build GRCh38
rs923937645 Chr.11: 66605744-66605744 on Build GRCh38
rs748697782 Chr.11: 66605838-66605838 on Build GRCh38
rs867758567 Chr.11: 66605838-66605838 on Build GRCh38
rs555054769 Chr.11: 66605667-66605667 on Build GRCh38
rs756979792 Chr.11: 66605771-66605771 on Build GRCh38
rs769281643 Chr.11: 66605808-66605808 on Build GRCh38
rs561125812 Chr.11: 66605766-66605766 on Build GRCh38
rs908092681 Chr.11: 66606011-66606011 on Build GRCh38
rs968590662 Chr.11: 66605716-66605716 on Build GRCh38
rs551837298 Chr.11: 66605865-66605865 on Build GRCh38
rs561632119 Chr.11: 66605669-66605669 on Build GRCh38
rs201533764 Chr.11: 66605841-66605841 on Build GRCh38
rs554910347 Chr.11: 66605833-66605833 on Build GRCh38
rs770444909 Chr.11: 66605695-66605695 on Build GRCh38
rs201606373 Chr.11: 66605768-66605768 on Build GRCh38
rs745458704 Chr.11: 66605875-66605875 on Build GRCh38
rs73505427 Chr.11: 66605672-66605672 on Build GRCh38
rs886764512 Chr.11: 66605807-66605807 on Build GRCh38
rs971734484 Chr.11: 66605890-66605890 on Build GRCh38
rs968581206 Chr.11: 66605880-66605880 on Build GRCh38
rs767604410 Chr.11: 66605729-66605729 on Build GRCh38
rs779777761 Chr.11: 66605893-66605893 on Build GRCh38
rs774566620 Chr.11: 66605724-66605724 on Build GRCh38
rs773363948 Chr.11: 66605895-66605895 on Build GRCh38
rs952231144 Chr.11: 66606002-66606003 on Build GRCh38
rs10061 Chr.11: 66605730-66605730 on Build GRCh38

View Full Product Details