Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67049039-67049562 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYT12
Entrez Gene ID
91683
Gene Name
synaptotagmin 12
Gene Aliases
SYT11, sytXII
UniGene
Hs.287636
Gene Chromosome Location
Chr.11: 67006778-67050863 on Build GRCh38
Transcripts
NM_001177880.1 NM_001318773.1 NM_001318775.1 NM_177963.3 XM_006718737.3 XM_011545346.2 XM_011545347.1 XM_017018547.1 XM_017018548.1

SNP Detail

SNP ID Chromosome Location
rs915126558 Chr.11: 67049115-67049115 on Build GRCh38
rs374834388 Chr.11: 67049458-67049458 on Build GRCh38
rs58201175 Chr.11: 67049286-67049286 on Build GRCh38
rs748562038 Chr.11: 67049421-67049421 on Build GRCh38
rs780355388 Chr.11: 67049288-67049288 on Build GRCh38
rs964518339 Chr.11: 67049338-67049338 on Build GRCh38
rs915967369 Chr.11: 67049422-67049422 on Build GRCh38
rs987177237 Chr.11: 67049083-67049083 on Build GRCh38
rs749830295 Chr.11: 67049289-67049289 on Build GRCh38
rs899173456 Chr.11: 67049254-67049254 on Build GRCh38
rs1049884888 Chr.11: 67049487-67049487 on Build GRCh38
rs530856839 Chr.11: 67049373-67049373 on Build GRCh38
rs932322095 Chr.11: 67049471-67049471 on Build GRCh38
rs983984725 Chr.11: 67049130-67049130 on Build GRCh38
rs575082917 Chr.11: 67049353-67049353 on Build GRCh38
rs922229787 Chr.11: 67049463-67049463 on Build GRCh38
rs900683354 Chr.11: 67049522-67049522 on Build GRCh38
rs998905585 Chr.11: 67049306-67049306 on Build GRCh38
rs890752291 Chr.11: 67049478-67049478 on Build GRCh38
rs148734389 Chr.11: 67049298-67049298 on Build GRCh38
rs927778579 Chr.11: 67049164-67049164 on Build GRCh38
rs756801748 Chr.11: 67049183-67049183 on Build GRCh38
rs889392400 Chr.11: 67049279-67049279 on Build GRCh38
rs1010977214 Chr.11: 67049346-67049346 on Build GRCh38
rs547244531 Chr.11: 67049347-67049347 on Build GRCh38
rs960086661 Chr.11: 67049311-67049311 on Build GRCh38
rs542756969 Chr.11: 67049154-67049154 on Build GRCh38
rs182582338 Chr.11: 67049442-67049442 on Build GRCh38
rs559912747 Chr.11: 67049389-67049389 on Build GRCh38
rs908274438 Chr.11: 67049137-67049137 on Build GRCh38
rs1035616870 Chr.11: 67049310-67049310 on Build GRCh38
rs1035689745 Chr.11: 67049068-67049068 on Build GRCh38
rs1037735969 Chr.11: 67049219-67049219 on Build GRCh38
rs927894525 Chr.11: 67049395-67049395 on Build GRCh38
rs77325998 Chr.11: 67049096-67049096 on Build GRCh38
rs993739633 Chr.11: 67049284-67049284 on Build GRCh38
rs758583902 Chr.11: 67049336-67049336 on Build GRCh38
rs939710040 Chr.11: 67049382-67049382 on Build GRCh38
rs187232030 Chr.11: 67049357-67049357 on Build GRCh38
rs954874479 Chr.11: 67049080-67049080 on Build GRCh38
rs964726460 Chr.11: 67049100-67049100 on Build GRCh38
rs939791854 Chr.11: 67049147-67049147 on Build GRCh38
rs1021785966 Chr.11: 67049293-67049293 on Build GRCh38
rs542437234 Chr.11: 67049488-67049488 on Build GRCh38
rs751016204 Chr.11: 67049177-67049177 on Build GRCh38
rs916035255 Chr.11: 67049188-67049188 on Build GRCh38
rs900841277 Chr.11: 67049170-67049170 on Build GRCh38
rs569838030 Chr.11: 67049264-67049264 on Build GRCh38
rs11827163 Chr.11: 67049489-67049489 on Build GRCh38
rs753020959 Chr.11: 67049182-67049182 on Build GRCh38
rs1017365459 Chr.11: 67049328-67049328 on Build GRCh38
rs772418510 Chr.11: 67049446-67049446 on Build GRCh38
rs1050713587 Chr.11: 67049204-67049204 on Build GRCh38
rs948710068 Chr.11: 67049216-67049216 on Build GRCh38
rs575652691 Chr.11: 67049106-67049106 on Build GRCh38
rs144238866 Chr.11: 67049095-67049095 on Build GRCh38
rs369774574 Chr.11: 67049509-67049509 on Build GRCh38
rs1047998415 Chr.11: 67049476-67049476 on Build GRCh38
rs931176299 Chr.11: 67049117-67049117 on Build GRCh38
rs908381422 Chr.11: 67049370-67049370 on Build GRCh38
rs939191151 Chr.11: 67049173-67049173 on Build GRCh38
rs779398008 Chr.11: 67049377-67049377 on Build GRCh38
rs890592324 Chr.11: 67049297-67049297 on Build GRCh38
rs866374516 Chr.11: 67049402-67049402 on Build GRCh38
rs945343760 Chr.11: 67049116-67049116 on Build GRCh38
rs192855665 Chr.11: 67049436-67049436 on Build GRCh38
rs976514264 Chr.11: 67049412-67049412 on Build GRCh38

View Full Product Details