Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67217651-67218149 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDM2A
Entrez Gene ID
22992
Gene Name
lysine demethylase 2A
Gene Aliases
CXXC8, FBL11, FBL7, FBXL11, JHDM1A, LILINA
UniGene
Hs.124147
Gene Chromosome Location
Chr.11: 67119210-67258082 on Build GRCh38
Transcripts
NM_012308.2 NR_027473.1 XM_006718480.3 XM_011544860.2 XM_011544861.1 XM_017017391.1 XM_017017392.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6692231 c.706C>T p.P236S Chr.11: 67217749-67217749 on Build GRCh38
931014 c.713C>T p.A238V Chr.11: 67217756-67217756 on Build GRCh38
3810194 c.828C>T p.F276F Chr.11: 67217871-67217871 on Build GRCh38
4508820 c.788C>G p.S263* Chr.11: 67217831-67217831 on Build GRCh38
6334031 c.794G>T p.C265F Chr.11: 67217837-67217837 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758160239 Chr.11: 67217787-67217787 on Build GRCh38
rs776008942 Chr.11: 67217702-67217702 on Build GRCh38
rs763341854 Chr.11: 67217706-67217706 on Build GRCh38
rs776370457 Chr.11: 67217714-67217714 on Build GRCh38
rs1009903493 Chr.11: 67217871-67217871 on Build GRCh38
rs922626500 Chr.11: 67218002-67218002 on Build GRCh38
rs549409682 Chr.11: 67218016-67218016 on Build GRCh38
rs763277369 Chr.11: 67217895-67217895 on Build GRCh38
rs951917285 Chr.11: 67217939-67217939 on Build GRCh38
rs140900090 Chr.11: 67218080-67218080 on Build GRCh38
rs998959473 Chr.11: 67217989-67217989 on Build GRCh38
rs753835308 Chr.11: 67217754-67217754 on Build GRCh38
rs750931088 Chr.11: 67218123-67218123 on Build GRCh38
rs766336172 Chr.11: 67217919-67217919 on Build GRCh38
rs767630504 Chr.11: 67217748-67217748 on Build GRCh38
rs531323377 Chr.11: 67217981-67217981 on Build GRCh38
rs889380009 Chr.11: 67217853-67217853 on Build GRCh38
rs774505188 Chr.11: 67217920-67217920 on Build GRCh38
rs772684146 Chr.11: 67217937-67217937 on Build GRCh38
rs764878559 Chr.11: 67217942-67217942 on Build GRCh38
rs769035071 Chr.11: 67217903-67217903 on Build GRCh38
rs1050002427 Chr.11: 67217676-67217676 on Build GRCh38
rs370448526 Chr.11: 67217829-67217829 on Build GRCh38
rs906863273 Chr.11: 67217913-67217913 on Build GRCh38
rs939865190 Chr.11: 67218114-67218114 on Build GRCh38
rs754925482 Chr.11: 67217756-67217756 on Build GRCh38
rs201135588 Chr.11: 67217708-67217708 on Build GRCh38
rs376276572 Chr.11: 67217793-67217793 on Build GRCh38
rs760667134 Chr.11: 67217905-67217906 on Build GRCh38
rs777336448 Chr.11: 67217788-67217788 on Build GRCh38
rs770340496 Chr.11: 67217685-67217685 on Build GRCh38
rs776098545 Chr.11: 67217888-67217888 on Build GRCh38
rs11227739 Chr.11: 67217923-67217923 on Build GRCh38
rs145472106 Chr.11: 67217963-67217963 on Build GRCh38
rs760708885 Chr.11: 67217928-67217928 on Build GRCh38
rs529668973 Chr.11: 67218085-67218085 on Build GRCh38
rs751785296 Chr.11: 67217712-67217712 on Build GRCh38
rs960369943 Chr.11: 67218011-67218011 on Build GRCh38
rs984078064 Chr.11: 67218094-67218094 on Build GRCh38
rs773288530 Chr.11: 67217924-67217924 on Build GRCh38
rs952683651 Chr.11: 67218091-67218091 on Build GRCh38
rs559989505 Chr.11: 67217929-67217929 on Build GRCh38
rs777314291 Chr.11: 67217682-67217682 on Build GRCh38
rs192963868 Chr.11: 67217680-67217680 on Build GRCh38
rs780835256 Chr.11: 67217844-67217844 on Build GRCh38
rs745316900 Chr.11: 67217855-67217855 on Build GRCh38
rs1001888352 Chr.11: 67217916-67217916 on Build GRCh38
rs986139558 Chr.11: 67218047-67218047 on Build GRCh38
rs936153497 Chr.11: 67217713-67217713 on Build GRCh38
rs762082758 Chr.11: 67217921-67217921 on Build GRCh38
rs770426802 Chr.11: 67217880-67217880 on Build GRCh38
rs151003407 Chr.11: 67218060-67218060 on Build GRCh38
rs12420763 Chr.11: 67217944-67217944 on Build GRCh38
rs752523087 Chr.11: 67217775-67217775 on Build GRCh38
rs908441873 Chr.11: 67218096-67218096 on Build GRCh38
rs868305626 Chr.11: 67217772-67217772 on Build GRCh38
rs547027121 Chr.11: 67217918-67217919 on Build GRCh38
rs972839270 Chr.11: 67217970-67217970 on Build GRCh38
rs778758911 Chr.11: 67217770-67217770 on Build GRCh38

View Full Product Details