Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67219342-67219890 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDM2A
Entrez Gene ID
22992
Gene Name
lysine demethylase 2A
Gene Aliases
CXXC8, FBL11, FBL7, FBXL11, JHDM1A, LILINA
UniGene
Hs.124147
Gene Chromosome Location
Chr.11: 67119210-67258082 on Build GRCh38
Transcripts
NM_012308.2 NR_027473.1 XM_006718480.3 XM_011544860.2 XM_011544861.1 XM_017017391.1 XM_017017392.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6863239 c.905G>C p.S302T Chr.11: 67219351-67219351 on Build GRCh38
6194351 c.928A>T p.K310* Chr.11: 67219374-67219374 on Build GRCh38
7283553 c.905G>A p.S302N Chr.11: 67219351-67219351 on Build GRCh38
4928810 c.938A>G p.N313S Chr.11: 67219384-67219384 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs960464628 Chr.11: 67219641-67219641 on Build GRCh38
rs185592869 Chr.11: 67219790-67219790 on Build GRCh38
rs958919133 Chr.11: 67219470-67219470 on Build GRCh38
rs201504546 Chr.11: 67219572-67219573 on Build GRCh38
rs969454678 Chr.11: 67219700-67219700 on Build GRCh38
rs897222322 Chr.11: 67219450-67219450 on Build GRCh38
rs575493371 Chr.11: 67219750-67219750 on Build GRCh38
rs987553044 Chr.11: 67219850-67219850 on Build GRCh38
rs201869182 Chr.11: 67219439-67219439 on Build GRCh38
rs1016406829 Chr.11: 67219523-67219523 on Build GRCh38
rs111732073 Chr.11: 67219827-67219827 on Build GRCh38
rs758577130 Chr.11: 67219760-67219760 on Build GRCh38
rs889827349 Chr.11: 67219863-67219863 on Build GRCh38
rs1053550598 Chr.11: 67219661-67219661 on Build GRCh38
rs372991424 Chr.11: 67219819-67219819 on Build GRCh38
rs764089486 Chr.11: 67219418-67219418 on Build GRCh38
rs980391873 Chr.11: 67219733-67219733 on Build GRCh38
rs918684210 Chr.11: 67219763-67219763 on Build GRCh38
rs868448219 Chr.11: 67219397-67219397 on Build GRCh38
rs767298797 Chr.11: 67219453-67219453 on Build GRCh38
rs1030721785 Chr.11: 67219629-67219629 on Build GRCh38
rs947506394 Chr.11: 67219693-67219693 on Build GRCh38
rs34898241 Chr.11: 67219686-67219687 on Build GRCh38
rs999302297 Chr.11: 67219572-67219572 on Build GRCh38
rs954458489 Chr.11: 67219843-67219843 on Build GRCh38
rs1042180232 Chr.11: 67219697-67219697 on Build GRCh38
rs182800538 Chr.11: 67219559-67219559 on Build GRCh38
rs763880528 Chr.11: 67219431-67219431 on Build GRCh38
rs993285994 Chr.11: 67219836-67219836 on Build GRCh38
rs766922545 Chr.11: 67219429-67219429 on Build GRCh38
rs1051079525 Chr.11: 67219481-67219481 on Build GRCh38
rs971800270 Chr.11: 67219546-67219546 on Build GRCh38
rs910475375 Chr.11: 67219617-67219617 on Build GRCh38
rs987134063 Chr.11: 67219603-67219603 on Build GRCh38
rs1016475642 Chr.11: 67219808-67219808 on Build GRCh38
rs910119821 Chr.11: 67219741-67219741 on Build GRCh38
rs557276041 Chr.11: 67219696-67219696 on Build GRCh38
rs903206313 Chr.11: 67219737-67219737 on Build GRCh38
rs111981707 Chr.11: 67219560-67219560 on Build GRCh38
rs748397968 Chr.11: 67219388-67219388 on Build GRCh38
rs200744093 Chr.11: 67219558-67219558 on Build GRCh38
rs915071665 Chr.11: 67219681-67219681 on Build GRCh38
rs774795323 Chr.11: 67219384-67219384 on Build GRCh38
rs1013172102 Chr.11: 67219762-67219762 on Build GRCh38
rs760790573 Chr.11: 67219424-67219424 on Build GRCh38
rs1026502948 Chr.11: 67219842-67219842 on Build GRCh38
rs561318984 Chr.11: 67219821-67219821 on Build GRCh38
rs534913690 Chr.11: 67219417-67219417 on Build GRCh38
rs941536771 Chr.11: 67219744-67219744 on Build GRCh38
rs115657829 Chr.11: 67219488-67219488 on Build GRCh38
rs963164304 Chr.11: 67219544-67219544 on Build GRCh38
rs761426200 Chr.11: 67219413-67219413 on Build GRCh38
rs578068501 Chr.11: 67219607-67219607 on Build GRCh38
rs762931066 Chr.11: 67219430-67219430 on Build GRCh38
rs768959966 Chr.11: 67219379-67219379 on Build GRCh38
rs539222913 Chr.11: 67219692-67219692 on Build GRCh38
rs1051542234 Chr.11: 67219852-67219852 on Build GRCh38
rs894738139 Chr.11: 67219761-67219761 on Build GRCh38
rs942700587 Chr.11: 67219864-67219864 on Build GRCh38
rs1036334301 Chr.11: 67219454-67219454 on Build GRCh38
rs941510489 Chr.11: 67219442-67219442 on Build GRCh38
rs954509434 Chr.11: 67219573-67219573 on Build GRCh38
rs943258057 Chr.11: 67219619-67219619 on Build GRCh38
rs757077539 Chr.11: 67219448-67219448 on Build GRCh38

View Full Product Details