Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 2797218-2797714 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XG
Entrez Gene ID
7499
Gene Name
Xg blood group
Gene Aliases
PBDX
UniGene
Hs.179675
Gene Chromosome Location
Chr.X: 2751108-2816500 on Build GRCh38
Transcripts
NM_001141919.1 NM_001141920.1 NM_175569.2 XM_005274587.3 XM_011545575.2 XM_017029787.1

SNP Detail

SNP ID Chromosome Location
rs774829003 Chr.X: 2797321-2797321 on Build GRCh38
rs367838820 Chr.X: 2797363-2797363 on Build GRCh38
rs370578832 Chr.X: 2797262-2797262 on Build GRCh38
rs768016709 Chr.X: 2797326-2797326 on Build GRCh38
rs138618142 Chr.X: 2797360-2797360 on Build GRCh38
rs372046947 Chr.X: 2797400-2797400 on Build GRCh38
rs769613383 Chr.X: 2797379-2797379 on Build GRCh38
rs141727718 Chr.X: 2797320-2797320 on Build GRCh38
rs746959859 Chr.X: 2797311-2797311 on Build GRCh38
rs977881148 Chr.X: 2797267-2797267 on Build GRCh38
rs773234690 Chr.X: 2797314-2797314 on Build GRCh38
rs779826523 Chr.X: 2797383-2797383 on Build GRCh38
rs757216217 Chr.X: 2797298-2797301 on Build GRCh38
rs150145174 Chr.X: 2797347-2797347 on Build GRCh38
rs755459001 Chr.X: 2797587-2797588 on Build GRCh38
rs754109442 Chr.X: 2797535-2797535 on Build GRCh38
rs778654776 Chr.X: 2797263-2797263 on Build GRCh38
rs1031285996 Chr.X: 2797287-2797287 on Build GRCh38
rs775353213 Chr.X: 2797308-2797308 on Build GRCh38
rs751037951 Chr.X: 2797608-2797608 on Build GRCh38
rs1047260126 Chr.X: 2797566-2797566 on Build GRCh38
rs969388580 Chr.X: 2797359-2797359 on Build GRCh38
rs370105441 Chr.X: 2797370-2797370 on Build GRCh38
rs771170430 Chr.X: 2797404-2797404 on Build GRCh38
rs756902043 Chr.X: 2797356-2797356 on Build GRCh38
rs753335679 Chr.X: 2797355-2797355 on Build GRCh38
rs763645598 Chr.X: 2797354-2797354 on Build GRCh38
rs368746982 Chr.X: 2797391-2797391 on Build GRCh38
rs760367775 Chr.X: 2797464-2797464 on Build GRCh38
rs767077270 Chr.X: 2797426-2797426 on Build GRCh38
rs769843327 Chr.X: 2797312-2797312 on Build GRCh38
rs765597274 Chr.X: 2797586-2797586 on Build GRCh38
rs780420114 Chr.X: 2797317-2797317 on Build GRCh38
rs192224164 Chr.X: 2797621-2797621 on Build GRCh38
rs771882283 Chr.X: 2797301-2797301 on Build GRCh38
rs311185 Chr.X: 2797283-2797283 on Build GRCh38
rs768563808 Chr.X: 2797573-2797573 on Build GRCh38
rs767436327 Chr.X: 2797364-2797365 on Build GRCh38
rs756584746 Chr.X: 2797348-2797348 on Build GRCh38
rs930248305 Chr.X: 2797548-2797548 on Build GRCh38
rs764487535 Chr.X: 2797514-2797514 on Build GRCh38
rs187425598 Chr.X: 2797422-2797422 on Build GRCh38
rs4484858 Chr.X: 2797384-2797384 on Build GRCh38
rs1025464733 Chr.X: 2797378-2797378 on Build GRCh38
rs745822343 Chr.X: 2797374-2797374 on Build GRCh38
rs200183426 Chr.X: 2797393-2797393 on Build GRCh38
rs757655392 Chr.X: 2797585-2797585 on Build GRCh38
rs780415616 Chr.X: 2797588-2797589 on Build GRCh38
rs761649585 Chr.X: 2797329-2797329 on Build GRCh38
rs775109605 Chr.X: 2797444-2797444 on Build GRCh38
rs759941200 Chr.X: 2797324-2797324 on Build GRCh38
rs758539551 Chr.X: 2797619-2797620 on Build GRCh38
rs944438982 Chr.X: 2797587-2797590 on Build GRCh38
rs1040541113 Chr.X: 2797588-2797588 on Build GRCh38

View Full Product Details