Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 108508591-108509120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXPH5
Entrez Gene ID
23086
Gene Name
exophilin 5
Gene Aliases
SLAC2-B, SLAC2B
UniGene
Hs.28540
Gene Chromosome Location
Chr.11: 108505431-108607514 on Build GRCh38
Transcripts
NM_001144763.1 NM_001144764.1 NM_001144765.1 NM_001308019.1 NM_015065.2 XM_011542696.2 XM_011542698.2 XM_011542700.2 XM_017017397.1 XM_017017398.1 XM_017017399.1 XM_017017400.1 XM_017017401.1 XM_017017402.1 XM_017017403.1 XM_017017404.1

SNP Detail

SNP ID Chromosome Location
rs942274806 Chr.11: 108508620-108508620 on Build GRCh38
rs748186021 Chr.11: 108508858-108508858 on Build GRCh38
rs184423642 Chr.11: 108508789-108508789 on Build GRCh38
rs986084679 Chr.11: 108509029-108509029 on Build GRCh38
rs946077841 Chr.11: 108508724-108508724 on Build GRCh38
rs751024320 Chr.11: 108508747-108508747 on Build GRCh38
rs547431656 Chr.11: 108508930-108508930 on Build GRCh38
rs1041730463 Chr.11: 108508733-108508733 on Build GRCh38
rs143561553 Chr.11: 108508722-108508722 on Build GRCh38
rs769740668 Chr.11: 108509069-108509069 on Build GRCh38
rs577345784 Chr.11: 108508938-108508939 on Build GRCh38
rs937455381 Chr.11: 108508660-108508660 on Build GRCh38
rs926020013 Chr.11: 108508937-108508937 on Build GRCh38
rs773463462 Chr.11: 108509054-108509054 on Build GRCh38
rs889696483 Chr.11: 108508719-108508719 on Build GRCh38
rs527944172 Chr.11: 108508743-108508743 on Build GRCh38
rs888226762 Chr.11: 108508849-108508849 on Build GRCh38
rs1034058125 Chr.11: 108508868-108508868 on Build GRCh38
rs983552850 Chr.11: 108508903-108508903 on Build GRCh38
rs942575260 Chr.11: 108509046-108509046 on Build GRCh38
rs749409645 Chr.11: 108508782-108508782 on Build GRCh38
rs531876244 Chr.11: 108508819-108508819 on Build GRCh38
rs1050408857 Chr.11: 108508678-108508678 on Build GRCh38
rs199727056 Chr.11: 108508949-108508949 on Build GRCh38
rs12366211 Chr.11: 108508922-108508922 on Build GRCh38
rs192525049 Chr.11: 108508742-108508742 on Build GRCh38
rs3832764 Chr.11: 108508939-108508940 on Build GRCh38
rs900304885 Chr.11: 108508790-108508790 on Build GRCh38
rs574632921 Chr.11: 108509053-108509055 on Build GRCh38
rs895818168 Chr.11: 108508723-108508723 on Build GRCh38
rs533803473 Chr.11: 108509013-108509013 on Build GRCh38
rs935988149 Chr.11: 108508673-108508673 on Build GRCh38
rs777359412 Chr.11: 108508652-108508652 on Build GRCh38
rs148050853 Chr.11: 108508882-108508882 on Build GRCh38
rs77741219 Chr.11: 108508649-108508649 on Build GRCh38
rs550226009 Chr.11: 108508905-108508905 on Build GRCh38
rs1024004023 Chr.11: 108508773-108508773 on Build GRCh38
rs76922339 Chr.11: 108508924-108508924 on Build GRCh38
rs188811283 Chr.11: 108508630-108508630 on Build GRCh38
rs942664992 Chr.11: 108508720-108508720 on Build GRCh38
rs910559462 Chr.11: 108509042-108509042 on Build GRCh38
rs566431089 Chr.11: 108508936-108508936 on Build GRCh38
rs1014510751 Chr.11: 108508902-108508902 on Build GRCh38
rs757296397 Chr.11: 108508865-108508865 on Build GRCh38
rs1030536054 Chr.11: 108508862-108508862 on Build GRCh38
rs543263641 Chr.11: 108508783-108508783 on Build GRCh38
rs973596747 Chr.11: 108508943-108508944 on Build GRCh38
rs995943804 Chr.11: 108508807-108508807 on Build GRCh38
rs397811390 Chr.11: 108508948-108508949 on Build GRCh38
rs901011427 Chr.11: 108508754-108508754 on Build GRCh38

View Full Product Details