Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 124987814-124988213 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC15
Entrez Gene ID
80071
Gene Name
coiled-coil domain containing 15
Gene Aliases
-
UniGene
Hs.287555
Gene Chromosome Location
Chr.11: 124954121-125041489 on Build GRCh38
Transcripts
NM_025004.2 XM_017018346.1 XM_017018347.1 XM_017018348.1 XM_017018349.1 XM_017018350.1 XM_017018351.1 XM_017018352.1 XM_017018353.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5885374 c.1713C>T p.P571P Chr.11: 124987939-124987939 on Build GRCh38
5838101 c.1748A>G p.Q583R Chr.11: 124987974-124987974 on Build GRCh38
390467 c.1762A>G p.R588G Chr.11: 124987988-124987988 on Build GRCh38
6991087 c.1783A>G p.K595E Chr.11: 124988009-124988009 on Build GRCh38
5838099 c.1706T>A p.V569D Chr.11: 124987932-124987932 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377372741 Chr.11: 124987845-124987845 on Build GRCh38
rs772719036 Chr.11: 124987957-124987957 on Build GRCh38
rs985026637 Chr.11: 124987925-124987925 on Build GRCh38
rs779117710 Chr.11: 124987933-124987933 on Build GRCh38
rs757252705 Chr.11: 124987932-124987932 on Build GRCh38
rs571571707 Chr.11: 124987855-124987855 on Build GRCh38
rs201316378 Chr.11: 124988127-124988127 on Build GRCh38
rs941104828 Chr.11: 124988001-124988001 on Build GRCh38
rs747337981 Chr.11: 124987941-124987941 on Build GRCh38
rs553577640 Chr.11: 124987963-124987963 on Build GRCh38
rs754851588 Chr.11: 124987907-124987907 on Build GRCh38
rs370955511 Chr.11: 124988029-124988029 on Build GRCh38
rs958907238 Chr.11: 124987883-124987883 on Build GRCh38
rs751330216 Chr.11: 124987896-124987896 on Build GRCh38
rs762722016 Chr.11: 124987879-124987879 on Build GRCh38
rs1032982544 Chr.11: 124987862-124987862 on Build GRCh38
rs199512085 Chr.11: 124988049-124988049 on Build GRCh38
rs750037111 Chr.11: 124987870-124987870 on Build GRCh38
rs757199874 Chr.11: 124988131-124988131 on Build GRCh38
rs370673006 Chr.11: 124987944-124987944 on Build GRCh38
rs776974998 Chr.11: 124987962-124987962 on Build GRCh38
rs200603729 Chr.11: 124988108-124988108 on Build GRCh38
rs868062031 Chr.11: 124987878-124987878 on Build GRCh38
rs755131889 Chr.11: 124988115-124988115 on Build GRCh38
rs958734083 Chr.11: 124988132-124988132 on Build GRCh38
rs756457704 Chr.11: 124988128-124988128 on Build GRCh38
rs933961508 Chr.11: 124987971-124987971 on Build GRCh38
rs61912661 Chr.11: 124987861-124987861 on Build GRCh38
rs763983988 Chr.11: 124987977-124987978 on Build GRCh38
rs748783104 Chr.11: 124988067-124988067 on Build GRCh38
rs373838890 Chr.11: 124988093-124988093 on Build GRCh38
rs762371908 Chr.11: 124987960-124987960 on Build GRCh38
rs59550486 Chr.11: 124988110-124988110 on Build GRCh38
rs749800937 Chr.11: 124988145-124988145 on Build GRCh38
rs753745832 Chr.11: 124988096-124988096 on Build GRCh38
rs758443949 Chr.11: 124988008-124988008 on Build GRCh38
rs770268494 Chr.11: 124988042-124988042 on Build GRCh38
rs376536200 Chr.11: 124987986-124987986 on Build GRCh38
rs191332524 Chr.11: 124987936-124987936 on Build GRCh38
rs758469275 Chr.11: 124988098-124988098 on Build GRCh38
rs781592429 Chr.11: 124988019-124988019 on Build GRCh38
rs777088244 Chr.11: 124987948-124987948 on Build GRCh38
rs748376438 Chr.11: 124988113-124988113 on Build GRCh38
rs772282016 Chr.11: 124987937-124987937 on Build GRCh38
rs775368917 Chr.11: 124988085-124988085 on Build GRCh38
rs373843708 Chr.11: 124988179-124988179 on Build GRCh38
rs774064065 Chr.11: 124988158-124988158 on Build GRCh38
rs910499636 Chr.11: 124987929-124987929 on Build GRCh38
rs747268569 Chr.11: 124988012-124988012 on Build GRCh38
rs770396707 Chr.11: 124988074-124988074 on Build GRCh38
rs752732058 Chr.11: 124987931-124987931 on Build GRCh38
rs753513654 Chr.11: 124988127-124988128 on Build GRCh38
rs760393725 Chr.11: 124988088-124988088 on Build GRCh38
rs1013156143 Chr.11: 124988157-124988157 on Build GRCh38
rs745385252 Chr.11: 124988160-124988160 on Build GRCh38
rs763353843 Chr.11: 124987880-124987880 on Build GRCh38
rs774116998 Chr.11: 124988078-124988078 on Build GRCh38
rs780205336 Chr.11: 124988009-124988009 on Build GRCh38
rs769170576 Chr.11: 124987953-124987953 on Build GRCh38
rs750453198 Chr.11: 124988006-124988006 on Build GRCh38
rs113750570 Chr.11: 124988037-124988037 on Build GRCh38
rs759329629 Chr.11: 124988079-124988079 on Build GRCh38
rs770561371 Chr.11: 124988152-124988152 on Build GRCh38
rs368706917 Chr.11: 124987949-124987949 on Build GRCh38
rs878916683 Chr.11: 124988000-124988000 on Build GRCh38
rs868117168 Chr.11: 124987877-124987877 on Build GRCh38
rs766015052 Chr.11: 124987961-124987961 on Build GRCh38
rs375126608 Chr.11: 124987964-124987964 on Build GRCh38
rs971060036 Chr.11: 124988161-124988161 on Build GRCh38
rs201340905 Chr.11: 124987881-124987881 on Build GRCh38
rs751001834 Chr.11: 124987969-124987969 on Build GRCh38
rs372783869 Chr.11: 124987928-124987928 on Build GRCh38
rs199841007 Chr.11: 124987974-124987974 on Build GRCh38
rs778320476 Chr.11: 124988144-124988144 on Build GRCh38
rs780264212 Chr.11: 124987938-124987938 on Build GRCh38
rs539276818 Chr.11: 124988080-124988080 on Build GRCh38
rs551396707 Chr.11: 124987853-124987853 on Build GRCh38
rs866316131 Chr.11: 124988043-124988043 on Build GRCh38
rs781396051 Chr.11: 124988117-124988117 on Build GRCh38
rs202196230 Chr.11: 124987893-124987893 on Build GRCh38
rs200807898 Chr.11: 124987939-124987939 on Build GRCh38
rs769089581 Chr.11: 124987851-124987851 on Build GRCh38
rs755327970 Chr.11: 124988015-124988015 on Build GRCh38

View Full Product Details