Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 19713351-19713899 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR11H2
Entrez Gene ID
79334
Gene Name
olfactory receptor family 11 subfamily H member 2
Gene Aliases
C14orf15, OR11H13, OR11H2P, OR11H8P
UniGene
Hs.554706
Gene Chromosome Location
Chr.14: 19712904-19714332 on Build GRCh38
Transcripts
NM_001197287.1

SNP Detail

SNP ID Chromosome Location
rs755680112 Chr.14: 19713825-19713825 on Build GRCh38
rs4983150 Chr.14: 19713450-19713450 on Build GRCh38
rs577202377 Chr.14: 19713440-19713440 on Build GRCh38
rs1056674540 Chr.14: 19713608-19713608 on Build GRCh38
rs755110313 Chr.14: 19713410-19713410 on Build GRCh38
rs201517204 Chr.14: 19713648-19713648 on Build GRCh38
rs779291084 Chr.14: 19713549-19713549 on Build GRCh38
rs762845839 Chr.14: 19713435-19713437 on Build GRCh38
rs780893137 Chr.14: 19713711-19713711 on Build GRCh38
rs529001888 Chr.14: 19713872-19713872 on Build GRCh38
rs541582055 Chr.14: 19713639-19713639 on Build GRCh38
rs768124684 Chr.14: 19713485-19713485 on Build GRCh38
rs761851829 Chr.14: 19713428-19713428 on Build GRCh38
rs760193968 Chr.14: 19713397-19713397 on Build GRCh38
rs765753700 Chr.14: 19713681-19713681 on Build GRCh38
rs534247969 Chr.14: 19713394-19713394 on Build GRCh38
rs775110144 Chr.14: 19713874-19713874 on Build GRCh38
rs774438253 Chr.14: 19713740-19713740 on Build GRCh38
rs754314626 Chr.14: 19713667-19713667 on Build GRCh38
rs750726972 Chr.14: 19713690-19713690 on Build GRCh38
rs761894230 Chr.14: 19713780-19713780 on Build GRCh38
rs766956795 Chr.14: 19713731-19713732 on Build GRCh38
rs568121769 Chr.14: 19713809-19713809 on Build GRCh38
rs750183452 Chr.14: 19713435-19713435 on Build GRCh38
rs558878120 Chr.14: 19713425-19713425 on Build GRCh38
rs749604213 Chr.14: 19713457-19713457 on Build GRCh38
rs773027834 Chr.14: 19713490-19713490 on Build GRCh38
rs752299270 Chr.14: 19713829-19713829 on Build GRCh38
rs531258908 Chr.14: 19713759-19713759 on Build GRCh38
rs765117771 Chr.14: 19713433-19713433 on Build GRCh38
rs771869242 Chr.14: 19713873-19713873 on Build GRCh38
rs556285351 Chr.14: 19713600-19713600 on Build GRCh38
rs766699718 Chr.14: 19713812-19713812 on Build GRCh38
rs780444553 Chr.14: 19713408-19713408 on Build GRCh38
rs751806869 Chr.14: 19713493-19713493 on Build GRCh38
rs777588169 Chr.14: 19713841-19713841 on Build GRCh38
rs752766103 Chr.14: 19713517-19713517 on Build GRCh38
rs776165251 Chr.14: 19713487-19713487 on Build GRCh38
rs559917075 Chr.14: 19713662-19713662 on Build GRCh38
rs775760370 Chr.14: 19713763-19713763 on Build GRCh38
rs769713930 Chr.14: 19713489-19713489 on Build GRCh38
rs766331692 Chr.14: 19713492-19713492 on Build GRCh38
rs765130362 Chr.14: 19713404-19713404 on Build GRCh38
rs757596273 Chr.14: 19713538-19713538 on Build GRCh38
rs753882709 Chr.14: 19713402-19713402 on Build GRCh38
rs763954638 Chr.14: 19713400-19713400 on Build GRCh38
rs973791423 Chr.14: 19713482-19713482 on Build GRCh38
rs748391730 Chr.14: 19713381-19713381 on Build GRCh38
rs761249623 Chr.14: 19713488-19713488 on Build GRCh38
rs773589321 Chr.14: 19713416-19713416 on Build GRCh38
rs779668861 Chr.14: 19713468-19713468 on Build GRCh38
rs778160617 Chr.14: 19713454-19713454 on Build GRCh38
rs759418864 Chr.14: 19713729-19713729 on Build GRCh38
rs760796289 Chr.14: 19713657-19713657 on Build GRCh38
rs778985082 Chr.14: 19713854-19713854 on Build GRCh38
rs774231144 Chr.14: 19713599-19713599 on Build GRCh38
rs748743874 Chr.14: 19713842-19713842 on Build GRCh38
rs767716308 Chr.14: 19713651-19713651 on Build GRCh38
rs564089287 Chr.14: 19713747-19713747 on Build GRCh38
rs758606563 Chr.14: 19713407-19713407 on Build GRCh38
rs962470992 Chr.14: 19713474-19713474 on Build GRCh38
rs766273475 Chr.14: 19713515-19713515 on Build GRCh38
rs759875736 Chr.14: 19713499-19713499 on Build GRCh38
rs527552593 Chr.14: 19713700-19713700 on Build GRCh38
rs778342917 Chr.14: 19713526-19713526 on Build GRCh38
rs754400787 Chr.14: 19713533-19713533 on Build GRCh38
rs199618256 Chr.14: 19713726-19713726 on Build GRCh38
rs760175410 Chr.14: 19713424-19713424 on Build GRCh38
rs771657519 Chr.14: 19713420-19713420 on Build GRCh38
rs758279658 Chr.14: 19713437-19713437 on Build GRCh38
rs755624245 Chr.14: 19713714-19713714 on Build GRCh38
rs773045103 Chr.14: 19713394-19713394 on Build GRCh38
rs746263590 Chr.14: 19713563-19713563 on Build GRCh38
rs756598282 Chr.14: 19713453-19713453 on Build GRCh38
rs768247230 Chr.14: 19713599-19713599 on Build GRCh38
rs752359256 Chr.14: 19713817-19713817 on Build GRCh38
rs759069772 Chr.14: 19713599-19713600 on Build GRCh38
rs771353050 Chr.14: 19713460-19713460 on Build GRCh38
rs771995552 Chr.14: 19713679-19713679 on Build GRCh38
rs1830970 Chr.14: 19713480-19713480 on Build GRCh38
rs760848161 Chr.14: 19713764-19713764 on Build GRCh38
rs767657698 Chr.14: 19713507-19713507 on Build GRCh38
rs776410461 Chr.14: 19713427-19713427 on Build GRCh38
rs756157269 Chr.14: 19713520-19713520 on Build GRCh38
rs756988934 Chr.14: 19713403-19713403 on Build GRCh38
rs200825460 Chr.14: 19713603-19713603 on Build GRCh38
rs775131797 Chr.14: 19713422-19713422 on Build GRCh38
rs747808926 Chr.14: 19713712-19713712 on Build GRCh38
rs747628064 Chr.14: 19713606-19713606 on Build GRCh38
rs758694856 Chr.14: 19713697-19713697 on Build GRCh38
rs759421420 Chr.14: 19713649-19713649 on Build GRCh38
rs549840728 Chr.14: 19713772-19713772 on Build GRCh38
rs751893938 Chr.14: 19713442-19713442 on Build GRCh38
rs574283556 Chr.14: 19713616-19713616 on Build GRCh38
rs748742484 Chr.14: 19713411-19713411 on Build GRCh38
rs61979610 Chr.14: 19713771-19713771 on Build GRCh38
rs756898409 Chr.14: 19713852-19713852 on Build GRCh38
rs3000029 Chr.14: 19713375-19713375 on Build GRCh38
rs770258388 Chr.14: 19713385-19713385 on Build GRCh38
rs777463395 Chr.14: 19713716-19713716 on Build GRCh38
rs773184011 Chr.14: 19713619-19713619 on Build GRCh38
rs544670557 Chr.14: 19713443-19713443 on Build GRCh38
rs765663664 Chr.14: 19713796-19713796 on Build GRCh38
rs759276984 Chr.14: 19713387-19713387 on Build GRCh38

View Full Product Details