Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 137908781-137909359 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PKD2L2
Entrez Gene ID
27039
Gene Name
polycystin 2 like 2, transient receptor potential cation channel
Gene Aliases
TRPP5
UniGene
Hs.716884
Gene Chromosome Location
Chr.5: 137889420-137942747 on Build GRCh38
Transcripts
NM_001258448.1 NM_001258449.1 NM_001300921.1 NM_014386.3 XM_011543318.2 XM_011543321.1 XM_017009343.1 XM_017009344.1 XM_017009345.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3850331 c.1290A>G p.G430G Chr.5: 137908908-137908908 on Build GRCh38
5537118 c.1288G>A p.G430R Chr.5: 137908906-137908906 on Build GRCh38
284045 c.1190C>A p.S397* Chr.5: 137908808-137908808 on Build GRCh38
4602126 c.1294C>A p.Q432K Chr.5: 137908912-137908912 on Build GRCh38
4602125 c.1294C>A p.Q432K Chr.5: 137908912-137908912 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373512810 Chr.5: 137909176-137909176 on Build GRCh38
rs754376683 Chr.5: 137908824-137908824 on Build GRCh38
rs1046163115 Chr.5: 137909136-137909136 on Build GRCh38
rs542708338 Chr.5: 137908972-137908972 on Build GRCh38
rs975557198 Chr.5: 137909114-137909114 on Build GRCh38
rs934220617 Chr.5: 137909167-137909167 on Build GRCh38
rs957979637 Chr.5: 137908840-137908840 on Build GRCh38
rs767088585 Chr.5: 137908957-137908957 on Build GRCh38
rs768431299 Chr.5: 137908939-137908939 on Build GRCh38
rs199670627 Chr.5: 137908823-137908823 on Build GRCh38
rs377579603 Chr.5: 137909245-137909245 on Build GRCh38
rs893598202 Chr.5: 137909190-137909190 on Build GRCh38
rs757796311 Chr.5: 137909094-137909094 on Build GRCh38
rs910163419 Chr.5: 137909063-137909063 on Build GRCh38
rs184162232 Chr.5: 137909085-137909085 on Build GRCh38
rs1030659149 Chr.5: 137909051-137909051 on Build GRCh38
rs375219991 Chr.5: 137908935-137908935 on Build GRCh38
rs201043281 Chr.5: 137908915-137908915 on Build GRCh38
rs532188798 Chr.5: 137908952-137908952 on Build GRCh38
rs767303843 Chr.5: 137908992-137908993 on Build GRCh38
rs964053354 Chr.5: 137909297-137909297 on Build GRCh38
rs1011841932 Chr.5: 137909102-137909102 on Build GRCh38
rs922652791 Chr.5: 137908960-137908960 on Build GRCh38
rs1030827744 Chr.5: 137909189-137909189 on Build GRCh38
rs758806204 Chr.5: 137908876-137908876 on Build GRCh38
rs560141358 Chr.5: 137908928-137908928 on Build GRCh38
rs894539563 Chr.5: 137909195-137909195 on Build GRCh38
rs765723305 Chr.5: 137908836-137908836 on Build GRCh38
rs534044832 Chr.5: 137909253-137909253 on Build GRCh38
rs1039905023 Chr.5: 137908819-137908819 on Build GRCh38
rs906409278 Chr.5: 137909300-137909300 on Build GRCh38
rs970599708 Chr.5: 137909008-137909008 on Build GRCh38
rs922791273 Chr.5: 137909156-137909156 on Build GRCh38
rs377634546 Chr.5: 137908999-137908999 on Build GRCh38
rs908795007 Chr.5: 137909178-137909178 on Build GRCh38
rs753251472 Chr.5: 137908998-137908998 on Build GRCh38
rs570073289 Chr.5: 137909326-137909326 on Build GRCh38
rs555425831 Chr.5: 137909281-137909281 on Build GRCh38
rs267600352 Chr.5: 137908821-137908821 on Build GRCh38
rs941323298 Chr.5: 137909186-137909186 on Build GRCh38
rs906276243 Chr.5: 137909140-137909140 on Build GRCh38
rs561097066 Chr.5: 137908976-137908976 on Build GRCh38
rs1010827313 Chr.5: 137909258-137909258 on Build GRCh38
rs114599444 Chr.5: 137909097-137909097 on Build GRCh38
rs774107485 Chr.5: 137908940-137908940 on Build GRCh38
rs1880458 Chr.5: 137908828-137908828 on Build GRCh38
rs773226695 Chr.5: 137908993-137908993 on Build GRCh38
rs761604235 Chr.5: 137908857-137908858 on Build GRCh38
rs757372723 Chr.5: 137908912-137908912 on Build GRCh38
rs188232477 Chr.5: 137909242-137909242 on Build GRCh38
rs999807039 Chr.5: 137909166-137909166 on Build GRCh38
rs756925881 Chr.5: 137909213-137909213 on Build GRCh38
rs989164947 Chr.5: 137909069-137909074 on Build GRCh38
rs747187186 Chr.5: 137908905-137908905 on Build GRCh38
rs745847797 Chr.5: 137908923-137908923 on Build GRCh38
rs753165363 Chr.5: 137908843-137908843 on Build GRCh38
rs772550534 Chr.5: 137908964-137908964 on Build GRCh38
rs201962100 Chr.5: 137908883-137908883 on Build GRCh38
rs551365115 Chr.5: 137909134-137909134 on Build GRCh38
rs894814473 Chr.5: 137909099-137909099 on Build GRCh38
rs948829677 Chr.5: 137909205-137909205 on Build GRCh38
rs1022430939 Chr.5: 137909080-137909083 on Build GRCh38
rs1038819927 Chr.5: 137908959-137908959 on Build GRCh38
rs549854202 Chr.5: 137909022-137909022 on Build GRCh38

View Full Product Details