Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 160199016-160199601 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FABP6
Entrez Gene ID
2172
Gene Name
fatty acid binding protein 6
Gene Aliases
I-15P, I-BABP, I-BALB, I-BAP, ILBP, ILBP3, ILLBP
UniGene
Hs.519719
Gene Chromosome Location
Chr.5: 160187367-160238722 on Build GRCh38
Transcripts
NM_001040442.1 NM_001130958.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5141494 c.22A>G p.M8V Chr.5: 160199128-160199128 on Build GRCh38
6436344 c.44T>A p.L15Q Chr.5: 160199150-160199150 on Build GRCh38
3613420 c.49C>A p.Q17K Chr.5: 160199155-160199155 on Build GRCh38
7148431 c.31G>A p.E11K Chr.5: 160199137-160199137 on Build GRCh38
6657894 c.41C>T p.A14V Chr.5: 160199147-160199147 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs949957425 Chr.5: 160199442-160199442 on Build GRCh38
rs774753160 Chr.5: 160199173-160199173 on Build GRCh38
rs750540506 Chr.5: 160199085-160199085 on Build GRCh38
rs777059808 Chr.5: 160199088-160199095 on Build GRCh38
rs990450815 Chr.5: 160199419-160199419 on Build GRCh38
rs765185372 Chr.5: 160199079-160199079 on Build GRCh38
rs767724967 Chr.5: 160199187-160199187 on Build GRCh38
rs971340029 Chr.5: 160199562-160199562 on Build GRCh38
rs983792986 Chr.5: 160199399-160199399 on Build GRCh38
rs751957245 Chr.5: 160199091-160199091 on Build GRCh38
rs755346751 Chr.5: 160199092-160199092 on Build GRCh38
rs753853647 Chr.5: 160199388-160199388 on Build GRCh38
rs932038839 Chr.5: 160199498-160199498 on Build GRCh38
rs766772386 Chr.5: 160199090-160199090 on Build GRCh38
rs902759466 Chr.5: 160199495-160199495 on Build GRCh38
rs550873727 Chr.5: 160199175-160199175 on Build GRCh38
rs974760839 Chr.5: 160199380-160199380 on Build GRCh38
rs1024660347 Chr.5: 160199571-160199571 on Build GRCh38
rs1027672238 Chr.5: 160199540-160199540 on Build GRCh38
rs138753107 Chr.5: 160199240-160199240 on Build GRCh38
rs938994849 Chr.5: 160199407-160199407 on Build GRCh38
rs771727828 Chr.5: 160199122-160199122 on Build GRCh38
rs546743603 Chr.5: 160199385-160199385 on Build GRCh38
rs989753996 Chr.5: 160199476-160199476 on Build GRCh38
rs1015294773 Chr.5: 160199560-160199560 on Build GRCh38
rs146032989 Chr.5: 160199121-160199121 on Build GRCh38
rs4388245 Chr.5: 160199509-160199509 on Build GRCh38
rs1054124789 Chr.5: 160199247-160199247 on Build GRCh38
rs760701869 Chr.5: 160199073-160199073 on Build GRCh38
rs894816227 Chr.5: 160199437-160199437 on Build GRCh38
rs995449897 Chr.5: 160199422-160199422 on Build GRCh38
rs775331841 Chr.5: 160199068-160199068 on Build GRCh38
rs890959629 Chr.5: 160199285-160199285 on Build GRCh38
rs531102670 Chr.5: 160199167-160199167 on Build GRCh38
rs983463100 Chr.5: 160199556-160199556 on Build GRCh38
rs773418482 Chr.5: 160199148-160199148 on Build GRCh38
rs746860569 Chr.5: 160199134-160199134 on Build GRCh38
rs772026716 Chr.5: 160199066-160199066 on Build GRCh38
rs945711296 Chr.5: 160199481-160199481 on Build GRCh38
rs1019075099 Chr.5: 160199508-160199508 on Build GRCh38
rs760036322 Chr.5: 160199117-160199118 on Build GRCh38
rs1012409225 Chr.5: 160199445-160199445 on Build GRCh38
rs575748642 Chr.5: 160199474-160199474 on Build GRCh38
rs768035620 Chr.5: 160199095-160199095 on Build GRCh38
rs55636938 Chr.5: 160199549-160199549 on Build GRCh38
rs759386302 Chr.5: 160199062-160199062 on Build GRCh38
rs763197384 Chr.5: 160199088-160199088 on Build GRCh38
rs1043062632 Chr.5: 160199487-160199487 on Build GRCh38
rs916300235 Chr.5: 160199439-160199439 on Build GRCh38
rs1019483380 Chr.5: 160199309-160199309 on Build GRCh38
rs756576198 Chr.5: 160199201-160199201 on Build GRCh38
rs111817900 Chr.5: 160199527-160199527 on Build GRCh38
rs775327708 Chr.5: 160199249-160199249 on Build GRCh38
rs952019368 Chr.5: 160199390-160199390 on Build GRCh38
rs964764268 Chr.5: 160199314-160199314 on Build GRCh38
rs774130643 Chr.5: 160199060-160199060 on Build GRCh38
rs34544641 Chr.5: 160199470-160199470 on Build GRCh38
rs1009520114 Chr.5: 160199503-160199503 on Build GRCh38
rs1011174407 Chr.5: 160199252-160199252 on Build GRCh38
rs907771497 Chr.5: 160199414-160199414 on Build GRCh38
rs753111139 Chr.5: 160199199-160199199 on Build GRCh38
rs764178238 Chr.5: 160199278-160199278 on Build GRCh38
rs566940906 Chr.5: 160199449-160199449 on Build GRCh38
rs1021180918 Chr.5: 160199260-160199260 on Build GRCh38
rs921887741 Chr.5: 160199489-160199489 on Build GRCh38
rs753104712 Chr.5: 160199103-160199103 on Build GRCh38
rs66840636 Chr.5: 160199237-160199237 on Build GRCh38
rs376905193 Chr.5: 160199459-160199459 on Build GRCh38
rs202217927 Chr.5: 160199132-160199132 on Build GRCh38
rs760561971 Chr.5: 160199261-160199261 on Build GRCh38
rs756666320 Chr.5: 160199105-160199105 on Build GRCh38
rs199868205 Chr.5: 160199131-160199131 on Build GRCh38
rs748865194 Chr.5: 160199112-160199112 on Build GRCh38
rs927072043 Chr.5: 160199456-160199456 on Build GRCh38
rs768640444 Chr.5: 160199146-160199146 on Build GRCh38
rs967302868 Chr.5: 160199270-160199270 on Build GRCh38
rs141290407 Chr.5: 160199473-160199473 on Build GRCh38
rs1009056595 Chr.5: 160199277-160199277 on Build GRCh38
rs987545370 Chr.5: 160199506-160199509 on Build GRCh38
rs770574469 Chr.5: 160199057-160199057 on Build GRCh38
rs368190475 Chr.5: 160199349-160199351 on Build GRCh38
rs369290281 Chr.5: 160199357-160199359 on Build GRCh38
rs774979117 Chr.5: 160199100-160199100 on Build GRCh38
rs896999637 Chr.5: 160199510-160199510 on Build GRCh38
rs1027033255 Chr.5: 160199425-160199425 on Build GRCh38
rs201223363 Chr.5: 160199120-160199120 on Build GRCh38
rs777082611 Chr.5: 160199106-160199106 on Build GRCh38
rs560214423 Chr.5: 160199042-160199042 on Build GRCh38

View Full Product Details