Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 175512973-175513550 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SFXN1
Entrez Gene ID
94081
Gene Name
sideroflexin 1
Gene Aliases
TCC
UniGene
Hs.369440
Gene Chromosome Location
Chr.5: 175478498-175529742 on Build GRCh38
Transcripts
NM_001322977.1 NM_001322978.1 NM_001322980.1 NM_001322981.1 NM_001322982.1 NM_001322983.1 NM_022754.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5538840 c.617C>T p.P206L Chr.5: 175513483-175513483 on Build GRCh38
4725391 c.661G>A p.A221T Chr.5: 175513527-175513527 on Build GRCh38
3919711 c.648G>A p.G216G Chr.5: 175513514-175513514 on Build GRCh38
6776472 c.618C>A p.P206P Chr.5: 175513484-175513484 on Build GRCh38
5656006 c.653C>T p.S218L Chr.5: 175513519-175513519 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375933160 Chr.5: 175513487-175513487 on Build GRCh38
rs544364580 Chr.5: 175513328-175513328 on Build GRCh38
rs1027735808 Chr.5: 175513118-175513118 on Build GRCh38
rs2644669 Chr.5: 175513233-175513233 on Build GRCh38
rs890142143 Chr.5: 175513309-175513309 on Build GRCh38
rs147084508 Chr.5: 175513509-175513509 on Build GRCh38
rs901963159 Chr.5: 175513508-175513508 on Build GRCh38
rs546491424 Chr.5: 175513321-175513324 on Build GRCh38
rs1013185550 Chr.5: 175513452-175513452 on Build GRCh38
rs926392925 Chr.5: 175513360-175513360 on Build GRCh38
rs897732703 Chr.5: 175513396-175513396 on Build GRCh38
rs554598453 Chr.5: 175513193-175513193 on Build GRCh38
rs71597334 Chr.5: 175513134-175513134 on Build GRCh38
rs552628184 Chr.5: 175513130-175513130 on Build GRCh38
rs993452303 Chr.5: 175513270-175513270 on Build GRCh38
rs1047096096 Chr.5: 175513009-175513009 on Build GRCh38
rs79095421 Chr.5: 175513323-175513326 on Build GRCh38
rs190858561 Chr.5: 175513027-175513027 on Build GRCh38
rs1050750773 Chr.5: 175513266-175513266 on Build GRCh38
rs753529247 Chr.5: 175513527-175513527 on Build GRCh38
rs548859151 Chr.5: 175513394-175513394 on Build GRCh38
rs763565479 Chr.5: 175513440-175513440 on Build GRCh38
rs533984393 Chr.5: 175513091-175513091 on Build GRCh38
rs776746253 Chr.5: 175513414-175513414 on Build GRCh38
rs910733741 Chr.5: 175513157-175513157 on Build GRCh38
rs944380639 Chr.5: 175513332-175513332 on Build GRCh38
rs746494221 Chr.5: 175513068-175513068 on Build GRCh38
rs866086193 Chr.5: 175513377-175513377 on Build GRCh38
rs575577589 Chr.5: 175513186-175513186 on Build GRCh38
rs1024483544 Chr.5: 175513457-175513457 on Build GRCh38
rs763427303 Chr.5: 175513520-175513520 on Build GRCh38
rs996490758 Chr.5: 175513058-175513058 on Build GRCh38
rs190209027 Chr.5: 175513434-175513434 on Build GRCh38
rs144516349 Chr.5: 175513526-175513526 on Build GRCh38
rs138497198 Chr.5: 175513512-175513512 on Build GRCh38
rs139000060 Chr.5: 175513191-175513191 on Build GRCh38
rs149270574 Chr.5: 175513522-175513522 on Build GRCh38
rs1017697587 Chr.5: 175513297-175513297 on Build GRCh38
rs750803136 Chr.5: 175513297-175513309 on Build GRCh38
rs897783340 Chr.5: 175513253-175513253 on Build GRCh38
rs902972571 Chr.5: 175513358-175513358 on Build GRCh38
rs567384168 Chr.5: 175513284-175513284 on Build GRCh38
rs748421606 Chr.5: 175513515-175513515 on Build GRCh38
rs905092726 Chr.5: 175513062-175513062 on Build GRCh38
rs902918846 Chr.5: 175513155-175513155 on Build GRCh38
rs759250389 Chr.5: 175513484-175513484 on Build GRCh38
rs757195521 Chr.5: 175513502-175513502 on Build GRCh38
rs1003390304 Chr.5: 175513066-175513066 on Build GRCh38
rs753720909 Chr.5: 175513497-175513497 on Build GRCh38
rs528079721 Chr.5: 175513351-175513351 on Build GRCh38
rs115315870 Chr.5: 175513177-175513177 on Build GRCh38
rs55902078 Chr.5: 175513048-175513048 on Build GRCh38
rs952261030 Chr.5: 175513129-175513129 on Build GRCh38
rs987807592 Chr.5: 175513319-175513319 on Build GRCh38
rs143175347 Chr.5: 175513507-175513507 on Build GRCh38
rs1056260968 Chr.5: 175513384-175513384 on Build GRCh38
rs893588501 Chr.5: 175513024-175513024 on Build GRCh38
rs386695265 Chr.5: 175513175-175513177 on Build GRCh38
rs539889045 Chr.5: 175513255-175513255 on Build GRCh38
rs149484695 Chr.5: 175513470-175513470 on Build GRCh38
rs367698228 Chr.5: 175513490-175513490 on Build GRCh38
rs956305952 Chr.5: 175513318-175513318 on Build GRCh38
rs544408946 Chr.5: 175513323-175513324 on Build GRCh38
rs961982749 Chr.5: 175513072-175513072 on Build GRCh38
rs369485721 Chr.5: 175513523-175513523 on Build GRCh38
rs182614076 Chr.5: 175513158-175513158 on Build GRCh38
rs748714826 Chr.5: 175513442-175513443 on Build GRCh38
rs187237840 Chr.5: 175513000-175513000 on Build GRCh38
rs868721329 Chr.5: 175513008-175513008 on Build GRCh38
rs993459197 Chr.5: 175513101-175513101 on Build GRCh38
rs55761424 Chr.5: 175513324-175513324 on Build GRCh38
rs552289806 Chr.5: 175513489-175513489 on Build GRCh38
rs890088439 Chr.5: 175513064-175513064 on Build GRCh38
rs920391358 Chr.5: 175513234-175513234 on Build GRCh38
rs924849617 Chr.5: 175513313-175513313 on Build GRCh38
rs976287316 Chr.5: 175513306-175513306 on Build GRCh38
rs777333892 Chr.5: 175513043-175513043 on Build GRCh38
rs769702628 Chr.5: 175513519-175513519 on Build GRCh38
rs566836682 Chr.5: 175513081-175513081 on Build GRCh38
rs777228417 Chr.5: 175513308-175513309 on Build GRCh38
rs907732811 Chr.5: 175513022-175513022 on Build GRCh38
rs781228529 Chr.5: 175513513-175513513 on Build GRCh38
rs1037921092 Chr.5: 175513026-175513026 on Build GRCh38
rs371212864 Chr.5: 175513454-175513454 on Build GRCh38
rs774922891 Chr.5: 175513478-175513478 on Build GRCh38
rs75624721 Chr.5: 175513320-175513320 on Build GRCh38
rs1045333547 Chr.5: 175513044-175513044 on Build GRCh38
rs1048989602 Chr.5: 175513441-175513441 on Build GRCh38
rs946414889 Chr.5: 175513336-175513336 on Build GRCh38
rs1008031794 Chr.5: 175513295-175513295 on Build GRCh38
rs924859664 Chr.5: 175513171-175513171 on Build GRCh38
rs187201182 Chr.5: 175513366-175513366 on Build GRCh38
rs777295891 Chr.5: 175513299-175513311 on Build GRCh38
rs1028098442 Chr.5: 175513279-175513279 on Build GRCh38
rs976298558 Chr.5: 175513161-175513161 on Build GRCh38
rs200390050 Chr.5: 175513485-175513485 on Build GRCh38
rs778679266 Chr.5: 175513505-175513505 on Build GRCh38
rs564954672 Chr.5: 175513277-175513277 on Build GRCh38
rs1044840464 Chr.5: 175513192-175513192 on Build GRCh38
rs373899701 Chr.5: 175513215-175513215 on Build GRCh38
rs769945554 Chr.5: 175513425-175513425 on Build GRCh38
rs116236863 Chr.5: 175513175-175513175 on Build GRCh38
rs748213702 Chr.5: 175513423-175513423 on Build GRCh38
rs765160945 Chr.5: 175513307-175513309 on Build GRCh38
rs966162443 Chr.5: 175513301-175513301 on Build GRCh38

View Full Product Details