Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 124950523-124950927 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX37
Entrez Gene ID
57647
Gene Name
DEAH-box helicase 37
Gene Aliases
DDX37
UniGene
Hs.107382
Gene Chromosome Location
Chr.12: 124946824-124989125 on Build GRCh38
Transcripts
NM_032656.3 XM_005253590.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3871074 c.2914C>T p.L972F Chr.12: 124950759-124950759 on Build GRCh38
5430523 c.2986G>A p.V996I Chr.12: 124950548-124950548 on Build GRCh38
6346337 c.2926C>A p.L976I Chr.12: 124950747-124950747 on Build GRCh38
936791 c.2971A>G p.M991V Chr.12: 124950702-124950702 on Build GRCh38
430675 c.2917T>A p.F973I Chr.12: 124950756-124950756 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756776212 Chr.12: 124950744-124950744 on Build GRCh38
rs772091220 Chr.12: 124950817-124950817 on Build GRCh38
rs910365763 Chr.12: 124950637-124950637 on Build GRCh38
rs779168447 Chr.12: 124950803-124950803 on Build GRCh38
rs755946339 Chr.12: 124950852-124950852 on Build GRCh38
rs770178274 Chr.12: 124950684-124950684 on Build GRCh38
rs184270416 Chr.12: 124950610-124950610 on Build GRCh38
rs745655247 Chr.12: 124950589-124950589 on Build GRCh38
rs372815146 Chr.12: 124950854-124950854 on Build GRCh38
rs61757600 Chr.12: 124950718-124950718 on Build GRCh38
rs761041095 Chr.12: 124950820-124950820 on Build GRCh38
rs749970499 Chr.12: 124950847-124950847 on Build GRCh38
rs768060240 Chr.12: 124950677-124950677 on Build GRCh38
rs199578145 Chr.12: 124950773-124950773 on Build GRCh38
rs111456585 Chr.12: 124950717-124950717 on Build GRCh38
rs201090732 Chr.12: 124950758-124950758 on Build GRCh38
rs565550244 Chr.12: 124950623-124950623 on Build GRCh38
rs748243390 Chr.12: 124950815-124950815 on Build GRCh38
rs372463222 Chr.12: 124950823-124950823 on Build GRCh38
rs773646108 Chr.12: 124950681-124950681 on Build GRCh38
rs751221066 Chr.12: 124950838-124950838 on Build GRCh38
rs910397015 Chr.12: 124950859-124950859 on Build GRCh38
rs770051885 Chr.12: 124950590-124950590 on Build GRCh38
rs752440046 Chr.12: 124950654-124950654 on Build GRCh38
rs757151866 Chr.12: 124950657-124950657 on Build GRCh38
rs778430577 Chr.12: 124950558-124950558 on Build GRCh38
rs114074306 Chr.12: 124950655-124950655 on Build GRCh38
rs750775063 Chr.12: 124950549-124950549 on Build GRCh38
rs370563951 Chr.12: 124950597-124950597 on Build GRCh38
rs762258581 Chr.12: 124950640-124950640 on Build GRCh38
rs775657558 Chr.12: 124950667-124950667 on Build GRCh38
rs375307340 Chr.12: 124950560-124950560 on Build GRCh38
rs752108250 Chr.12: 124950546-124950546 on Build GRCh38
rs761075434 Chr.12: 124950685-124950685 on Build GRCh38
rs181779649 Chr.12: 124950855-124950855 on Build GRCh38
rs115221248 Chr.12: 124950763-124950763 on Build GRCh38
rs760331969 Chr.12: 124950559-124950560 on Build GRCh38
rs375604344 Chr.12: 124950555-124950555 on Build GRCh38
rs750729526 Chr.12: 124950645-124950645 on Build GRCh38
rs768887553 Chr.12: 124950675-124950675 on Build GRCh38
rs984513157 Chr.12: 124950848-124950848 on Build GRCh38
rs189467805 Chr.12: 124950676-124950676 on Build GRCh38
rs775747831 Chr.12: 124950593-124950593 on Build GRCh38
rs964502047 Chr.12: 124950902-124950902 on Build GRCh38
rs151265243 Chr.12: 124950762-124950762 on Build GRCh38
rs960441547 Chr.12: 124950793-124950793 on Build GRCh38
rs892531538 Chr.12: 124950694-124950694 on Build GRCh38
rs367848710 Chr.12: 124950656-124950656 on Build GRCh38
rs536491190 Chr.12: 124950581-124950581 on Build GRCh38
rs372044227 Chr.12: 124950730-124950730 on Build GRCh38
rs773374631 Chr.12: 124950819-124950819 on Build GRCh38
rs10773125 Chr.12: 124950842-124950842 on Build GRCh38
rs932056979 Chr.12: 124950587-124950587 on Build GRCh38
rs575682659 Chr.12: 124950580-124950580 on Build GRCh38
rs930383419 Chr.12: 124950629-124950629 on Build GRCh38
rs775962605 Chr.12: 124950574-124950574 on Build GRCh38
rs768799595 Chr.12: 124950792-124950792 on Build GRCh38
rs140554912 Chr.12: 124950789-124950789 on Build GRCh38
rs763826752 Chr.12: 124950825-124950825 on Build GRCh38
rs142360847 Chr.12: 124950742-124950742 on Build GRCh38
rs763829040 Chr.12: 124950600-124950600 on Build GRCh38
rs766895090 Chr.12: 124950845-124950845 on Build GRCh38
rs776433771 Chr.12: 124950584-124950584 on Build GRCh38
rs781058615 Chr.12: 124950661-124950661 on Build GRCh38
rs148565171 Chr.12: 124950638-124950638 on Build GRCh38
rs890439440 Chr.12: 124950626-124950626 on Build GRCh38
rs745664037 Chr.12: 124950663-124950663 on Build GRCh38
rs749574178 Chr.12: 124950899-124950899 on Build GRCh38
rs555181449 Chr.12: 124950583-124950583 on Build GRCh38
rs369525506 Chr.12: 124950759-124950759 on Build GRCh38
rs1029211112 Chr.12: 124950904-124950904 on Build GRCh38
rs375762765 Chr.12: 124950779-124950779 on Build GRCh38
rs1018842597 Chr.12: 124950851-124950851 on Build GRCh38
rs376940167 Chr.12: 124950741-124950741 on Build GRCh38
rs966355190 Chr.12: 124950863-124950863 on Build GRCh38
rs974638316 Chr.12: 124950874-124950874 on Build GRCh38
rs774443764 Chr.12: 124950598-124950598 on Build GRCh38
rs375705373 Chr.12: 124950687-124950687 on Build GRCh38
rs746906254 Chr.12: 124950582-124950582 on Build GRCh38
rs370122962 Chr.12: 124950821-124950821 on Build GRCh38
rs976036579 Chr.12: 124950903-124950903 on Build GRCh38
rs544154799 Chr.12: 124950643-124950643 on Build GRCh38
rs757584327 Chr.12: 124950547-124950547 on Build GRCh38
rs879028523 Chr.12: 124950897-124950897 on Build GRCh38
rs35498490 Chr.12: 124950594-124950594 on Build GRCh38
rs756498981 Chr.12: 124950647-124950647 on Build GRCh38
rs377211297 Chr.12: 124950648-124950648 on Build GRCh38
rs781579759 Chr.12: 124950548-124950548 on Build GRCh38
rs754596894 Chr.12: 124950556-124950556 on Build GRCh38
rs749470961 Chr.12: 124950672-124950672 on Build GRCh38
rs865823686 Chr.12: 124950585-124950585 on Build GRCh38
rs777073142 Chr.12: 124950822-124950822 on Build GRCh38

View Full Product Details