Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 128982841-128983342 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLT1D1
Entrez Gene ID
144423
Gene Name
glycosyltransferase 1 domain containing 1
Gene Aliases
-
UniGene
Hs.655668
Gene Chromosome Location
Chr.12: 128853323-128984964 on Build GRCh38
Transcripts
NM_144669.2 NR_133646.1 XM_005253553.4 XM_011537956.2 XM_011537957.2 XM_017018856.1 XM_017018857.1 XM_017018858.1 XM_017018860.1 XM_017018861.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4687256 c.937G>A p.V313I Chr.12: 128982986-128982986 on Build GRCh38
7369576 c.1017G>T p.K339N Chr.12: 128983066-128983066 on Build GRCh38
468001 c.687A>G p.E229E Chr.12: 128982976-128982976 on Build GRCh38
459612 c.698T>A p.V233E Chr.12: 128982987-128982987 on Build GRCh38
5907604 c.785G>A p.G262E Chr.12: 128983074-128983074 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755191964 Chr.12: 128983049-128983049 on Build GRCh38
rs371240067 Chr.12: 128982912-128982912 on Build GRCh38
rs781124895 Chr.12: 128982953-128982953 on Build GRCh38
rs200864217 Chr.12: 128983115-128983115 on Build GRCh38
rs201184250 Chr.12: 128983078-128983078 on Build GRCh38
rs756338558 Chr.12: 128982881-128982881 on Build GRCh38
rs147418750 Chr.12: 128983103-128983103 on Build GRCh38
rs755831469 Chr.12: 128983041-128983041 on Build GRCh38
rs762349285 Chr.12: 128982926-128982926 on Build GRCh38
rs750155607 Chr.12: 128982956-128982956 on Build GRCh38
rs764757497 Chr.12: 128982907-128982907 on Build GRCh38
rs747593622 Chr.12: 128983077-128983077 on Build GRCh38
rs778402828 Chr.12: 128983064-128983064 on Build GRCh38
rs763635818 Chr.12: 128982937-128982937 on Build GRCh38
rs777114679 Chr.12: 128983081-128983081 on Build GRCh38
rs959036539 Chr.12: 128983130-128983130 on Build GRCh38
rs753991383 Chr.12: 128983046-128983046 on Build GRCh38
rs1026142050 Chr.12: 128983290-128983290 on Build GRCh38
rs866790089 Chr.12: 128983023-128983023 on Build GRCh38
rs780207577 Chr.12: 128982911-128982913 on Build GRCh38
rs779281722 Chr.12: 128983050-128983050 on Build GRCh38
rs751016784 Chr.12: 128983035-128983035 on Build GRCh38
rs773016021 Chr.12: 128983111-128983111 on Build GRCh38
rs535059991 Chr.12: 128983069-128983069 on Build GRCh38
rs767390154 Chr.12: 128983040-128983040 on Build GRCh38
rs766239540 Chr.12: 128983120-128983120 on Build GRCh38
rs947608945 Chr.12: 128983167-128983167 on Build GRCh38
rs140092627 Chr.12: 128983006-128983006 on Build GRCh38
rs1043276870 Chr.12: 128983174-128983174 on Build GRCh38
rs757245039 Chr.12: 128982952-128982952 on Build GRCh38
rs201839480 Chr.12: 128983296-128983296 on Build GRCh38
rs774975656 Chr.12: 128983015-128983015 on Build GRCh38
rs755956013 Chr.12: 128982966-128982966 on Build GRCh38
rs377377789 Chr.12: 128982986-128982986 on Build GRCh38
rs772920154 Chr.12: 128983276-128983276 on Build GRCh38
rs927621942 Chr.12: 128983129-128983129 on Build GRCh38
rs576547710 Chr.12: 128983270-128983270 on Build GRCh38
rs774026819 Chr.12: 128983097-128983097 on Build GRCh38
rs764639021 Chr.12: 128983226-128983226 on Build GRCh38
rs35993969 Chr.12: 128983162-128983162 on Build GRCh38
rs78799732 Chr.12: 128982985-128982985 on Build GRCh38
rs903292971 Chr.12: 128982962-128982962 on Build GRCh38
rs1007532097 Chr.12: 128983261-128983261 on Build GRCh38
rs947550706 Chr.12: 128982919-128982919 on Build GRCh38
rs1006248398 Chr.12: 128982977-128982977 on Build GRCh38
rs145605197 Chr.12: 128983007-128983007 on Build GRCh38
rs746235160 Chr.12: 128983087-128983087 on Build GRCh38
rs755238228 Chr.12: 128983034-128983037 on Build GRCh38
rs773653592 Chr.12: 128982890-128982890 on Build GRCh38
rs374628572 Chr.12: 128983025-128983025 on Build GRCh38
rs111512414 Chr.12: 128983139-128983139 on Build GRCh38
rs768235031 Chr.12: 128983094-128983094 on Build GRCh38
rs374640326 Chr.12: 128982957-128982957 on Build GRCh38
rs779962467 Chr.12: 128983042-128983042 on Build GRCh38
rs75920323 Chr.12: 128983219-128983219 on Build GRCh38
rs747250421 Chr.12: 128982918-128982919 on Build GRCh38
rs751496255 Chr.12: 128982944-128982944 on Build GRCh38
rs148480797 Chr.12: 128983176-128983176 on Build GRCh38
rs934772308 Chr.12: 128983204-128983204 on Build GRCh38
rs994704054 Chr.12: 128983278-128983278 on Build GRCh38
rs773886460 Chr.12: 128982999-128982999 on Build GRCh38
rs370400269 Chr.12: 128982995-128982995 on Build GRCh38
rs373877202 Chr.12: 128983136-128983136 on Build GRCh38
rs772301070 Chr.12: 128983061-128983061 on Build GRCh38
rs748249350 Chr.12: 128982994-128982994 on Build GRCh38
rs761287278 Chr.12: 128983102-128983102 on Build GRCh38
rs201627526 Chr.12: 128983032-128983032 on Build GRCh38
rs111578125 Chr.12: 128983062-128983062 on Build GRCh38
rs761105698 Chr.12: 128982893-128982893 on Build GRCh38
rs1021028404 Chr.12: 128983031-128983031 on Build GRCh38
rs748197949 Chr.12: 128983057-128983057 on Build GRCh38
rs1039383672 Chr.12: 128983269-128983269 on Build GRCh38
rs761320197 Chr.12: 128983039-128983039 on Build GRCh38

View Full Product Details