Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 131728201-131728682 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SFSWAP
Entrez Gene ID
6433
Gene Name
splicing factor SWAP homolog
Gene Aliases
SFRS8, SWAP
UniGene
Hs.308171
Gene Chromosome Location
Chr.12: 131711087-131799738 on Build GRCh38
Transcripts
NM_001261411.1 NM_004592.3 XM_011538653.1 XM_011538654.2 XM_011538655.2 XM_017019798.1 XM_017019799.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4444398 c.1073C>T p.T358M Chr.12: 131728420-131728420 on Build GRCh38
7279082 c.1021A>G p.T341A Chr.12: 131728368-131728368 on Build GRCh38
937077 c.1032C>T p.N344N Chr.12: 131728379-131728379 on Build GRCh38
4725368 c.1075G>T p.A359S Chr.12: 131728422-131728422 on Build GRCh38
3782707 c.1054C>T p.P352S Chr.12: 131728401-131728401 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779995526 Chr.12: 131728280-131728281 on Build GRCh38
rs368173659 Chr.12: 131728277-131728277 on Build GRCh38
rs542020329 Chr.12: 131728379-131728379 on Build GRCh38
rs755872590 Chr.12: 131728326-131728326 on Build GRCh38
rs768946019 Chr.12: 131728449-131728450 on Build GRCh38
rs930829103 Chr.12: 131728618-131728618 on Build GRCh38
rs371449895 Chr.12: 131728536-131728536 on Build GRCh38
rs140262217 Chr.12: 131728390-131728390 on Build GRCh38
rs752728726 Chr.12: 131728450-131728450 on Build GRCh38
rs529405050 Chr.12: 131728656-131728656 on Build GRCh38
rs576562646 Chr.12: 131728376-131728376 on Build GRCh38
rs766214247 Chr.12: 131728421-131728421 on Build GRCh38
rs564129125 Chr.12: 131728423-131728423 on Build GRCh38
rs996659585 Chr.12: 131728426-131728426 on Build GRCh38
rs748257985 Chr.12: 131728369-131728369 on Build GRCh38
rs952517573 Chr.12: 131728367-131728367 on Build GRCh38
rs200747413 Chr.12: 131728405-131728405 on Build GRCh38
rs756349766 Chr.12: 131728243-131728243 on Build GRCh38
rs775212941 Chr.12: 131728283-131728283 on Build GRCh38
rs762582212 Chr.12: 131728284-131728284 on Build GRCh38
rs1052827860 Chr.12: 131728456-131728456 on Build GRCh38
rs747147320 Chr.12: 131728441-131728442 on Build GRCh38
rs899379315 Chr.12: 131728272-131728272 on Build GRCh38
rs774864277 Chr.12: 131728257-131728257 on Build GRCh38
rs982224793 Chr.12: 131728549-131728549 on Build GRCh38
rs757180902 Chr.12: 131728304-131728304 on Build GRCh38
rs779090220 Chr.12: 131728442-131728442 on Build GRCh38
rs966329293 Chr.12: 131728545-131728545 on Build GRCh38
rs146357379 Chr.12: 131728334-131728334 on Build GRCh38
rs767477305 Chr.12: 131728322-131728322 on Build GRCh38
rs754014252 Chr.12: 131728338-131728338 on Build GRCh38
rs533199470 Chr.12: 131728431-131728431 on Build GRCh38
rs796950098 Chr.12: 131728561-131728561 on Build GRCh38
rs780193419 Chr.12: 131728246-131728246 on Build GRCh38
rs372391452 Chr.12: 131728355-131728355 on Build GRCh38
rs1008219735 Chr.12: 131728484-131728484 on Build GRCh38
rs990964342 Chr.12: 131728571-131728571 on Build GRCh38
rs151066029 Chr.12: 131728325-131728325 on Build GRCh38
rs772509588 Chr.12: 131728375-131728375 on Build GRCh38
rs989367914 Chr.12: 131728476-131728476 on Build GRCh38
rs758434491 Chr.12: 131728451-131728451 on Build GRCh38
rs116484519 Chr.12: 131728271-131728271 on Build GRCh38
rs751089015 Chr.12: 131728301-131728301 on Build GRCh38
rs772604240 Chr.12: 131728263-131728263 on Build GRCh38
rs774700613 Chr.12: 131728544-131728544 on Build GRCh38
rs11246775 Chr.12: 131728563-131728563 on Build GRCh38
rs764806553 Chr.12: 131728278-131728278 on Build GRCh38
rs1015800209 Chr.12: 131728642-131728642 on Build GRCh38
rs748657233 Chr.12: 131728260-131728260 on Build GRCh38
rs760410108 Chr.12: 131728415-131728415 on Build GRCh38
rs375791954 Chr.12: 131728477-131728477 on Build GRCh38
rs1027136576 Chr.12: 131728357-131728357 on Build GRCh38
rs368504316 Chr.12: 131728385-131728385 on Build GRCh38
rs537136358 Chr.12: 131728290-131728290 on Build GRCh38
rs200297330 Chr.12: 131728372-131728372 on Build GRCh38
rs1026836671 Chr.12: 131728601-131728601 on Build GRCh38
rs139185938 Chr.12: 131728380-131728380 on Build GRCh38
rs755174782 Chr.12: 131728339-131728339 on Build GRCh38
rs886730826 Chr.12: 131728627-131728627 on Build GRCh38
rs762743632 Chr.12: 131728386-131728386 on Build GRCh38
rs773799126 Chr.12: 131728391-131728391 on Build GRCh38
rs777001604 Chr.12: 131728477-131728477 on Build GRCh38
rs112989257 Chr.12: 131728550-131728550 on Build GRCh38
rs772838996 Chr.12: 131728253-131728253 on Build GRCh38
rs753601458 Chr.12: 131728427-131728427 on Build GRCh38
rs888163639 Chr.12: 131728602-131728602 on Build GRCh38
rs868340742 Chr.12: 131728623-131728623 on Build GRCh38
rs918207143 Chr.12: 131728397-131728397 on Build GRCh38
rs764404750 Chr.12: 131728463-131728463 on Build GRCh38
rs938190338 Chr.12: 131728242-131728242 on Build GRCh38
rs747499684 Chr.12: 131728471-131728471 on Build GRCh38
rs541335391 Chr.12: 131728403-131728403 on Build GRCh38
rs996616172 Chr.12: 131728599-131728599 on Build GRCh38
rs746570772 Chr.12: 131728254-131728254 on Build GRCh38

View Full Product Details