Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 131908795-131909293 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ULK1
Entrez Gene ID
8408
Gene Name
unc-51 like autophagy activating kinase 1
Gene Aliases
ATG1, ATG1A, UNC51, Unc51.1, hATG1
UniGene
Hs.47061
Gene Chromosome Location
Chr.12: 131894734-131923162 on Build GRCh38
Transcripts
NM_003565.2 XM_011538798.2 XM_011538799.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5341959 c.554C>T p.P185L Chr.12: 131908961-131908961 on Build GRCh38
6816587 c.621C>T p.I207I Chr.12: 131909192-131909192 on Build GRCh38
937116 c.504C>T p.F168F Chr.12: 131908911-131908911 on Build GRCh38
4040204 c.529A>G p.M177V Chr.12: 131908936-131908936 on Build GRCh38
5361946 c.479G>T p.R160L Chr.12: 131908806-131908806 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767234260 Chr.12: 131909240-131909240 on Build GRCh38
rs774525992 Chr.12: 131908999-131908999 on Build GRCh38
rs759903037 Chr.12: 131908848-131908848 on Build GRCh38
rs367995902 Chr.12: 131908923-131908923 on Build GRCh38
rs747974505 Chr.12: 131908867-131908867 on Build GRCh38
rs765932904 Chr.12: 131908905-131908905 on Build GRCh38
rs757057620 Chr.12: 131909023-131909023 on Build GRCh38
rs781329369 Chr.12: 131909096-131909096 on Build GRCh38
rs767331303 Chr.12: 131908853-131908853 on Build GRCh38
rs548126062 Chr.12: 131909254-131909254 on Build GRCh38
rs1039237766 Chr.12: 131909085-131909085 on Build GRCh38
rs779346547 Chr.12: 131909169-131909169 on Build GRCh38
rs1057170606 Chr.12: 131909007-131909007 on Build GRCh38
rs748566843 Chr.12: 131909202-131909202 on Build GRCh38
rs146729534 Chr.12: 131909219-131909219 on Build GRCh38
rs369403948 Chr.12: 131909118-131909118 on Build GRCh38
rs779442736 Chr.12: 131909113-131909113 on Build GRCh38
rs781448917 Chr.12: 131908988-131908988 on Build GRCh38
rs886562902 Chr.12: 131909131-131909131 on Build GRCh38
rs947198242 Chr.12: 131908947-131908947 on Build GRCh38
rs771055804 Chr.12: 131908871-131908871 on Build GRCh38
rs910428495 Chr.12: 131908893-131908893 on Build GRCh38
rs754479000 Chr.12: 131908917-131908917 on Build GRCh38
rs776524514 Chr.12: 131908872-131908872 on Build GRCh38
rs143236084 Chr.12: 131909203-131909203 on Build GRCh38
rs747277457 Chr.12: 131908980-131908980 on Build GRCh38
rs562665511 Chr.12: 131909176-131909176 on Build GRCh38
rs373988299 Chr.12: 131908839-131908839 on Build GRCh38
rs749174743 Chr.12: 131908820-131908820 on Build GRCh38
rs533542000 Chr.12: 131908868-131908868 on Build GRCh38
rs368400125 Chr.12: 131909001-131909001 on Build GRCh38
rs1035049384 Chr.12: 131909162-131909162 on Build GRCh38
rs769152398 Chr.12: 131908998-131908998 on Build GRCh38
rs761747976 Chr.12: 131909234-131909234 on Build GRCh38
rs370177188 Chr.12: 131908870-131908870 on Build GRCh38
rs759131114 Chr.12: 131909119-131909119 on Build GRCh38
rs777488587 Chr.12: 131908968-131908968 on Build GRCh38
rs756264600 Chr.12: 131909102-131909102 on Build GRCh38
rs748508281 Chr.12: 131909115-131909115 on Build GRCh38
rs780309745 Chr.12: 131909165-131909165 on Build GRCh38
rs567759604 Chr.12: 131908849-131908849 on Build GRCh38
rs750084575 Chr.12: 131908854-131908854 on Build GRCh38
rs776322558 Chr.12: 131909003-131909003 on Build GRCh38
rs780094178 Chr.12: 131908816-131908816 on Build GRCh38
rs143962966 Chr.12: 131909064-131909064 on Build GRCh38
rs567992862 Chr.12: 131909257-131909257 on Build GRCh38
rs990724453 Chr.12: 131908941-131908941 on Build GRCh38
rs761230658 Chr.12: 131908864-131908865 on Build GRCh38
rs748391153 Chr.12: 131908828-131908828 on Build GRCh38
rs753823741 Chr.12: 131908861-131908861 on Build GRCh38
rs1022667099 Chr.12: 131909028-131909028 on Build GRCh38
rs745552807 Chr.12: 131909201-131909201 on Build GRCh38
rs367989623 Chr.12: 131908862-131908862 on Build GRCh38
rs753167888 Chr.12: 131909267-131909267 on Build GRCh38
rs769242645 Chr.12: 131909108-131909108 on Build GRCh38
rs1050751677 Chr.12: 131909058-131909058 on Build GRCh38
rs774163203 Chr.12: 131909228-131909228 on Build GRCh38
rs139527932 Chr.12: 131909141-131909141 on Build GRCh38
rs747134855 Chr.12: 131908869-131908869 on Build GRCh38
rs544961930 Chr.12: 131909060-131909060 on Build GRCh38
rs758195265 Chr.12: 131908934-131908934 on Build GRCh38
rs754009822 Chr.12: 131909166-131909166 on Build GRCh38
rs200288395 Chr.12: 131909127-131909127 on Build GRCh38
rs201483006 Chr.12: 131908882-131908882 on Build GRCh38
rs778617910 Chr.12: 131908827-131908827 on Build GRCh38
rs553480723 Chr.12: 131908894-131908894 on Build GRCh38
rs753245967 Chr.12: 131908916-131908916 on Build GRCh38
rs758703423 Chr.12: 131909181-131909181 on Build GRCh38
rs758899209 Chr.12: 131909272-131909272 on Build GRCh38
rs755175672 Chr.12: 131909262-131909262 on Build GRCh38
rs575857025 Chr.12: 131909006-131909006 on Build GRCh38
rs555295917 Chr.12: 131909071-131909071 on Build GRCh38
rs772451739 Chr.12: 131908824-131908825 on Build GRCh38
rs967907861 Chr.12: 131909268-131909268 on Build GRCh38
rs983819114 Chr.12: 131908902-131908902 on Build GRCh38
rs950755092 Chr.12: 131908895-131908895 on Build GRCh38
rs201590771 Chr.12: 131909101-131909101 on Build GRCh38
rs773099095 Chr.12: 131909252-131909252 on Build GRCh38
rs748857333 Chr.12: 131909218-131909218 on Build GRCh38
rs532612536 Chr.12: 131909150-131909150 on Build GRCh38
rs776121172 Chr.12: 131908874-131908874 on Build GRCh38
rs747630517 Chr.12: 131909008-131909008 on Build GRCh38
rs373850625 Chr.12: 131908873-131908873 on Build GRCh38
rs769316601 Chr.12: 131909123-131909123 on Build GRCh38
rs750620536 Chr.12: 131909158-131909158 on Build GRCh38
rs778113484 Chr.12: 131909273-131909273 on Build GRCh38
rs752616966 Chr.12: 131908927-131908927 on Build GRCh38
rs375108812 Chr.12: 131909000-131909000 on Build GRCh38
rs766755852 Chr.12: 131909256-131909256 on Build GRCh38
rs769984036 Chr.12: 131908994-131908994 on Build GRCh38
rs1053373552 Chr.12: 131908832-131908832 on Build GRCh38
rs763473156 Chr.12: 131908876-131908876 on Build GRCh38
rs984376648 Chr.12: 131908850-131908850 on Build GRCh38
rs757382289 Chr.12: 131908987-131908987 on Build GRCh38
rs756352855 Chr.12: 131909161-131909161 on Build GRCh38
rs768497731 Chr.12: 131908826-131908826 on Build GRCh38
rs762420187 Chr.12: 131909132-131909132 on Build GRCh38
rs755714351 Chr.12: 131908858-131908858 on Build GRCh38
rs775767036 Chr.12: 131908844-131908865 on Build GRCh38
rs747139719 Chr.12: 131908834-131908834 on Build GRCh38
rs545685871 Chr.12: 131909168-131909168 on Build GRCh38
rs372632934 Chr.12: 131909004-131909004 on Build GRCh38
rs370336430 Chr.12: 131908863-131908863 on Build GRCh38
rs763381010 Chr.12: 131908852-131908852 on Build GRCh38
rs377240298 Chr.12: 131909014-131909014 on Build GRCh38
rs373368474 Chr.12: 131909255-131909255 on Build GRCh38
rs778150876 Chr.12: 131909197-131909197 on Build GRCh38
rs765293939 Chr.12: 131909264-131909264 on Build GRCh38
rs780223294 Chr.12: 131909103-131909103 on Build GRCh38
rs751354182 Chr.12: 131909017-131909017 on Build GRCh38
rs375826891 Chr.12: 131909104-131909104 on Build GRCh38
rs531590488 Chr.12: 131908833-131908833 on Build GRCh38
rs964829441 Chr.12: 131908875-131908875 on Build GRCh38

View Full Product Details