Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 132144366-132144864 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOC4L
Entrez Gene ID
79050
Gene Name
nucleolar complex associated 4 homolog
Gene Aliases
NET49, NOC4, UTP19
UniGene
Hs.558536
Gene Chromosome Location
Chr.12: 132144421-132152473 on Build GRCh38
Transcripts
NM_024078.2 XM_017019960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7317773 c.40C>T p.L14L Chr.12: 132144528-132144528 on Build GRCh38
6249352 c.124G>T p.D42Y Chr.12: 132144860-132144860 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs922513257 Chr.12: 132144839-132144839 on Build GRCh38
rs769293164 Chr.12: 132144455-132144455 on Build GRCh38
rs752669340 Chr.12: 132144585-132144585 on Build GRCh38
rs576327785 Chr.12: 132144570-132144570 on Build GRCh38
rs547395275 Chr.12: 132144801-132144801 on Build GRCh38
rs1014403442 Chr.12: 132144685-132144685 on Build GRCh38
rs768290552 Chr.12: 132144519-132144519 on Build GRCh38
rs764488335 Chr.12: 132144595-132144595 on Build GRCh38
rs894662515 Chr.12: 132144664-132144664 on Build GRCh38
rs1053662768 Chr.12: 132144384-132144384 on Build GRCh38
rs778220342 Chr.12: 132144500-132144500 on Build GRCh38
rs756373349 Chr.12: 132144548-132144548 on Build GRCh38
rs994195767 Chr.12: 132144414-132144414 on Build GRCh38
rs982733285 Chr.12: 132144767-132144767 on Build GRCh38
rs999105405 Chr.12: 132144795-132144795 on Build GRCh38
rs906743279 Chr.12: 132144634-132144634 on Build GRCh38
rs1047134593 Chr.12: 132144596-132144596 on Build GRCh38
rs1047063225 Chr.12: 132144842-132144842 on Build GRCh38
rs770717133 Chr.12: 132144836-132144836 on Build GRCh38
rs370351959 Chr.12: 132144417-132144417 on Build GRCh38
rs967642589 Chr.12: 132144768-132144768 on Build GRCh38
rs534111025 Chr.12: 132144466-132144466 on Build GRCh38
rs759774718 Chr.12: 132144840-132144840 on Build GRCh38
rs771866737 Chr.12: 132144523-132144523 on Build GRCh38
rs773097883 Chr.12: 132144526-132144526 on Build GRCh38
rs753205984 Chr.12: 132144495-132144495 on Build GRCh38
rs868755926 Chr.12: 132144429-132144429 on Build GRCh38
rs568655385 Chr.12: 132144434-132144434 on Build GRCh38
rs1005643872 Chr.12: 132144636-132144636 on Build GRCh38
rs1025723193 Chr.12: 132144415-132144415 on Build GRCh38
rs556531728 Chr.12: 132144522-132144522 on Build GRCh38
rs1053469107 Chr.12: 132144590-132144590 on Build GRCh38
rs765800396 Chr.12: 132144492-132144492 on Build GRCh38
rs1034527907 Chr.12: 132144428-132144428 on Build GRCh38
rs111232707 Chr.12: 132144799-132144799 on Build GRCh38
rs973996354 Chr.12: 132144420-132144420 on Build GRCh38
rs975176433 Chr.12: 132144796-132144796 on Build GRCh38
rs753964630 Chr.12: 132144486-132144486 on Build GRCh38
rs888491135 Chr.12: 132144614-132144614 on Build GRCh38
rs577120175 Chr.12: 132144475-132144475 on Build GRCh38
rs989261495 Chr.12: 132144821-132144821 on Build GRCh38
rs781466597 Chr.12: 132144841-132144841 on Build GRCh38
rs774430255 Chr.12: 132144464-132144464 on Build GRCh38
rs111829499 Chr.12: 132144701-132144701 on Build GRCh38
rs892695344 Chr.12: 132144597-132144597 on Build GRCh38
rs541224864 Chr.12: 132144760-132144760 on Build GRCh38
rs572231341 Chr.12: 132144726-132144726 on Build GRCh38
rs912398108 Chr.12: 132144770-132144770 on Build GRCh38
rs1041036330 Chr.12: 132144398-132144398 on Build GRCh38
rs535540311 Chr.12: 132144413-132144413 on Build GRCh38
rs889660612 Chr.12: 132144390-132144390 on Build GRCh38
rs777554422 Chr.12: 132144831-132144831 on Build GRCh38
rs766554183 Chr.12: 132144485-132144485 on Build GRCh38
rs752019782 Chr.12: 132144599-132144599 on Build GRCh38
rs748786527 Chr.12: 132144463-132144463 on Build GRCh38
rs201538803 Chr.12: 132144509-132144509 on Build GRCh38
rs548777530 Chr.12: 132144418-132144418 on Build GRCh38
rs995224704 Chr.12: 132144723-132144723 on Build GRCh38
rs751841077 Chr.12: 132144508-132144508 on Build GRCh38
rs761592853 Chr.12: 132144465-132144465 on Build GRCh38
rs771197580 Chr.12: 132144541-132144541 on Build GRCh38
rs775182226 Chr.12: 132144458-132144458 on Build GRCh38
rs747577997 Chr.12: 132144827-132144827 on Build GRCh38
rs533284385 Chr.12: 132144394-132144394 on Build GRCh38
rs1046513358 Chr.12: 132144631-132144631 on Build GRCh38
rs187880223 Chr.12: 132144820-132144820 on Build GRCh38
rs148548142 Chr.12: 132144469-132144469 on Build GRCh38
rs965231961 Chr.12: 132144774-132144774 on Build GRCh38
rs776352145 Chr.12: 132144837-132144837 on Build GRCh38
rs763557627 Chr.12: 132144591-132144591 on Build GRCh38
rs757692953 Chr.12: 132144630-132144630 on Build GRCh38
rs911068495 Chr.12: 132144506-132144506 on Build GRCh38
rs542322714 Chr.12: 132144695-132144695 on Build GRCh38
rs759594921 Chr.12: 132144491-132144491 on Build GRCh38
rs1002239931 Chr.12: 132144650-132144650 on Build GRCh38
rs748874647 Chr.12: 132144513-132144513 on Build GRCh38
rs746884473 Chr.12: 132144833-132144833 on Build GRCh38
rs758101024 Chr.12: 132144466-132144467 on Build GRCh38
rs532409250 Chr.12: 132144478-132144478 on Build GRCh38
rs1056338826 Chr.12: 132144643-132144643 on Build GRCh38
rs765313291 Chr.12: 132144572-132144572 on Build GRCh38
rs1033491486 Chr.12: 132144830-132144830 on Build GRCh38
rs1026319031 Chr.12: 132144742-132144742 on Build GRCh38
rs745545565 Chr.12: 132144449-132144449 on Build GRCh38
rs778512981 Chr.12: 132144520-132144520 on Build GRCh38
rs758772704 Chr.12: 132144496-132144496 on Build GRCh38
rs1034201110 Chr.12: 132144699-132144699 on Build GRCh38
rs760360991 Chr.12: 132144540-132144540 on Build GRCh38
rs778600944 Chr.12: 132144825-132144825 on Build GRCh38
rs1009661948 Chr.12: 132144388-132144388 on Build GRCh38
rs781638000 Chr.12: 132144804-132144804 on Build GRCh38
rs776828511 Chr.12: 132144556-132144556 on Build GRCh38
rs533274722 Chr.12: 132144828-132144828 on Build GRCh38
rs1024535987 Chr.12: 132144435-132144435 on Build GRCh38

View Full Product Details