Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42386064-42386562 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN3
Entrez Gene ID
825
Gene Name
calpain 3
Gene Aliases
CANP3, CANPL3, LGMD2, LGMD2A, nCL-1, p94
UniGene
Hs.143261
Gene Chromosome Location
Chr.15: 42359500-42412317 on Build GRCh38
Transcripts
NM_000070.2 NM_024344.1 NM_173087.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3501278 c.433C>T p.L145F Chr.15: 42386220-42386220 on Build GRCh38
6221271 c.140C>T p.A47V Chr.15: 42386188-42386188 on Build GRCh38
6221270 c.401C>T p.A134V Chr.15: 42386188-42386188 on Build GRCh38
7078992 c.419T>C p.L140P Chr.15: 42386206-42386206 on Build GRCh38
7078991 c.419T>C p.L140P Chr.15: 42386206-42386206 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1055901249 Chr.15: 42386388-42386388 on Build GRCh38
rs200469580 Chr.15: 42386288-42386288 on Build GRCh38
rs867162013 Chr.15: 42386347-42386347 on Build GRCh38
rs35855730 Chr.15: 42386256-42386257 on Build GRCh38
rs768543005 Chr.15: 42386310-42386320 on Build GRCh38
rs1041476214 Chr.15: 42386333-42386333 on Build GRCh38
rs863224963 Chr.15: 42386270-42386270 on Build GRCh38
rs1801324 Chr.15: 42386282-42386282 on Build GRCh38
rs1042151947 Chr.15: 42386199-42386199 on Build GRCh38
rs753376577 Chr.15: 42386335-42386335 on Build GRCh38
rs982848892 Chr.15: 42386144-42386144 on Build GRCh38
rs747738726 Chr.15: 42386134-42386134 on Build GRCh38
rs904229952 Chr.15: 42386372-42386372 on Build GRCh38
rs551835368 Chr.15: 42386125-42386125 on Build GRCh38
rs775324529 Chr.15: 42386408-42386408 on Build GRCh38
rs375163994 Chr.15: 42386120-42386120 on Build GRCh38
rs539965498 Chr.15: 42386301-42386301 on Build GRCh38
rs139671324 Chr.15: 42386227-42386227 on Build GRCh38
rs977860525 Chr.15: 42386511-42386511 on Build GRCh38
rs749697583 Chr.15: 42386265-42386265 on Build GRCh38
rs886051148 Chr.15: 42386238-42386238 on Build GRCh38
rs886043431 Chr.15: 42386189-42386189 on Build GRCh38
rs774685118 Chr.15: 42386185-42386185 on Build GRCh38
rs760975555 Chr.15: 42386205-42386205 on Build GRCh38
rs143942248 Chr.15: 42386255-42386255 on Build GRCh38
rs886043025 Chr.15: 42386263-42386263 on Build GRCh38
rs746311570 Chr.15: 42386279-42386279 on Build GRCh38
rs560110587 Chr.15: 42386402-42386402 on Build GRCh38
rs878854364 Chr.15: 42386226-42386226 on Build GRCh38
rs879146589 Chr.15: 42386113-42386113 on Build GRCh38
rs761027377 Chr.15: 42386285-42386285 on Build GRCh38
rs772347683 Chr.15: 42386280-42386280 on Build GRCh38
rs200996550 Chr.15: 42386131-42386131 on Build GRCh38
rs746935735 Chr.15: 42386188-42386188 on Build GRCh38
rs28364395 Chr.15: 42386088-42386088 on Build GRCh38
rs185202757 Chr.15: 42386307-42386307 on Build GRCh38
rs765001171 Chr.15: 42386218-42386218 on Build GRCh38
rs191314388 Chr.15: 42386096-42386096 on Build GRCh38
rs770055574 Chr.15: 42386138-42386138 on Build GRCh38
rs886044404 Chr.15: 42386220-42386220 on Build GRCh38
rs754494701 Chr.15: 42386244-42386244 on Build GRCh38
rs762837895 Chr.15: 42386232-42386232 on Build GRCh38
rs759713838 Chr.15: 42386189-42386189 on Build GRCh38
rs376337969 Chr.15: 42386305-42386305 on Build GRCh38
rs991353376 Chr.15: 42386108-42386108 on Build GRCh38
rs555686014 Chr.15: 42386334-42386334 on Build GRCh38
rs1004756898 Chr.15: 42386407-42386407 on Build GRCh38
rs766369281 Chr.15: 42386239-42386239 on Build GRCh38
rs899265656 Chr.15: 42386326-42386326 on Build GRCh38
rs751448923 Chr.15: 42386243-42386243 on Build GRCh38
rs12438453 Chr.15: 42386411-42386411 on Build GRCh38
rs886042672 Chr.15: 42386229-42386229 on Build GRCh38
rs562241410 Chr.15: 42386480-42386480 on Build GRCh38
rs967702617 Chr.15: 42386451-42386451 on Build GRCh38
rs779579276 Chr.15: 42386267-42386267 on Build GRCh38
rs999943646 Chr.15: 42386381-42386381 on Build GRCh38
rs28364398 Chr.15: 42386317-42386317 on Build GRCh38
rs574653247 Chr.15: 42386397-42386397 on Build GRCh38
rs568666121 Chr.15: 42386192-42386192 on Build GRCh38
rs955334059 Chr.15: 42386508-42386508 on Build GRCh38
rs28364399 Chr.15: 42386320-42386320 on Build GRCh38
rs200944841 Chr.15: 42386256-42386256 on Build GRCh38
rs946415346 Chr.15: 42386184-42386184 on Build GRCh38
rs886043254 Chr.15: 42386149-42386164 on Build GRCh38
rs923701668 Chr.15: 42386537-42386537 on Build GRCh38
rs548911867 Chr.15: 42386141-42386141 on Build GRCh38
rs1022003111 Chr.15: 42386415-42386415 on Build GRCh38
rs28364396 Chr.15: 42386140-42386140 on Build GRCh38
rs780611743 Chr.15: 42386253-42386253 on Build GRCh38
rs28364400 Chr.15: 42386398-42386398 on Build GRCh38
rs185374433 Chr.15: 42386101-42386101 on Build GRCh38
rs775861630 Chr.15: 42386195-42386195 on Build GRCh38
rs28364401 Chr.15: 42386412-42386412 on Build GRCh38
rs931921898 Chr.15: 42386291-42386291 on Build GRCh38
rs1022861629 Chr.15: 42386114-42386114 on Build GRCh38
rs959308080 Chr.15: 42386129-42386129 on Build GRCh38
rs17592 Chr.15: 42386266-42386266 on Build GRCh38

View Full Product Details