Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42728513-42729015 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDAN1
Entrez Gene ID
146059
Gene Name
codanin 1
Gene Aliases
CDA1, CDAI, CDAN1A, DLT, PRO1295
UniGene
Hs.599232
Gene Chromosome Location
Chr.15: 42723557-42737219 on Build GRCh38
Transcripts
NM_138477.2 XM_005254176.4 XM_011521270.2 XM_011521271.2 XM_011521274.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5849191 c.2725G>A p.G909R Chr.15: 42728731-42728731 on Build GRCh38
3754317 c.2671C>T p.R891C Chr.15: 42728785-42728785 on Build GRCh38
6142144 c.2802G>A p.R934R Chr.15: 42728654-42728654 on Build GRCh38
6410033 c.2797G>T p.G933W Chr.15: 42728659-42728659 on Build GRCh38
3690399 c.2746G>A p.E916K Chr.15: 42728710-42728710 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370115296 Chr.15: 42728977-42728977 on Build GRCh38
rs768327667 Chr.15: 42728685-42728685 on Build GRCh38
rs763487399 Chr.15: 42728761-42728761 on Build GRCh38
rs149472555 Chr.15: 42728713-42728713 on Build GRCh38
rs557295691 Chr.15: 42728778-42728778 on Build GRCh38
rs138273028 Chr.15: 42728568-42728568 on Build GRCh38
rs747712384 Chr.15: 42728745-42728745 on Build GRCh38
rs374837444 Chr.15: 42728948-42728950 on Build GRCh38
rs1010698341 Chr.15: 42728559-42728561 on Build GRCh38
rs762229479 Chr.15: 42728687-42728687 on Build GRCh38
rs543119360 Chr.15: 42728560-42728560 on Build GRCh38
rs774350952 Chr.15: 42728779-42728779 on Build GRCh38
rs577649668 Chr.15: 42728551-42728551 on Build GRCh38
rs765530912 Chr.15: 42728691-42728691 on Build GRCh38
rs745824109 Chr.15: 42728731-42728731 on Build GRCh38
rs572787333 Chr.15: 42728851-42728851 on Build GRCh38
rs962284435 Chr.15: 42728577-42728577 on Build GRCh38
rs771650996 Chr.15: 42728611-42728611 on Build GRCh38
rs776929659 Chr.15: 42728686-42728686 on Build GRCh38
rs754845295 Chr.15: 42728983-42728983 on Build GRCh38
rs773630644 Chr.15: 42728823-42728823 on Build GRCh38
rs1057167706 Chr.15: 42728712-42728712 on Build GRCh38
rs150657573 Chr.15: 42728794-42728794 on Build GRCh38
rs905412308 Chr.15: 42728916-42728916 on Build GRCh38
rs771226283 Chr.15: 42728833-42728833 on Build GRCh38
rs993298768 Chr.15: 42728925-42728925 on Build GRCh38
rs750630547 Chr.15: 42728774-42728775 on Build GRCh38
rs183816495 Chr.15: 42728583-42728583 on Build GRCh38
rs779007555 Chr.15: 42728679-42728679 on Build GRCh38
rs369518566 Chr.15: 42728813-42728813 on Build GRCh38
rs965647472 Chr.15: 42728613-42728613 on Build GRCh38
rs377408193 Chr.15: 42728854-42728854 on Build GRCh38
rs1056298858 Chr.15: 42728911-42728911 on Build GRCh38
rs993416302 Chr.15: 42728726-42728726 on Build GRCh38
rs150468048 Chr.15: 42728566-42728566 on Build GRCh38
rs904929368 Chr.15: 42728859-42728859 on Build GRCh38
rs76947588 Chr.15: 42728718-42728718 on Build GRCh38
rs774925862 Chr.15: 42728835-42728835 on Build GRCh38
rs148040924 Chr.15: 42728946-42728948 on Build GRCh38
rs1010049752 Chr.15: 42728962-42728962 on Build GRCh38
rs200401359 Chr.15: 42728680-42728680 on Build GRCh38
rs887708389 Chr.15: 42728964-42728972 on Build GRCh38
rs780031588 Chr.15: 42728803-42728803 on Build GRCh38
rs753653211 Chr.15: 42728575-42728576 on Build GRCh38
rs754525241 Chr.15: 42728808-42728808 on Build GRCh38
rs376721314 Chr.15: 42728744-42728744 on Build GRCh38
rs777468443 Chr.15: 42728670-42728670 on Build GRCh38
rs988354822 Chr.15: 42728797-42728797 on Build GRCh38
rs768815034 Chr.15: 42728843-42728845 on Build GRCh38
rs1019726571 Chr.15: 42728987-42728987 on Build GRCh38
rs761020947 Chr.15: 42728848-42728848 on Build GRCh38
rs542817784 Chr.15: 42728580-42728580 on Build GRCh38
rs750563812 Chr.15: 42728643-42728643 on Build GRCh38
rs1000891073 Chr.15: 42728553-42728553 on Build GRCh38
rs756122419 Chr.15: 42728625-42728625 on Build GRCh38
rs975377036 Chr.15: 42728638-42728638 on Build GRCh38
rs765733042 Chr.15: 42728642-42728642 on Build GRCh38
rs762083726 Chr.15: 42728976-42728977 on Build GRCh38
rs138627747 Chr.15: 42728846-42728846 on Build GRCh38
rs754211126 Chr.15: 42728632-42728632 on Build GRCh38
rs371548844 Chr.15: 42728652-42728652 on Build GRCh38
rs772114815 Chr.15: 42728733-42728733 on Build GRCh38
rs61745640 Chr.15: 42728756-42728756 on Build GRCh38
rs201131708 Chr.15: 42728806-42728806 on Build GRCh38
rs367943698 Chr.15: 42728617-42728617 on Build GRCh38
rs528418245 Chr.15: 42728597-42728597 on Build GRCh38
rs778811422 Chr.15: 42728730-42728730 on Build GRCh38
rs533382887 Chr.15: 42728615-42728615 on Build GRCh38
rs775586084 Chr.15: 42728632-42728633 on Build GRCh38
rs762269501 Chr.15: 42728637-42728637 on Build GRCh38
rs143086237 Chr.15: 42728721-42728721 on Build GRCh38
rs760295520 Chr.15: 42728682-42728682 on Build GRCh38
rs201778116 Chr.15: 42728681-42728681 on Build GRCh38
rs374649955 Chr.15: 42728991-42728991 on Build GRCh38
rs892656090 Chr.15: 42728947-42728947 on Build GRCh38
rs201407882 Chr.15: 42728782-42728782 on Build GRCh38
rs577917690 Chr.15: 42728920-42728920 on Build GRCh38
rs759919615 Chr.15: 42728836-42728836 on Build GRCh38
rs921081061 Chr.15: 42728639-42728639 on Build GRCh38
rs111609226 Chr.15: 42728784-42728784 on Build GRCh38
rs751799284 Chr.15: 42728656-42728656 on Build GRCh38
rs769402481 Chr.15: 42728819-42728819 on Build GRCh38
rs921227010 Chr.15: 42728788-42728788 on Build GRCh38
rs776992129 Chr.15: 42728859-42728860 on Build GRCh38
rs200089015 Chr.15: 42728860-42728860 on Build GRCh38
rs372240589 Chr.15: 42728722-42728722 on Build GRCh38
rs972747648 Chr.15: 42728578-42728579 on Build GRCh38
rs142713606 Chr.15: 42728988-42728988 on Build GRCh38
rs747879397 Chr.15: 42728814-42728814 on Build GRCh38
rs543111906 Chr.15: 42728576-42728577 on Build GRCh38
rs758478579 Chr.15: 42728800-42728800 on Build GRCh38
rs747225543 Chr.15: 42728688-42728689 on Build GRCh38
rs755202657 Chr.15: 42728662-42728662 on Build GRCh38
rs199753863 Chr.15: 42728655-42728655 on Build GRCh38
rs767658967 Chr.15: 42728839-42728839 on Build GRCh38
rs984204825 Chr.15: 42728601-42728601 on Build GRCh38
rs912570519 Chr.15: 42728589-42728589 on Build GRCh38
rs749638337 Chr.15: 42728831-42728831 on Build GRCh38
rs199886860 Chr.15: 42728583-42728584 on Build GRCh38
rs758571284 Chr.15: 42728972-42728973 on Build GRCh38
rs747651097 Chr.15: 42728986-42728986 on Build GRCh38
rs749102959 Chr.15: 42728676-42728676 on Build GRCh38
rs564837925 Chr.15: 42728907-42728907 on Build GRCh38
rs770830490 Chr.15: 42728677-42728677 on Build GRCh38
rs773540721 Chr.15: 42728759-42728759 on Build GRCh38
rs946371123 Chr.15: 42728705-42728705 on Build GRCh38
rs746526678 Chr.15: 42728734-42728734 on Build GRCh38
rs1052097537 Chr.15: 42728668-42728668 on Build GRCh38
rs952797061 Chr.15: 42728587-42728587 on Build GRCh38
rs529452785 Chr.15: 42728706-42728706 on Build GRCh38
rs1005495851 Chr.15: 42728966-42728966 on Build GRCh38
rs766865099 Chr.15: 42728765-42728765 on Build GRCh38
rs764390055 Chr.15: 42728627-42728627 on Build GRCh38
rs1045720349 Chr.15: 42728906-42728906 on Build GRCh38
rs8023524 Chr.15: 42728785-42728785 on Build GRCh38
rs564104534 Chr.15: 42728600-42728600 on Build GRCh38
rs939755578 Chr.15: 42728708-42728708 on Build GRCh38
rs1053850745 Chr.15: 42728937-42728937 on Build GRCh38
rs375343604 Chr.15: 42728796-42728796 on Build GRCh38
rs541005975 Chr.15: 42728599-42728599 on Build GRCh38
rs768254953 Chr.15: 42728743-42728743 on Build GRCh38
rs766492829 Chr.15: 42728714-42728714 on Build GRCh38

View Full Product Details