Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 25391794-25392263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RHCE
Entrez Gene ID
6006
Gene Name
Rh blood group CcEe antigens
Gene Aliases
CD240CE, RH, RH30A, RHC, RHE, RHIXB, RHPI, Rh4, RhIVb(J), RhVI, RhVIII
UniGene
Hs.449968
Gene Chromosome Location
Chr.1: 25362249-25430193 on Build GRCh38
Transcripts
NM_020485.4 NM_138616.3 NM_138617.3 NM_138618.3 XM_005245957.3 XM_006710810.3 XM_011541888.2 XM_011541889.2 XM_011541890.2 XM_011541891.2 XM_017002014.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5554860 c.586G>T p.G196C Chr.1: 25391994-25391994 on Build GRCh38
5941568 c.569C>T p.P190L Chr.1: 25392059-25392059 on Build GRCh38
5888618 c.493T>A p.Y165N Chr.1: 25392135-25392135 on Build GRCh38
7387468 c.577A>G p.K193E Chr.1: 25392051-25392051 on Build GRCh38
5941566 c.569C>T p.P190L Chr.1: 25392059-25392059 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772521070 Chr.1: 25391878-25391878 on Build GRCh38
rs146306079 Chr.1: 25392108-25392108 on Build GRCh38
rs1037378632 Chr.1: 25392220-25392220 on Build GRCh38
rs184101067 Chr.1: 25392136-25392136 on Build GRCh38
rs773423995 Chr.1: 25392158-25392158 on Build GRCh38
rs777377612 Chr.1: 25392124-25392124 on Build GRCh38
rs749326190 Chr.1: 25392157-25392158 on Build GRCh38
rs913271782 Chr.1: 25391866-25391866 on Build GRCh38
rs181678482 Chr.1: 25392212-25392212 on Build GRCh38
rs746303049 Chr.1: 25392134-25392134 on Build GRCh38
rs756120130 Chr.1: 25392169-25392169 on Build GRCh38
rs765506405 Chr.1: 25392152-25392152 on Build GRCh38
rs764256566 Chr.1: 25392095-25392095 on Build GRCh38
rs567798161 Chr.1: 25391909-25391909 on Build GRCh38
rs112970979 Chr.1: 25391965-25391965 on Build GRCh38
rs760149624 Chr.1: 25392002-25392002 on Build GRCh38
rs567575075 Chr.1: 25391999-25391999 on Build GRCh38
rs370087814 Chr.1: 25391988-25391988 on Build GRCh38
rs774947339 Chr.1: 25392000-25392000 on Build GRCh38
rs926070456 Chr.1: 25391887-25391887 on Build GRCh38
rs766776085 Chr.1: 25391996-25391996 on Build GRCh38
rs188935839 Chr.1: 25392175-25392175 on Build GRCh38
rs552953999 Chr.1: 25392189-25392189 on Build GRCh38
rs189225585 Chr.1: 25392115-25392115 on Build GRCh38
rs777709497 Chr.1: 25392141-25392142 on Build GRCh38
rs1053349 Chr.1: 25392114-25392114 on Build GRCh38
rs764289876 Chr.1: 25392021-25392021 on Build GRCh38
rs1053354 Chr.1: 25392034-25392034 on Build GRCh38
rs1020280601 Chr.1: 25392123-25392123 on Build GRCh38
rs376509301 Chr.1: 25392103-25392103 on Build GRCh38
rs747449449 Chr.1: 25392192-25392192 on Build GRCh38
rs368228913 Chr.1: 25392109-25392109 on Build GRCh38
rs898902121 Chr.1: 25392241-25392241 on Build GRCh38
rs200305051 Chr.1: 25392094-25392094 on Build GRCh38
rs767672600 Chr.1: 25392006-25392006 on Build GRCh38
rs940311309 Chr.1: 25392213-25392213 on Build GRCh38
rs1053350 Chr.1: 25392084-25392084 on Build GRCh38
rs573097889 Chr.1: 25392198-25392198 on Build GRCh38
rs768585920 Chr.1: 25392145-25392145 on Build GRCh38
rs199815681 Chr.1: 25392174-25392174 on Build GRCh38
rs754102926 Chr.1: 25392022-25392022 on Build GRCh38
rs779408591 Chr.1: 25392129-25392129 on Build GRCh38
rs769261867 Chr.1: 25392193-25392193 on Build GRCh38
rs1004570437 Chr.1: 25391997-25391997 on Build GRCh38
rs534550543 Chr.1: 25392060-25392060 on Build GRCh38
rs979281070 Chr.1: 25391880-25391880 on Build GRCh38
rs776819527 Chr.1: 25392146-25392146 on Build GRCh38
rs895062961 Chr.1: 25391916-25391916 on Build GRCh38
rs761275424 Chr.1: 25392141-25392141 on Build GRCh38
rs143372707 Chr.1: 25392019-25392019 on Build GRCh38
rs760100539 Chr.1: 25392071-25392071 on Build GRCh38
rs760614194 Chr.1: 25392088-25392088 on Build GRCh38
rs916442804 Chr.1: 25392179-25392179 on Build GRCh38
rs781037009 Chr.1: 25392116-25392116 on Build GRCh38
rs375227984 Chr.1: 25392171-25392171 on Build GRCh38
rs778641970 Chr.1: 25392180-25392180 on Build GRCh38
rs755533657 Chr.1: 25392121-25392121 on Build GRCh38
rs769695705 Chr.1: 25391976-25391977 on Build GRCh38
rs745537829 Chr.1: 25392186-25392186 on Build GRCh38
rs775811870 Chr.1: 25392139-25392139 on Build GRCh38
rs771385162 Chr.1: 25392050-25392050 on Build GRCh38
rs372119551 Chr.1: 25392083-25392083 on Build GRCh38
rs753965768 Chr.1: 25392102-25392102 on Build GRCh38
rs181246382 Chr.1: 25392118-25392118 on Build GRCh38
rs776160667 Chr.1: 25391949-25391949 on Build GRCh38
rs762006807 Chr.1: 25392147-25392147 on Build GRCh38
rs141398055 Chr.1: 25392026-25392026 on Build GRCh38
rs780664726 Chr.1: 25392040-25392040 on Build GRCh38
rs775130441 Chr.1: 25392161-25392161 on Build GRCh38
rs368705839 Chr.1: 25392044-25392044 on Build GRCh38
rs140421430 Chr.1: 25392128-25392128 on Build GRCh38
rs367815430 Chr.1: 25391983-25391983 on Build GRCh38
rs758079594 Chr.1: 25392023-25392023 on Build GRCh38
rs987753117 Chr.1: 25392119-25392119 on Build GRCh38
rs758794355 Chr.1: 25392188-25392188 on Build GRCh38
rs747817751 Chr.1: 25391964-25391964 on Build GRCh38
rs145624225 Chr.1: 25392163-25392163 on Build GRCh38
rs754954217 Chr.1: 25392039-25392039 on Build GRCh38
rs374591957 Chr.1: 25392043-25392043 on Build GRCh38
rs760937331 Chr.1: 25392020-25392020 on Build GRCh38
rs372268880 Chr.1: 25392156-25392156 on Build GRCh38
rs987468410 Chr.1: 25391852-25391852 on Build GRCh38
rs556663345 Chr.1: 25391900-25391900 on Build GRCh38
rs747674448 Chr.1: 25392042-25392042 on Build GRCh38
rs770920427 Chr.1: 25392189-25392189 on Build GRCh38

View Full Product Details