Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 26854406-26854902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZDHHC18
Entrez Gene ID
84243
Gene Name
zinc finger DHHC-type containing 18
Gene Aliases
DHHC-18, DHHC18
UniGene
Hs.523710
Gene Chromosome Location
Chr.1: 26826710-26855720 on Build GRCh38
Transcripts
NM_032283.2

SNP Detail

SNP ID Chromosome Location
rs139859763 Chr.1: 26854765-26854765 on Build GRCh38
rs149864376 Chr.1: 26854867-26854867 on Build GRCh38
rs920440359 Chr.1: 26854641-26854641 on Build GRCh38
rs1000770926 Chr.1: 26854513-26854513 on Build GRCh38
rs886543393 Chr.1: 26854529-26854529 on Build GRCh38
rs917112785 Chr.1: 26854496-26854496 on Build GRCh38
rs530015271 Chr.1: 26854850-26854850 on Build GRCh38
rs932037484 Chr.1: 26854644-26854644 on Build GRCh38
rs978515876 Chr.1: 26854734-26854736 on Build GRCh38
rs41303777 Chr.1: 26854553-26854553 on Build GRCh38
rs865917876 Chr.1: 26854827-26854827 on Build GRCh38
rs76153219 Chr.1: 26854574-26854574 on Build GRCh38
rs940797044 Chr.1: 26854843-26854843 on Build GRCh38
rs763761940 Chr.1: 26854614-26854614 on Build GRCh38
rs996872204 Chr.1: 26854568-26854568 on Build GRCh38
rs544421966 Chr.1: 26854651-26854651 on Build GRCh38
rs1037790295 Chr.1: 26854857-26854857 on Build GRCh38
rs772834662 Chr.1: 26854833-26854834 on Build GRCh38
rs74062531 Chr.1: 26854589-26854589 on Build GRCh38
rs754339418 Chr.1: 26854779-26854779 on Build GRCh38
rs903462565 Chr.1: 26854508-26854508 on Build GRCh38
rs530617618 Chr.1: 26854618-26854618 on Build GRCh38
rs973609023 Chr.1: 26854623-26854623 on Build GRCh38
rs958389725 Chr.1: 26854736-26854736 on Build GRCh38
rs1041779672 Chr.1: 26854721-26854721 on Build GRCh38
rs894656022 Chr.1: 26854798-26854798 on Build GRCh38
rs1033216675 Chr.1: 26854519-26854519 on Build GRCh38
rs145056973 Chr.1: 26854850-26854851 on Build GRCh38
rs890481129 Chr.1: 26854492-26854492 on Build GRCh38
rs560135890 Chr.1: 26854450-26854450 on Build GRCh38
rs745434310 Chr.1: 26854534-26854534 on Build GRCh38
rs1003152809 Chr.1: 26854876-26854876 on Build GRCh38
rs866259658 Chr.1: 26854831-26854831 on Build GRCh38
rs944672774 Chr.1: 26854708-26854708 on Build GRCh38
rs1038138548 Chr.1: 26854551-26854551 on Build GRCh38
rs944914025 Chr.1: 26854501-26854501 on Build GRCh38
rs1012001070 Chr.1: 26854548-26854548 on Build GRCh38
rs1023321683 Chr.1: 26854862-26854862 on Build GRCh38
rs773488685 Chr.1: 26854653-26854653 on Build GRCh38
rs766852055 Chr.1: 26854693-26854693 on Build GRCh38
rs949893734 Chr.1: 26854500-26854500 on Build GRCh38
rs893513257 Chr.1: 26854532-26854532 on Build GRCh38
rs41307935 Chr.1: 26854576-26854576 on Build GRCh38
rs925648959 Chr.1: 26854734-26854734 on Build GRCh38
rs903428856 Chr.1: 26854753-26854753 on Build GRCh38
rs1041981137 Chr.1: 26854504-26854504 on Build GRCh38
rs762531266 Chr.1: 26854608-26854608 on Build GRCh38
rs991080351 Chr.1: 26854755-26854755 on Build GRCh38
rs146134557 Chr.1: 26854654-26854654 on Build GRCh38
rs1013530718 Chr.1: 26854853-26854853 on Build GRCh38
rs962019836 Chr.1: 26854613-26854613 on Build GRCh38

View Full Product Details