Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 27100565-27101068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC9A1
Entrez Gene ID
6548
Gene Name
solute carrier family 9 member A1
Gene Aliases
APNH, LIKNS, NHE-1, NHE1, PPP1R143
UniGene
Hs.469116
Gene Chromosome Location
Chr.1: 27098809-27155130 on Build GRCh38
Transcripts
NM_003047.4 NR_046474.1 XM_011542021.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
680156 c.2171C>A p.P724Q Chr.1: 27100584-27100584 on Build GRCh38
5508663 c.2148C>T p.V716V Chr.1: 27100607-27100607 on Build GRCh38
6783676 c.2149A>G p.I717V Chr.1: 27100606-27100606 on Build GRCh38
5007271 c.2163G>A p.P721P Chr.1: 27100592-27100592 on Build GRCh38
5483810 c.2172G>A p.P724P Chr.1: 27100583-27100583 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1043358238 Chr.1: 27100666-27100666 on Build GRCh38
rs756281745 Chr.1: 27100783-27100783 on Build GRCh38
rs758416733 Chr.1: 27100584-27100584 on Build GRCh38
rs745617798 Chr.1: 27100593-27100593 on Build GRCh38
rs547713563 Chr.1: 27100792-27100792 on Build GRCh38
rs939479824 Chr.1: 27100934-27100934 on Build GRCh38
rs1043797356 Chr.1: 27100866-27100866 on Build GRCh38
rs376446353 Chr.1: 27100693-27100693 on Build GRCh38
rs1018085203 Chr.1: 27101013-27101013 on Build GRCh38
rs374837943 Chr.1: 27100662-27100662 on Build GRCh38
rs115896271 Chr.1: 27101023-27101023 on Build GRCh38
rs554798640 Chr.1: 27100949-27100949 on Build GRCh38
rs570823886 Chr.1: 27100797-27100797 on Build GRCh38
rs960551877 Chr.1: 27100714-27100714 on Build GRCh38
rs1007476176 Chr.1: 27101015-27101015 on Build GRCh38
rs752154321 Chr.1: 27100679-27100679 on Build GRCh38
rs759289316 Chr.1: 27100697-27100713 on Build GRCh38
rs915460896 Chr.1: 27100766-27100766 on Build GRCh38
rs775458580 Chr.1: 27100597-27100597 on Build GRCh38
rs910522190 Chr.1: 27100752-27100752 on Build GRCh38
rs924158884 Chr.1: 27100749-27100749 on Build GRCh38
rs548100387 Chr.1: 27100691-27100691 on Build GRCh38
rs779075735 Chr.1: 27100592-27100592 on Build GRCh38
rs574710633 Chr.1: 27100983-27100983 on Build GRCh38
rs1006280170 Chr.1: 27100999-27100999 on Build GRCh38
rs985201169 Chr.1: 27100744-27100744 on Build GRCh38
rs1034834708 Chr.1: 27100709-27100709 on Build GRCh38
rs577092208 Chr.1: 27101018-27101018 on Build GRCh38
rs148527788 Chr.1: 27100619-27100619 on Build GRCh38
rs34091185 Chr.1: 27101034-27101035 on Build GRCh38
rs913655787 Chr.1: 27100841-27100841 on Build GRCh38
rs757684057 Chr.1: 27100590-27100590 on Build GRCh38
rs778134656 Chr.1: 27100585-27100585 on Build GRCh38
rs545437423 Chr.1: 27101022-27101022 on Build GRCh38
rs1056337972 Chr.1: 27100938-27100938 on Build GRCh38
rs993331563 Chr.1: 27100977-27100977 on Build GRCh38
rs527976968 Chr.1: 27100782-27100782 on Build GRCh38
rs1036836372 Chr.1: 27100945-27100946 on Build GRCh38
rs554076679 Chr.1: 27101006-27101006 on Build GRCh38
rs1049013439 Chr.1: 27101002-27101002 on Build GRCh38
rs772086866 Chr.1: 27100595-27100595 on Build GRCh38
rs373292370 Chr.1: 27100656-27100656 on Build GRCh38
rs948060686 Chr.1: 27100871-27100871 on Build GRCh38
rs747523057 Chr.1: 27101046-27101046 on Build GRCh38
rs200339796 Chr.1: 27100657-27100657 on Build GRCh38
rs1053276200 Chr.1: 27100795-27100795 on Build GRCh38
rs946452735 Chr.1: 27100856-27100856 on Build GRCh38
rs993278864 Chr.1: 27100716-27100716 on Build GRCh38
rs182987443 Chr.1: 27100984-27100984 on Build GRCh38
rs769612061 Chr.1: 27100757-27100757 on Build GRCh38
rs1045136024 Chr.1: 27100589-27100589 on Build GRCh38
rs112347271 Chr.1: 27100758-27100758 on Build GRCh38
rs749600897 Chr.1: 27100586-27100586 on Build GRCh38
rs374596916 Chr.1: 27100678-27100678 on Build GRCh38
rs372414685 Chr.1: 27100687-27100687 on Build GRCh38
rs769630690 Chr.1: 27100606-27100606 on Build GRCh38
rs539421302 Chr.1: 27100812-27100812 on Build GRCh38
rs535981935 Chr.1: 27100935-27100935 on Build GRCh38
rs568356938 Chr.1: 27100770-27100770 on Build GRCh38
rs142887458 Chr.1: 27100626-27100626 on Build GRCh38
rs570011849 Chr.1: 27100924-27100924 on Build GRCh38
rs953036137 Chr.1: 27100711-27100711 on Build GRCh38
rs777894033 Chr.1: 27100713-27100713 on Build GRCh38
rs774129278 Chr.1: 27100648-27100648 on Build GRCh38
rs141836522 Chr.1: 27101043-27101043 on Build GRCh38
rs747200882 Chr.1: 27101014-27101014 on Build GRCh38
rs748049158 Chr.1: 27100598-27100598 on Build GRCh38
rs754407329 Chr.1: 27100690-27100690 on Build GRCh38
rs762655180 Chr.1: 27100625-27100625 on Build GRCh38
rs556523594 Chr.1: 27100923-27100923 on Build GRCh38
rs1026745412 Chr.1: 27100789-27100789 on Build GRCh38
rs952129356 Chr.1: 27100742-27100742 on Build GRCh38
rs1026392120 Chr.1: 27100741-27100741 on Build GRCh38
rs368913703 Chr.1: 27100683-27100683 on Build GRCh38
rs896401118 Chr.1: 27100972-27100972 on Build GRCh38
rs965347707 Chr.1: 27100753-27100753 on Build GRCh38
rs548335504 Chr.1: 27100701-27100701 on Build GRCh38

View Full Product Details