Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 40133739-40134243 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MSL1
Entrez Gene ID
339287
Gene Name
male specific lethal 1 homolog
Gene Aliases
MSL-1, hMSL1
UniGene
Hs.532786
Gene Chromosome Location
Chr.17: 40121409-40136789 on Build GRCh38
Transcripts
NM_001012241.1 XM_005257298.4 XM_005257299.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4443300 c.1150C>T p.Q384* Chr.17: 40133898-40133898 on Build GRCh38
4579656 c.1099C>T p.P367S Chr.17: 40133847-40133847 on Build GRCh38
5543135 c.1129C>T p.P377S Chr.17: 40133877-40133877 on Build GRCh38
5543134 c.1732C>T p.P578S Chr.17: 40133877-40133877 on Build GRCh38
5196885 c.1701C>T p.T567T Chr.17: 40133846-40133846 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs930082146 Chr.17: 40134019-40134019 on Build GRCh38
rs765964916 Chr.17: 40133806-40133806 on Build GRCh38
rs749617221 Chr.17: 40133908-40133908 on Build GRCh38
rs748650610 Chr.17: 40133918-40133918 on Build GRCh38
rs549926783 Chr.17: 40134020-40134020 on Build GRCh38
rs770581167 Chr.17: 40133924-40133924 on Build GRCh38
rs772629439 Chr.17: 40133921-40133921 on Build GRCh38
rs1038234012 Chr.17: 40134201-40134201 on Build GRCh38
rs1001001211 Chr.17: 40134153-40134153 on Build GRCh38
rs766983487 Chr.17: 40133803-40133803 on Build GRCh38
rs768987630 Chr.17: 40133909-40133909 on Build GRCh38
rs34201108 Chr.17: 40133888-40133888 on Build GRCh38
rs920604070 Chr.17: 40134217-40134217 on Build GRCh38
rs1045767935 Chr.17: 40134092-40134092 on Build GRCh38
rs912958371 Chr.17: 40134063-40134063 on Build GRCh38
rs970332011 Chr.17: 40133977-40133977 on Build GRCh38
rs369952395 Chr.17: 40133931-40133931 on Build GRCh38
rs942661149 Chr.17: 40134134-40134134 on Build GRCh38
rs899164342 Chr.17: 40133772-40133772 on Build GRCh38
rs760111354 Chr.17: 40133922-40133922 on Build GRCh38
rs368270175 Chr.17: 40133808-40133808 on Build GRCh38
rs986480354 Chr.17: 40133847-40133847 on Build GRCh38
rs753604986 Chr.17: 40133807-40133807 on Build GRCh38
rs772668925 Chr.17: 40133804-40133804 on Build GRCh38
rs954630702 Chr.17: 40133844-40133844 on Build GRCh38
rs988981126 Chr.17: 40134055-40134055 on Build GRCh38
rs17678817 Chr.17: 40133769-40133769 on Build GRCh38
rs943383821 Chr.17: 40133766-40133766 on Build GRCh38
rs1026368077 Chr.17: 40133832-40133832 on Build GRCh38
rs760366047 Chr.17: 40133805-40133805 on Build GRCh38
rs780141762 Chr.17: 40133907-40133907 on Build GRCh38
rs1032701281 Chr.17: 40134018-40134018 on Build GRCh38
rs753585016 Chr.17: 40133911-40133911 on Build GRCh38
rs762970474 Chr.17: 40133945-40133945 on Build GRCh38
rs986565535 Chr.17: 40134111-40134111 on Build GRCh38
rs1047035354 Chr.17: 40134028-40134028 on Build GRCh38
rs774862245 Chr.17: 40133801-40133801 on Build GRCh38
rs765214816 Chr.17: 40133839-40133839 on Build GRCh38
rs1037498385 Chr.17: 40133911-40133914 on Build GRCh38
rs765090079 Chr.17: 40133939-40133939 on Build GRCh38
rs769114201 Chr.17: 40133793-40133793 on Build GRCh38
rs957046665 Chr.17: 40134030-40134030 on Build GRCh38
rs377095062 Chr.17: 40133910-40133910 on Build GRCh38
rs955386564 Chr.17: 40134081-40134081 on Build GRCh38
rs563904895 Chr.17: 40133802-40133802 on Build GRCh38
rs376914100 Chr.17: 40133779-40133779 on Build GRCh38
rs866430268 Chr.17: 40134128-40134128 on Build GRCh38
rs201242174 Chr.17: 40133848-40133848 on Build GRCh38
rs896508868 Chr.17: 40134039-40134039 on Build GRCh38
rs752656422 Chr.17: 40133846-40133846 on Build GRCh38
rs775909472 Chr.17: 40133777-40133777 on Build GRCh38
rs757222416 Chr.17: 40133944-40133944 on Build GRCh38
rs752601576 Chr.17: 40133942-40133942 on Build GRCh38
rs571333662 Chr.17: 40134080-40134080 on Build GRCh38
rs980347679 Chr.17: 40133995-40133995 on Build GRCh38
rs775805285 Chr.17: 40133973-40133973 on Build GRCh38
rs756300093 Chr.17: 40133878-40133878 on Build GRCh38
rs759307442 Chr.17: 40133935-40133935 on Build GRCh38
rs750524872 Chr.17: 40133859-40133859 on Build GRCh38

View Full Product Details