Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 40558801-40559296 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCR7
Entrez Gene ID
1236
Gene Name
C-C motif chemokine receptor 7
Gene Aliases
BLR2, CC-CKR-7, CCR-7, CD197, CDw197, CMKBR7, EBI1
UniGene
Hs.370036
Gene Chromosome Location
Chr.17: 40553769-40565484 on Build GRCh38
Transcripts
NM_001301714.1 NM_001301716.1 NM_001301717.1 NM_001301718.1 NM_001838.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
978912 c.27C>T p.S9S Chr.17: 40558926-40558926 on Build GRCh38
7303121 c.60+9C>A p.? Chr.17: 40558884-40558884 on Build GRCh38
978911 c.35_36insT p.V13fs*15 Chr.17: 40558917-40558918 on Build GRCh38
3970026 c.50T>C p.V17A Chr.17: 40558903-40558903 on Build GRCh38
6623225 c.36G>T p.V12V Chr.17: 40558917-40558917 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750914678 Chr.17: 40558849-40558849 on Build GRCh38
rs771787906 Chr.17: 40559129-40559129 on Build GRCh38
rs747940899 Chr.17: 40558873-40558873 on Build GRCh38
rs557760418 Chr.17: 40558881-40558881 on Build GRCh38
rs567673364 Chr.17: 40559241-40559241 on Build GRCh38
rs112837758 Chr.17: 40559147-40559147 on Build GRCh38
rs561365689 Chr.17: 40559021-40559021 on Build GRCh38
rs777476348 Chr.17: 40558987-40558987 on Build GRCh38
rs1018793447 Chr.17: 40558884-40558884 on Build GRCh38
rs371359979 Chr.17: 40558981-40558981 on Build GRCh38
rs989837772 Chr.17: 40559149-40559149 on Build GRCh38
rs996657496 Chr.17: 40559016-40559016 on Build GRCh38
rs764917514 Chr.17: 40558967-40558967 on Build GRCh38
rs374537899 Chr.17: 40558907-40558907 on Build GRCh38
rs1042084720 Chr.17: 40559167-40559167 on Build GRCh38
rs946443403 Chr.17: 40559128-40559128 on Build GRCh38
rs376828366 Chr.17: 40558864-40558864 on Build GRCh38
rs975917370 Chr.17: 40558929-40558929 on Build GRCh38
rs768892070 Chr.17: 40558917-40558917 on Build GRCh38
rs3136689 Chr.17: 40558850-40558850 on Build GRCh38
rs1034421857 Chr.17: 40559088-40559088 on Build GRCh38
rs984119925 Chr.17: 40558992-40558992 on Build GRCh38
rs971472819 Chr.17: 40559083-40559083 on Build GRCh38
rs991036420 Chr.17: 40558869-40558869 on Build GRCh38
rs902143088 Chr.17: 40559266-40559266 on Build GRCh38
rs1055681207 Chr.17: 40559108-40559108 on Build GRCh38
rs764482222 Chr.17: 40558976-40558977 on Build GRCh38
rs923958590 Chr.17: 40558918-40558918 on Build GRCh38
rs958508766 Chr.17: 40559119-40559119 on Build GRCh38
rs370620921 Chr.17: 40558865-40558865 on Build GRCh38
rs2228015 Chr.17: 40558934-40558934 on Build GRCh38
rs774486933 Chr.17: 40558919-40558919 on Build GRCh38
rs113678754 Chr.17: 40559117-40559117 on Build GRCh38
rs767047433 Chr.17: 40558952-40558952 on Build GRCh38
rs778723333 Chr.17: 40559026-40559026 on Build GRCh38
rs762121218 Chr.17: 40558925-40558925 on Build GRCh38
rs76257997 Chr.17: 40559096-40559096 on Build GRCh38
rs368547056 Chr.17: 40558975-40558975 on Build GRCh38
rs111607645 Chr.17: 40558933-40558933 on Build GRCh38
rs938220061 Chr.17: 40559101-40559101 on Build GRCh38
rs960609761 Chr.17: 40559059-40559059 on Build GRCh38
rs778493479 Chr.17: 40558870-40558870 on Build GRCh38
rs553853532 Chr.17: 40559054-40559054 on Build GRCh38
rs749396477 Chr.17: 40558913-40558913 on Build GRCh38
rs770858393 Chr.17: 40558902-40558902 on Build GRCh38
rs193282140 Chr.17: 40558926-40558926 on Build GRCh38
rs754483254 Chr.17: 40558965-40558965 on Build GRCh38
rs773718925 Chr.17: 40558927-40558927 on Build GRCh38
rs563050018 Chr.17: 40559210-40559210 on Build GRCh38
rs1002581350 Chr.17: 40559086-40559086 on Build GRCh38
rs747608043 Chr.17: 40558993-40558993 on Build GRCh38
rs753391794 Chr.17: 40558963-40558963 on Build GRCh38
rs908516002 Chr.17: 40559013-40559013 on Build GRCh38
rs746907260 Chr.17: 40558890-40558890 on Build GRCh38
rs588019 Chr.17: 40558855-40558855 on Build GRCh38
rs928134245 Chr.17: 40559066-40559066 on Build GRCh38
rs1004822486 Chr.17: 40559064-40559064 on Build GRCh38
rs968161127 Chr.17: 40559175-40559175 on Build GRCh38
rs183729168 Chr.17: 40559029-40559029 on Build GRCh38
rs771811231 Chr.17: 40558875-40558875 on Build GRCh38
rs2228015 Chr.17: 40558934-40558934 on Build GRCh38

View Full Product Details