Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41381635-41382132 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT34
Entrez Gene ID
3885
Gene Name
keratin 34
Gene Aliases
HA4, Ha-4, KRTHA4, hHa4
UniGene
Hs.296942
Gene Chromosome Location
Chr.17: 41377650-41383816 on Build GRCh38
Transcripts
NM_021013.3 XM_011524793.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5548519 c.371C>T p.A124V Chr.17: 41382002-41382002 on Build GRCh38
5533537 c.475A>G p.I159V Chr.17: 41381795-41381795 on Build GRCh38
6497671 c.338T>A p.L113Q Chr.17: 41382035-41382035 on Build GRCh38
6697999 c.525T>A p.N175K Chr.17: 41381745-41381745 on Build GRCh38
5558580 c.421C>T p.L141L Chr.17: 41381952-41381952 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150445234 Chr.17: 41381971-41381971 on Build GRCh38
rs768417842 Chr.17: 41382107-41382107 on Build GRCh38
rs779566184 Chr.17: 41382102-41382102 on Build GRCh38
rs139387232 Chr.17: 41382050-41382050 on Build GRCh38
rs761506935 Chr.17: 41382055-41382055 on Build GRCh38
rs759729353 Chr.17: 41382067-41382067 on Build GRCh38
rs746122385 Chr.17: 41381678-41381678 on Build GRCh38
rs754012680 Chr.17: 41382003-41382003 on Build GRCh38
rs953702826 Chr.17: 41382096-41382096 on Build GRCh38
rs368667266 Chr.17: 41381930-41381930 on Build GRCh38
rs148769068 Chr.17: 41381691-41381691 on Build GRCh38
rs547092908 Chr.17: 41382089-41382089 on Build GRCh38
rs767289297 Chr.17: 41382002-41382002 on Build GRCh38
rs1007825614 Chr.17: 41381883-41381883 on Build GRCh38
rs756190282 Chr.17: 41381942-41381942 on Build GRCh38
rs770009258 Chr.17: 41381679-41381679 on Build GRCh38
rs906418605 Chr.17: 41381907-41381907 on Build GRCh38
rs201960842 Chr.17: 41381773-41381773 on Build GRCh38
rs892488665 Chr.17: 41381739-41381739 on Build GRCh38
rs756210024 Chr.17: 41381863-41381863 on Build GRCh38
rs752494449 Chr.17: 41381918-41381918 on Build GRCh38
rs766005733 Chr.17: 41381790-41381790 on Build GRCh38
rs762672987 Chr.17: 41381989-41381989 on Build GRCh38
rs769559689 Chr.17: 41381899-41381899 on Build GRCh38
rs374899064 Chr.17: 41381687-41381687 on Build GRCh38
rs140507109 Chr.17: 41381949-41381949 on Build GRCh38
rs772849368 Chr.17: 41381788-41381788 on Build GRCh38
rs200621003 Chr.17: 41381674-41381674 on Build GRCh38
rs779430132 Chr.17: 41381879-41381879 on Build GRCh38
rs758708346 Chr.17: 41382082-41382082 on Build GRCh38
rs752936495 Chr.17: 41382018-41382018 on Build GRCh38
rs752775083 Chr.17: 41382081-41382081 on Build GRCh38
rs763848349 Chr.17: 41381992-41381992 on Build GRCh38
rs745441964 Chr.17: 41382048-41382048 on Build GRCh38
rs112831734 Chr.17: 41381976-41381976 on Build GRCh38
rs202028743 Chr.17: 41381710-41381710 on Build GRCh38
rs775113601 Chr.17: 41381988-41381988 on Build GRCh38
rs760227102 Chr.17: 41381789-41381789 on Build GRCh38
rs770974651 Chr.17: 41381672-41381672 on Build GRCh38
rs769275757 Chr.17: 41382049-41382049 on Build GRCh38
rs755251502 Chr.17: 41381952-41381952 on Build GRCh38
rs753642654 Chr.17: 41381721-41381721 on Build GRCh38
rs144496585 Chr.17: 41381688-41381688 on Build GRCh38
rs1051664964 Chr.17: 41381780-41381780 on Build GRCh38
rs775041402 Chr.17: 41381775-41381775 on Build GRCh38
rs753505605 Chr.17: 41381794-41381794 on Build GRCh38
rs767544620 Chr.17: 41382059-41382059 on Build GRCh38
rs779269498 Chr.17: 41381965-41381965 on Build GRCh38
rs202220823 Chr.17: 41382054-41382054 on Build GRCh38
rs1026009939 Chr.17: 41381684-41381684 on Build GRCh38
rs781274428 Chr.17: 41381735-41381735 on Build GRCh38
rs976418612 Chr.17: 41381745-41381745 on Build GRCh38
rs749498137 Chr.17: 41381873-41381873 on Build GRCh38
rs762604171 Chr.17: 41381873-41381874 on Build GRCh38
rs79458943 Chr.17: 41381777-41381777 on Build GRCh38
rs768708651 Chr.17: 41381875-41381875 on Build GRCh38
rs759287985 Chr.17: 41381803-41381803 on Build GRCh38
rs748580306 Chr.17: 41381967-41381967 on Build GRCh38
rs543631425 Chr.17: 41381709-41381709 on Build GRCh38
rs532669839 Chr.17: 41381975-41381975 on Build GRCh38
rs370145628 Chr.17: 41381689-41381689 on Build GRCh38
rs61740672 Chr.17: 41382028-41382028 on Build GRCh38
rs1026068051 Chr.17: 41381941-41381941 on Build GRCh38
rs866168029 Chr.17: 41381770-41381770 on Build GRCh38
rs866260015 Chr.17: 41381792-41381792 on Build GRCh38
rs773148078 Chr.17: 41381683-41381683 on Build GRCh38
rs757550438 Chr.17: 41382087-41382087 on Build GRCh38
rs751022055 Chr.17: 41381693-41381693 on Build GRCh38
rs911824010 Chr.17: 41382032-41382032 on Build GRCh38
rs745309735 Chr.17: 41381979-41381979 on Build GRCh38
rs775780633 Chr.17: 41381752-41381752 on Build GRCh38
rs748453377 Chr.17: 41381880-41381880 on Build GRCh38
rs747148046 Chr.17: 41381666-41381666 on Build GRCh38
rs778220460 Chr.17: 41382025-41382025 on Build GRCh38
rs764813910 Chr.17: 41381916-41381916 on Build GRCh38
rs757789836 Chr.17: 41382034-41382034 on Build GRCh38
rs780191013 Chr.17: 41381867-41381867 on Build GRCh38
rs761821120 Chr.17: 41382001-41382001 on Build GRCh38
rs780216860 Chr.17: 41381946-41381946 on Build GRCh38
rs777621565 Chr.17: 41381859-41381859 on Build GRCh38
rs773841508 Chr.17: 41381888-41381888 on Build GRCh38
rs778414119 Chr.17: 41382111-41382111 on Build GRCh38
rs534771994 Chr.17: 41382070-41382070 on Build GRCh38
rs752030183 Chr.17: 41382085-41382085 on Build GRCh38
rs181152719 Chr.17: 41381694-41381694 on Build GRCh38
rs781749096 Chr.17: 41382090-41382090 on Build GRCh38
rs755092027 Chr.17: 41382004-41382004 on Build GRCh38
rs369259644 Chr.17: 41382060-41382060 on Build GRCh38
rs886149901 Chr.17: 41382110-41382110 on Build GRCh38
rs781546162 Chr.17: 41382045-41382045 on Build GRCh38
rs186118320 Chr.17: 41381887-41381887 on Build GRCh38
rs3744390 Chr.17: 41381932-41381932 on Build GRCh38
rs199776619 Chr.17: 41381727-41381727 on Build GRCh38
rs939828173 Chr.17: 41381874-41381874 on Build GRCh38
rs199859296 Chr.17: 41381707-41381707 on Build GRCh38
rs967299714 Chr.17: 41381977-41381977 on Build GRCh38
rs190446535 Chr.17: 41382097-41382097 on Build GRCh38
rs748973716 Chr.17: 41382051-41382051 on Build GRCh38
rs769504488 Chr.17: 41381981-41381981 on Build GRCh38
rs750572236 Chr.17: 41381862-41381862 on Build GRCh38
rs765605567 Chr.17: 41382012-41382012 on Build GRCh38
rs747581274 Chr.17: 41381973-41381973 on Build GRCh38
rs767163858 Chr.17: 41381699-41381699 on Build GRCh38
rs753799735 Chr.17: 41381947-41381947 on Build GRCh38
rs144045423 Chr.17: 41382024-41382024 on Build GRCh38
rs773980852 Chr.17: 41381997-41381997 on Build GRCh38
rs148317991 Chr.17: 41381933-41381933 on Build GRCh38
rs111441823 Chr.17: 41382103-41382103 on Build GRCh38
rs756577185 Chr.17: 41382100-41382100 on Build GRCh38
rs375355411 Chr.17: 41381978-41381978 on Build GRCh38
rs16966754 Chr.17: 41381660-41381660 on Build GRCh38
rs749676550 Chr.17: 41381755-41381755 on Build GRCh38
rs759256671 Chr.17: 41381911-41381911 on Build GRCh38
rs112408316 Chr.17: 41381813-41381813 on Build GRCh38
rs35040340 Chr.17: 41381667-41381667 on Build GRCh38
rs752443261 Chr.17: 41381812-41381812 on Build GRCh38
rs770442215 Chr.17: 41381891-41381891 on Build GRCh38
rs372070920 Chr.17: 41381726-41381726 on Build GRCh38
rs768075825 Chr.17: 41381781-41381781 on Build GRCh38
rs765049721 Chr.17: 41381810-41381810 on Build GRCh38
rs769913513 Chr.17: 41381748-41381748 on Build GRCh38
rs760152250 Chr.17: 41381890-41381890 on Build GRCh38
rs1017411589 Chr.17: 41381885-41381885 on Build GRCh38
rs375061597 Chr.17: 41381852-41381852 on Build GRCh38
rs758293346 Chr.17: 41381731-41381731 on Build GRCh38
rs758346381 Chr.17: 41381823-41381823 on Build GRCh38
rs895141786 Chr.17: 41381956-41381956 on Build GRCh38

View Full Product Details