Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41437277-41437825 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT38
Entrez Gene ID
8687
Gene Name
keratin 38
Gene Aliases
HA8, KRTHA8, hHa8
UniGene
Hs.248188
Gene Chromosome Location
Chr.17: 41436369-41441344 on Build GRCh38
Transcripts
NM_006771.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6572454 c.1283C>T p.T428I Chr.17: 41437500-41437500 on Build GRCh38
7303261 c.1364G>A p.R455Q Chr.17: 41437419-41437419 on Build GRCh38
7374382 c.1303C>T p.P435S Chr.17: 41437480-41437480 on Build GRCh38
6281546 c.1356C>T p.T452T Chr.17: 41437427-41437427 on Build GRCh38
5386333 c.1335C>T p.P445P Chr.17: 41437448-41437448 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs56120541 Chr.17: 41437470-41437470 on Build GRCh38
rs769280282 Chr.17: 41437530-41437530 on Build GRCh38
rs373418175 Chr.17: 41437422-41437422 on Build GRCh38
rs1000143898 Chr.17: 41437632-41437632 on Build GRCh38
rs748434953 Chr.17: 41437742-41437742 on Build GRCh38
rs1055459189 Chr.17: 41437569-41437569 on Build GRCh38
rs141275633 Chr.17: 41437734-41437734 on Build GRCh38
rs9894044 Chr.17: 41437368-41437368 on Build GRCh38
rs201870112 Chr.17: 41437466-41437466 on Build GRCh38
rs866736039 Chr.17: 41437557-41437557 on Build GRCh38
rs762648893 Chr.17: 41437536-41437536 on Build GRCh38
rs557586825 Chr.17: 41437600-41437600 on Build GRCh38
rs375989251 Chr.17: 41437379-41437379 on Build GRCh38
rs747582005 Chr.17: 41437523-41437523 on Build GRCh38
rs985134789 Chr.17: 41437467-41437467 on Build GRCh38
rs1036581095 Chr.17: 41437337-41437337 on Build GRCh38
rs780324275 Chr.17: 41437504-41437504 on Build GRCh38
rs184262664 Chr.17: 41437604-41437604 on Build GRCh38
rs529308915 Chr.17: 41437419-41437419 on Build GRCh38
rs940181668 Chr.17: 41437601-41437601 on Build GRCh38
rs146369884 Chr.17: 41437518-41437518 on Build GRCh38
rs775016296 Chr.17: 41437535-41437535 on Build GRCh38
rs188375863 Chr.17: 41437608-41437608 on Build GRCh38
rs921961536 Chr.17: 41437432-41437432 on Build GRCh38
rs897416 Chr.17: 41437516-41437516 on Build GRCh38
rs751600455 Chr.17: 41437486-41437486 on Build GRCh38
rs765120123 Chr.17: 41437411-41437411 on Build GRCh38
rs777074027 Chr.17: 41437526-41437526 on Build GRCh38
rs753502725 Chr.17: 41437400-41437400 on Build GRCh38
rs775857390 Chr.17: 41437375-41437375 on Build GRCh38
rs764819105 Chr.17: 41437473-41437473 on Build GRCh38
rs2071614 Chr.17: 41437770-41437770 on Build GRCh38
rs984821454 Chr.17: 41437763-41437763 on Build GRCh38
rs773714417 Chr.17: 41437457-41437457 on Build GRCh38
rs938469045 Chr.17: 41437529-41437529 on Build GRCh38
rs201031533 Chr.17: 41437538-41437538 on Build GRCh38
rs953418685 Chr.17: 41437760-41437760 on Build GRCh38
rs201381536 Chr.17: 41437420-41437420 on Build GRCh38
rs141861762 Chr.17: 41437427-41437427 on Build GRCh38
rs546085284 Chr.17: 41437325-41437325 on Build GRCh38
rs530701592 Chr.17: 41437431-41437431 on Build GRCh38
rs768733190 Chr.17: 41437438-41437438 on Build GRCh38
rs772445542 Chr.17: 41437451-41437451 on Build GRCh38
rs776403446 Chr.17: 41437448-41437467 on Build GRCh38
rs757253265 Chr.17: 41437493-41437493 on Build GRCh38
rs771404275 Chr.17: 41437524-41437524 on Build GRCh38
rs557804376 Chr.17: 41437350-41437350 on Build GRCh38
rs117706286 Chr.17: 41437386-41437386 on Build GRCh38
rs192615532 Chr.17: 41437644-41437644 on Build GRCh38
rs897415 Chr.17: 41437620-41437620 on Build GRCh38
rs779249140 Chr.17: 41437633-41437633 on Build GRCh38
rs754575104 Chr.17: 41437406-41437406 on Build GRCh38
rs140093590 Chr.17: 41437482-41437482 on Build GRCh38
rs771513194 Chr.17: 41437723-41437723 on Build GRCh38
rs765931907 Chr.17: 41437463-41437463 on Build GRCh38
rs372908459 Chr.17: 41437393-41437393 on Build GRCh38
rs777882647 Chr.17: 41437424-41437424 on Build GRCh38
rs756273135 Chr.17: 41437429-41437429 on Build GRCh38
rs748485967 Chr.17: 41437450-41437450 on Build GRCh38
rs749460012 Chr.17: 41437377-41437377 on Build GRCh38
rs751364962 Chr.17: 41437563-41437563 on Build GRCh38
rs573131570 Chr.17: 41437636-41437636 on Build GRCh38
rs573929481 Chr.17: 41437745-41437745 on Build GRCh38
rs774999607 Chr.17: 41437357-41437357 on Build GRCh38
rs984333448 Chr.17: 41437499-41437499 on Build GRCh38
rs552415442 Chr.17: 41437437-41437437 on Build GRCh38
rs749309611 Chr.17: 41437505-41437505 on Build GRCh38
rs781381424 Chr.17: 41437361-41437361 on Build GRCh38
rs976708652 Chr.17: 41437455-41437455 on Build GRCh38
rs966191312 Chr.17: 41437708-41437708 on Build GRCh38
rs1021683958 Chr.17: 41437610-41437610 on Build GRCh38
rs371518902 Chr.17: 41437527-41437527 on Build GRCh38
rs146550898 Chr.17: 41437380-41437380 on Build GRCh38
rs377077955 Chr.17: 41437517-41437517 on Build GRCh38
rs544249300 Chr.17: 41437575-41437575 on Build GRCh38
rs1002539244 Chr.17: 41437356-41437356 on Build GRCh38
rs1011261986 Chr.17: 41437609-41437609 on Build GRCh38
rs761115467 Chr.17: 41437462-41437462 on Build GRCh38
rs763573091 Chr.17: 41437427-41437429 on Build GRCh38
rs940516744 Chr.17: 41437326-41437326 on Build GRCh38
rs778098533 Chr.17: 41437520-41437520 on Build GRCh38
rs369193583 Chr.17: 41437388-41437388 on Build GRCh38
rs1048963444 Chr.17: 41437305-41437305 on Build GRCh38
rs772338762 Chr.17: 41437519-41437519 on Build GRCh38
rs148086126 Chr.17: 41437426-41437426 on Build GRCh38
rs763921085 Chr.17: 41437483-41437483 on Build GRCh38
rs774832023 Chr.17: 41437446-41437446 on Build GRCh38

View Full Product Details