Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41502236-41502771 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT13
Entrez Gene ID
3860
Gene Name
keratin 13
Gene Aliases
CK13, K13, WSN2
UniGene
Hs.654550
Gene Chromosome Location
Chr.17: 41500981-41505613 on Build GRCh38
Transcripts
NM_002274.3 NM_153490.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
107392 c.1023G>A p.M341I Chr.17: 41502687-41502687 on Build GRCh38
6697827 c.970C>G p.L324V Chr.17: 41502740-41502740 on Build GRCh38
706376 c.977G>A p.R326H Chr.17: 41502733-41502733 on Build GRCh38
2840855 c.1133G>A p.R378H Chr.17: 41502485-41502485 on Build GRCh38
6989103 c.1017G>A p.L339L Chr.17: 41502693-41502693 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1051122025 Chr.17: 41502309-41502309 on Build GRCh38
rs773710186 Chr.17: 41502627-41502627 on Build GRCh38
rs145167376 Chr.17: 41502686-41502686 on Build GRCh38
rs779293187 Chr.17: 41502617-41502617 on Build GRCh38
rs1036893212 Chr.17: 41502275-41502275 on Build GRCh38
rs751003803 Chr.17: 41502358-41502358 on Build GRCh38
rs377025441 Chr.17: 41502745-41502745 on Build GRCh38
rs1005644386 Chr.17: 41502343-41502343 on Build GRCh38
rs767038726 Chr.17: 41502498-41502498 on Build GRCh38
rs769874748 Chr.17: 41502478-41502478 on Build GRCh38
rs747067918 Chr.17: 41502388-41502388 on Build GRCh38
rs368051143 Chr.17: 41502352-41502352 on Build GRCh38
rs369660028 Chr.17: 41502659-41502659 on Build GRCh38
rs752701638 Chr.17: 41502534-41502534 on Build GRCh38
rs570689594 Chr.17: 41502378-41502378 on Build GRCh38
rs749916295 Chr.17: 41502499-41502499 on Build GRCh38
rs148296285 Chr.17: 41502554-41502554 on Build GRCh38
rs560509278 Chr.17: 41502723-41502723 on Build GRCh38
rs200309164 Chr.17: 41502485-41502485 on Build GRCh38
rs779527087 Chr.17: 41502372-41502373 on Build GRCh38
rs777839874 Chr.17: 41502599-41502599 on Build GRCh38
rs556542745 Chr.17: 41502605-41502605 on Build GRCh38
rs758084444 Chr.17: 41502367-41502371 on Build GRCh38
rs778562268 Chr.17: 41502357-41502359 on Build GRCh38
rs996483538 Chr.17: 41502287-41502288 on Build GRCh38
rs760111749 Chr.17: 41502568-41502568 on Build GRCh38
rs531211732 Chr.17: 41502724-41502724 on Build GRCh38
rs267604870 Chr.17: 41502747-41502747 on Build GRCh38
rs765844638 Chr.17: 41502571-41502571 on Build GRCh38
rs754996481 Chr.17: 41502432-41502432 on Build GRCh38
rs748028479 Chr.17: 41502462-41502462 on Build GRCh38
rs745938353 Chr.17: 41502548-41502548 on Build GRCh38
rs755226206 Chr.17: 41502677-41502677 on Build GRCh38
rs748580603 Chr.17: 41502380-41502380 on Build GRCh38
rs758476417 Chr.17: 41502598-41502598 on Build GRCh38
rs756153506 Chr.17: 41502604-41502604 on Build GRCh38
rs751138953 Chr.17: 41502412-41502412 on Build GRCh38
rs771164488 Chr.17: 41502471-41502471 on Build GRCh38
rs749496713 Chr.17: 41502606-41502606 on Build GRCh38
rs972267166 Chr.17: 41502735-41502735 on Build GRCh38
rs770113117 Chr.17: 41502549-41502549 on Build GRCh38
rs750110704 Chr.17: 41502426-41502426 on Build GRCh38
rs894459277 Chr.17: 41502310-41502310 on Build GRCh38
rs762358997 Chr.17: 41502562-41502562 on Build GRCh38
rs1034328946 Chr.17: 41502305-41502305 on Build GRCh38
rs762855075 Chr.17: 41502664-41502664 on Build GRCh38
rs746141279 Chr.17: 41502477-41502477 on Build GRCh38
rs776182292 Chr.17: 41502643-41502643 on Build GRCh38
rs200269035 Chr.17: 41502380-41502380 on Build GRCh38
rs748299083 Chr.17: 41502683-41502683 on Build GRCh38
rs369742463 Chr.17: 41502428-41502428 on Build GRCh38
rs983806783 Chr.17: 41502706-41502706 on Build GRCh38
rs375900245 Chr.17: 41502535-41502535 on Build GRCh38
rs764975817 Chr.17: 41502652-41502652 on Build GRCh38
rs1014546775 Chr.17: 41502410-41502410 on Build GRCh38
rs1011567467 Chr.17: 41502645-41502645 on Build GRCh38
rs138885186 Chr.17: 41502402-41502402 on Build GRCh38
rs192772396 Chr.17: 41502603-41502603 on Build GRCh38
rs201273553 Chr.17: 41502493-41502493 on Build GRCh38
rs748511951 Chr.17: 41502625-41502625 on Build GRCh38
rs756886209 Chr.17: 41502349-41502352 on Build GRCh38
rs767962920 Chr.17: 41502393-41502393 on Build GRCh38
rs780811887 Chr.17: 41502365-41502365 on Build GRCh38
rs771074447 Chr.17: 41502390-41502390 on Build GRCh38
rs766421222 Chr.17: 41502668-41502668 on Build GRCh38
rs139318123 Chr.17: 41502701-41502701 on Build GRCh38
rs753468933 Chr.17: 41502519-41502519 on Build GRCh38
rs372223120 Chr.17: 41502436-41502436 on Build GRCh38
rs577900764 Chr.17: 41502637-41502637 on Build GRCh38
rs1028069325 Chr.17: 41502690-41502690 on Build GRCh38
rs749461317 Chr.17: 41502552-41502552 on Build GRCh38
rs765717279 Chr.17: 41502399-41502399 on Build GRCh38
rs545219692 Chr.17: 41502658-41502658 on Build GRCh38
rs749009134 Chr.17: 41502429-41502429 on Build GRCh38
rs149077503 Chr.17: 41502391-41502391 on Build GRCh38
rs771435962 Chr.17: 41502719-41502719 on Build GRCh38
rs141432690 Chr.17: 41502563-41502563 on Build GRCh38
rs961139675 Chr.17: 41502364-41502364 on Build GRCh38
rs1002881454 Chr.17: 41502302-41502302 on Build GRCh38
rs761201809 Chr.17: 41502628-41502628 on Build GRCh38
rs199762312 Chr.17: 41502590-41502590 on Build GRCh38
rs757172151 Chr.17: 41502667-41502667 on Build GRCh38
rs768904310 Chr.17: 41502616-41502616 on Build GRCh38
rs147022064 Chr.17: 41502508-41502508 on Build GRCh38
rs150845230 Chr.17: 41502589-41502589 on Build GRCh38
rs371531703 Chr.17: 41502279-41502279 on Build GRCh38
rs768459234 Chr.17: 41502382-41502382 on Build GRCh38
rs758386353 Chr.17: 41502537-41502537 on Build GRCh38
rs554915056 Chr.17: 41502526-41502526 on Build GRCh38
rs762655823 Chr.17: 41502733-41502733 on Build GRCh38
rs923312445 Chr.17: 41502523-41502523 on Build GRCh38
rs149134550 Chr.17: 41502687-41502687 on Build GRCh38
rs764004897 Chr.17: 41502734-41502734 on Build GRCh38
rs754732492 Chr.17: 41502525-41502525 on Build GRCh38
rs780154003 Chr.17: 41502551-41502551 on Build GRCh38
rs548989486 Chr.17: 41502328-41502328 on Build GRCh38
rs775287167 Chr.17: 41502730-41502730 on Build GRCh38
rs141462482 Chr.17: 41502715-41502715 on Build GRCh38
rs368888147 Chr.17: 41502564-41502564 on Build GRCh38
rs930982219 Chr.17: 41502531-41502531 on Build GRCh38
rs142895125 Chr.17: 41502476-41502476 on Build GRCh38
rs369135048 Chr.17: 41502327-41502327 on Build GRCh38
rs753462261 Chr.17: 41502573-41502573 on Build GRCh38
rs902007287 Chr.17: 41502262-41502262 on Build GRCh38
rs935439174 Chr.17: 41502456-41502456 on Build GRCh38
rs201356802 Chr.17: 41502492-41502492 on Build GRCh38
rs928832284 Chr.17: 41502737-41502737 on Build GRCh38
rs755910617 Chr.17: 41502370-41502370 on Build GRCh38
rs115695615 Chr.17: 41502347-41502347 on Build GRCh38
rs766948707 Chr.17: 41502422-41502422 on Build GRCh38
rs373085505 Chr.17: 41502665-41502665 on Build GRCh38
rs763708756 Chr.17: 41502357-41502357 on Build GRCh38
rs769371388 Chr.17: 41502727-41502727 on Build GRCh38
rs761699251 Chr.17: 41502744-41502744 on Build GRCh38
rs770118226 Chr.17: 41502663-41502663 on Build GRCh38
rs538303362 Chr.17: 41502486-41502486 on Build GRCh38
rs145983286 Chr.17: 41502401-41502401 on Build GRCh38
rs749838803 Chr.17: 41502361-41502362 on Build GRCh38
rs779098221 Chr.17: 41502681-41502681 on Build GRCh38
rs188713834 Chr.17: 41502288-41502288 on Build GRCh38
rs753925971 Chr.17: 41502674-41502674 on Build GRCh38
rs781460720 Chr.17: 41502545-41502545 on Build GRCh38
rs759178223 Chr.17: 41502575-41502575 on Build GRCh38
rs759150262 Chr.17: 41502648-41502648 on Build GRCh38
rs371850724 Chr.17: 41502453-41502453 on Build GRCh38
rs756970882 Chr.17: 41502362-41502362 on Build GRCh38
rs762270993 Chr.17: 41502496-41502496 on Build GRCh38
rs552137773 Chr.17: 41502379-41502379 on Build GRCh38
rs1021997832 Chr.17: 41502653-41502653 on Build GRCh38
rs773405109 Chr.17: 41502386-41502386 on Build GRCh38
rs758902395 Chr.17: 41502555-41502555 on Build GRCh38

View Full Product Details