Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 59321563-59322048 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGGY
Entrez Gene ID
55277
Gene Name
FGGY carbohydrate kinase domain containing
Gene Aliases
-
UniGene
Hs.444301
Gene Chromosome Location
Chr.1: 59296405-59762730 on Build GRCh38
Transcripts
NM_001113411.1 NM_001244714.1 NM_018291.3 NR_103473.1 XM_011541730.1 XM_011541731.1 XM_011541732.1 XM_011541733.1 XM_011541735.1 XM_011541736.2 XM_017001643.1 XM_017001644.1 XM_017001645.1 XM_017001646.1 XM_017001647.1 XM_017001648.1 XM_017001649.1 XM_017001650.1 XM_017001651.1 XM_017001652.1 XM_017001653.1 XM_017001654.1 XM_017001655.1 XM_017001656.1 XM_017001658.1 XM_017001659.1 XM_017001660.1 XM_017001661.1 XM_017001663.1 XM_017001664.1 XM_017001665.1 XM_017001666.1 XM_017001667.1 XM_017001668.1 XM_017001669.1 XM_017001671.1 XM_017001672.1 XM_017001673.1 XM_017001674.1 XM_017001675.1 XM_017001676.1 XM_017001679.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4009121 c.67C>T p.R23C Chr.1: 59321616-59321616 on Build GRCh38
7201596 c.62G>T p.S21I Chr.1: 59321611-59321611 on Build GRCh38
1602583 c.54A>G p.G18G Chr.1: 59321603-59321603 on Build GRCh38
5847085 c.49G>A p.V17I Chr.1: 59321598-59321598 on Build GRCh38
6063592 c.17A>T p.Q6L Chr.1: 59321566-59321566 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1010317388 Chr.1: 59321693-59321693 on Build GRCh38
rs766518834 Chr.1: 59321617-59321617 on Build GRCh38
rs1054472247 Chr.1: 59321670-59321670 on Build GRCh38
rs773950262 Chr.1: 59321609-59321609 on Build GRCh38
rs140149476 Chr.1: 59321757-59321757 on Build GRCh38
rs532734870 Chr.1: 59321829-59321829 on Build GRCh38
rs200490773 Chr.1: 59321766-59321766 on Build GRCh38
rs369669169 Chr.1: 59321719-59321719 on Build GRCh38
rs368626109 Chr.1: 59321590-59321590 on Build GRCh38
rs750581053 Chr.1: 59321614-59321614 on Build GRCh38
rs201564247 Chr.1: 59321742-59321742 on Build GRCh38
rs835409 Chr.1: 59321678-59321678 on Build GRCh38
rs199980192 Chr.1: 59321598-59321598 on Build GRCh38
rs143791102 Chr.1: 59321804-59321804 on Build GRCh38
rs534054341 Chr.1: 59321942-59321942 on Build GRCh38
rs916442678 Chr.1: 59321720-59321720 on Build GRCh38
rs946038461 Chr.1: 59321982-59321982 on Build GRCh38
rs748071727 Chr.1: 59321771-59321771 on Build GRCh38
rs918573355 Chr.1: 59321692-59321692 on Build GRCh38
rs182624572 Chr.1: 59321644-59321644 on Build GRCh38
rs990805120 Chr.1: 59321925-59321925 on Build GRCh38
rs376917600 Chr.1: 59321801-59321801 on Build GRCh38
rs548488191 Chr.1: 59321878-59321878 on Build GRCh38
rs200358653 Chr.1: 59321707-59321707 on Build GRCh38
rs916516066 Chr.1: 59321981-59321981 on Build GRCh38
rs538580665 Chr.1: 59321846-59321846 on Build GRCh38
rs563762898 Chr.1: 59321823-59321823 on Build GRCh38
rs928200231 Chr.1: 59321792-59321792 on Build GRCh38
rs747251682 Chr.1: 59321785-59321785 on Build GRCh38
rs1051118518 Chr.1: 59321699-59321699 on Build GRCh38
rs759506495 Chr.1: 59321796-59321796 on Build GRCh38
rs148995992 Chr.1: 59321964-59321964 on Build GRCh38
rs369929167 Chr.1: 59321714-59321714 on Build GRCh38
rs146690473 Chr.1: 59321737-59321737 on Build GRCh38
rs190529789 Chr.1: 59321805-59321805 on Build GRCh38
rs773705363 Chr.1: 59321715-59321715 on Build GRCh38
rs755480158 Chr.1: 59321633-59321633 on Build GRCh38
rs764515786 Chr.1: 59321862-59321862 on Build GRCh38
rs773592418 Chr.1: 59321710-59321710 on Build GRCh38
rs773076423 Chr.1: 59321783-59321783 on Build GRCh38
rs187579685 Chr.1: 59321728-59321728 on Build GRCh38
rs771268862 Chr.1: 59321791-59321791 on Build GRCh38
rs771796919 Chr.1: 59321773-59321773 on Build GRCh38
rs184067447 Chr.1: 59321822-59321822 on Build GRCh38
rs749228110 Chr.1: 59321682-59321682 on Build GRCh38
rs778683737 Chr.1: 59321767-59321767 on Build GRCh38
rs767618630 Chr.1: 59321610-59321610 on Build GRCh38
rs139993207 Chr.1: 59322001-59322001 on Build GRCh38
rs753148274 Chr.1: 59321731-59321731 on Build GRCh38
rs532672011 Chr.1: 59321597-59321597 on Build GRCh38
rs779552113 Chr.1: 59321751-59321751 on Build GRCh38
rs761528063 Chr.1: 59321614-59321614 on Build GRCh38
rs779353611 Chr.1: 59321634-59321634 on Build GRCh38
rs568600959 Chr.1: 59321937-59321937 on Build GRCh38
rs890019703 Chr.1: 59321807-59321807 on Build GRCh38
rs927588351 Chr.1: 59321642-59321642 on Build GRCh38
rs570891961 Chr.1: 59321993-59321993 on Build GRCh38
rs369790341 Chr.1: 59321741-59321741 on Build GRCh38
rs531980422 Chr.1: 59321877-59321877 on Build GRCh38
rs780674255 Chr.1: 59321662-59321662 on Build GRCh38
rs375711757 Chr.1: 59321631-59321631 on Build GRCh38
rs201589585 Chr.1: 59321646-59321646 on Build GRCh38
rs963080541 Chr.1: 59321669-59321669 on Build GRCh38
rs146878633 Chr.1: 59321833-59321833 on Build GRCh38
rs960979844 Chr.1: 59321896-59321896 on Build GRCh38
rs768420286 Chr.1: 59321697-59321697 on Build GRCh38
rs1017082415 Chr.1: 59321830-59321830 on Build GRCh38
rs769312503 Chr.1: 59321667-59321667 on Build GRCh38
rs779678533 Chr.1: 59321605-59321605 on Build GRCh38
rs776919965 Chr.1: 59321793-59321793 on Build GRCh38
rs761507232 Chr.1: 59321701-59321701 on Build GRCh38
rs199810745 Chr.1: 59321732-59321732 on Build GRCh38
rs760816361 Chr.1: 59321616-59321616 on Build GRCh38
rs760151330 Chr.1: 59321729-59321729 on Build GRCh38

View Full Product Details