Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 62212281-62212819 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
L1TD1
Entrez Gene ID
54596
Gene Name
LINE1 type transposase domain containing 1
Gene Aliases
ECAT11
UniGene
Hs.685462
Gene Chromosome Location
Chr.1: 62194802-62212328 on Build GRCh38
Transcripts
NM_001164835.1 NM_019079.4

SNP Detail

SNP ID Chromosome Location
rs909244457 Chr.1: 62212626-62212628 on Build GRCh38
rs997693662 Chr.1: 62212781-62212781 on Build GRCh38
rs41302758 Chr.1: 62212337-62212337 on Build GRCh38
rs1019936744 Chr.1: 62212361-62212361 on Build GRCh38
rs986991901 Chr.1: 62212372-62212372 on Build GRCh38
rs1046268432 Chr.1: 62212471-62212471 on Build GRCh38
rs769401911 Chr.1: 62212517-62212517 on Build GRCh38
rs981469911 Chr.1: 62212620-62212620 on Build GRCh38
rs543861362 Chr.1: 62212630-62212630 on Build GRCh38
rs966998876 Chr.1: 62212364-62212364 on Build GRCh38
rs937384041 Chr.1: 62212489-62212489 on Build GRCh38
rs962574956 Chr.1: 62212618-62212618 on Build GRCh38
rs909307437 Chr.1: 62212362-62212363 on Build GRCh38
rs183333677 Chr.1: 62212609-62212609 on Build GRCh38
rs892957167 Chr.1: 62212507-62212507 on Build GRCh38
rs561252937 Chr.1: 62212607-62212607 on Build GRCh38
rs138694434 Chr.1: 62212388-62212388 on Build GRCh38
rs968585751 Chr.1: 62212315-62212315 on Build GRCh38
rs775259518 Chr.1: 62212676-62212676 on Build GRCh38
rs140032598 Chr.1: 62212448-62212448 on Build GRCh38
rs950716263 Chr.1: 62212354-62212354 on Build GRCh38
rs903110851 Chr.1: 62212730-62212730 on Build GRCh38
rs966215719 Chr.1: 62212563-62212563 on Build GRCh38
rs572442416 Chr.1: 62212585-62212585 on Build GRCh38
rs765454543 Chr.1: 62212690-62212692 on Build GRCh38
rs907070678 Chr.1: 62212338-62212338 on Build GRCh38
rs1001368633 Chr.1: 62212343-62212343 on Build GRCh38
rs7367812 Chr.1: 62212500-62212500 on Build GRCh38
rs773068445 Chr.1: 62212541-62212541 on Build GRCh38
rs12760006 Chr.1: 62212577-62212577 on Build GRCh38
rs75356199 Chr.1: 62212352-62212352 on Build GRCh38
rs907901631 Chr.1: 62212438-62212438 on Build GRCh38
rs1021375884 Chr.1: 62212304-62212304 on Build GRCh38
rs1019656801 Chr.1: 62212550-62212550 on Build GRCh38
rs41306641 Chr.1: 62212360-62212360 on Build GRCh38
rs71751969 Chr.1: 62212780-62212780 on Build GRCh38
rs551278183 Chr.1: 62212666-62212666 on Build GRCh38
rs940670198 Chr.1: 62212452-62212452 on Build GRCh38
rs147128295 Chr.1: 62212350-62212350 on Build GRCh38
rs369404343 Chr.1: 62212779-62212780 on Build GRCh38
rs907293800 Chr.1: 62212485-62212485 on Build GRCh38
rs573063453 Chr.1: 62212331-62212331 on Build GRCh38
rs1007073470 Chr.1: 62212524-62212524 on Build GRCh38
rs71053307 Chr.1: 62212792-62212793 on Build GRCh38
rs1041495361 Chr.1: 62212725-62212725 on Build GRCh38
rs1011470708 Chr.1: 62212515-62212515 on Build GRCh38
rs190134833 Chr.1: 62212592-62212592 on Build GRCh38
rs762756600 Chr.1: 62212418-62212418 on Build GRCh38
rs1040837119 Chr.1: 62212768-62212768 on Build GRCh38
rs142004069 Chr.1: 62212417-62212417 on Build GRCh38
rs1040731038 Chr.1: 62212791-62212791 on Build GRCh38
rs1008537333 Chr.1: 62212745-62212745 on Build GRCh38
rs538813731 Chr.1: 62212555-62212555 on Build GRCh38
rs7367828 Chr.1: 62212716-62212716 on Build GRCh38
rs560342482 Chr.1: 62212682-62212682 on Build GRCh38
rs774131648 Chr.1: 62212590-62212590 on Build GRCh38
rs72921870 Chr.1: 62212641-62212641 on Build GRCh38
rs567043022 Chr.1: 62212793-62212793 on Build GRCh38
rs368358176 Chr.1: 62212394-62212394 on Build GRCh38
rs562595835 Chr.1: 62212640-62212640 on Build GRCh38
rs146819336 Chr.1: 62212764-62212765 on Build GRCh38
rs947328759 Chr.1: 62212697-62212697 on Build GRCh38
rs772965906 Chr.1: 62212398-62212398 on Build GRCh38
rs938521149 Chr.1: 62212649-62212649 on Build GRCh38
rs868166252 Chr.1: 62212333-62212333 on Build GRCh38
rs913140546 Chr.1: 62212692-62212692 on Build GRCh38
rs985031016 Chr.1: 62212430-62212430 on Build GRCh38
rs942437820 Chr.1: 62212767-62212767 on Build GRCh38
rs548632682 Chr.1: 62212780-62212780 on Build GRCh38
rs975541034 Chr.1: 62212580-62212580 on Build GRCh38
rs576473922 Chr.1: 62212627-62212627 on Build GRCh38
rs34759928 Chr.1: 62212334-62212334 on Build GRCh38
rs901974750 Chr.1: 62212775-62212775 on Build GRCh38
rs902888494 Chr.1: 62212534-62212534 on Build GRCh38
rs1019349696 Chr.1: 62212377-62212377 on Build GRCh38
rs1007716289 Chr.1: 62212322-62212322 on Build GRCh38
rs760901505 Chr.1: 62212556-62212556 on Build GRCh38
rs779587170 Chr.1: 62212646-62212648 on Build GRCh38
rs1007899943 Chr.1: 62212548-62212548 on Build GRCh38

View Full Product Details