Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 64210607-64211154 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBE2U
Entrez Gene ID
148581
Gene Name
ubiquitin conjugating enzyme E2 U (putative)
Gene Aliases
-
UniGene
Hs.388746
Gene Chromosome Location
Chr.1: 64202802-64267381 on Build GRCh38
Transcripts
NM_152489.1 XM_005270498.4 XM_006710379.3 XM_011540764.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6848843 c.242-1G>A p.? Chr.1: 64210741-64210741 on Build GRCh38
3724066 c.252C>G p.H84Q Chr.1: 64210752-64210752 on Build GRCh38
3724065 c.252C>G p.H84Q Chr.1: 64210752-64210752 on Build GRCh38
4577519 c.277T>C p.L93L Chr.1: 64210777-64210777 on Build GRCh38
6540496 c.247C>T p.P83S Chr.1: 64210747-64210747 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs938979394 Chr.1: 64210655-64210655 on Build GRCh38
rs980689645 Chr.1: 64210927-64210927 on Build GRCh38
rs988195545 Chr.1: 64211048-64211048 on Build GRCh38
rs761799747 Chr.1: 64210866-64210869 on Build GRCh38
rs1005659418 Chr.1: 64211073-64211073 on Build GRCh38
rs994791743 Chr.1: 64211008-64211008 on Build GRCh38
rs370452729 Chr.1: 64210866-64210866 on Build GRCh38
rs888590042 Chr.1: 64211047-64211047 on Build GRCh38
rs192044358 Chr.1: 64210786-64210786 on Build GRCh38
rs537795877 Chr.1: 64210988-64210988 on Build GRCh38
rs769812253 Chr.1: 64210880-64210880 on Build GRCh38
rs186010531 Chr.1: 64210656-64210656 on Build GRCh38
rs768907735 Chr.1: 64210757-64210757 on Build GRCh38
rs969440323 Chr.1: 64211081-64211081 on Build GRCh38
rs368864268 Chr.1: 64210860-64210860 on Build GRCh38
rs776567811 Chr.1: 64210784-64210784 on Build GRCh38
rs770759142 Chr.1: 64210862-64210862 on Build GRCh38
rs200327318 Chr.1: 64210797-64210797 on Build GRCh38
rs946930759 Chr.1: 64210717-64210717 on Build GRCh38
rs867599810 Chr.1: 64210820-64210820 on Build GRCh38
rs775176230 Chr.1: 64210805-64210805 on Build GRCh38
rs745764796 Chr.1: 64210867-64210867 on Build GRCh38
rs756664831 Chr.1: 64210851-64210851 on Build GRCh38
rs750929939 Chr.1: 64210834-64210834 on Build GRCh38
rs928605114 Chr.1: 64210747-64210747 on Build GRCh38
rs377528173 Chr.1: 64211062-64211062 on Build GRCh38
rs755076575 Chr.1: 64210857-64210857 on Build GRCh38
rs756180464 Chr.1: 64210749-64210749 on Build GRCh38
rs778247542 Chr.1: 64210712-64210712 on Build GRCh38
rs942376471 Chr.1: 64210815-64210815 on Build GRCh38
rs764816048 Chr.1: 64210796-64210796 on Build GRCh38
rs1034904632 Chr.1: 64210948-64210948 on Build GRCh38
rs370723679 Chr.1: 64210756-64210756 on Build GRCh38
rs748326204 Chr.1: 64210861-64210861 on Build GRCh38
rs757573369 Chr.1: 64210811-64210811 on Build GRCh38
rs35535097 Chr.1: 64210921-64210922 on Build GRCh38
rs376895959 Chr.1: 64210725-64210725 on Build GRCh38
rs1003588241 Chr.1: 64211086-64211086 on Build GRCh38
rs969386508 Chr.1: 64210670-64210670 on Build GRCh38
rs770646556 Chr.1: 64210777-64210777 on Build GRCh38
rs751908084 Chr.1: 64210714-64210714 on Build GRCh38
rs767835718 Chr.1: 64210818-64210818 on Build GRCh38
rs889525948 Chr.1: 64210807-64210807 on Build GRCh38
rs960420466 Chr.1: 64211041-64211041 on Build GRCh38
rs1047238804 Chr.1: 64211046-64211046 on Build GRCh38
rs774472046 Chr.1: 64210766-64210766 on Build GRCh38
rs913617167 Chr.1: 64210706-64210706 on Build GRCh38
rs781681553 Chr.1: 64210723-64210723 on Build GRCh38
rs768561715 Chr.1: 64210785-64210785 on Build GRCh38
rs776725227 Chr.1: 64210851-64210851 on Build GRCh38
rs969168942 Chr.1: 64210921-64210921 on Build GRCh38
rs1016774234 Chr.1: 64210916-64210916 on Build GRCh38
rs1023705984 Chr.1: 64211076-64211076 on Build GRCh38
rs369382687 Chr.1: 64210809-64210809 on Build GRCh38
rs927614229 Chr.1: 64210652-64210652 on Build GRCh38
rs7532933 Chr.1: 64210769-64210769 on Build GRCh38
rs375399932 Chr.1: 64210810-64210810 on Build GRCh38
rs775587685 Chr.1: 64210885-64210885 on Build GRCh38
rs757584898 Chr.1: 64210720-64210720 on Build GRCh38
rs367642527 Chr.1: 64210855-64210855 on Build GRCh38

View Full Product Details