Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 65147936-65148427 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AK4
Entrez Gene ID
205
Gene Name
adenylate kinase 4
Gene Aliases
AK 4, AK3, AK3L1, AK3L2
UniGene
Hs.10862
Gene Chromosome Location
Chr.1: 65147342-65232145 on Build GRCh38
Transcripts
NM_001005353.2 NM_013410.3 NM_203464.2 XM_017000613.1

SNP Detail

SNP ID Chromosome Location
rs114871912 Chr.1: 65148218-65148218 on Build GRCh38
rs1025300969 Chr.1: 65148385-65148385 on Build GRCh38
rs1049452577 Chr.1: 65148171-65148171 on Build GRCh38
rs1048678089 Chr.1: 65148015-65148015 on Build GRCh38
rs1044461967 Chr.1: 65148219-65148219 on Build GRCh38
rs1008123672 Chr.1: 65148192-65148192 on Build GRCh38
rs1011770520 Chr.1: 65148106-65148106 on Build GRCh38
rs752340573 Chr.1: 65148141-65148158 on Build GRCh38
rs61781805 Chr.1: 65148376-65148376 on Build GRCh38
rs7551080 Chr.1: 65148133-65148133 on Build GRCh38
rs150746311 Chr.1: 65148160-65148160 on Build GRCh38
rs961520261 Chr.1: 65148034-65148034 on Build GRCh38
rs564944065 Chr.1: 65148211-65148211 on Build GRCh38
rs1012237229 Chr.1: 65148304-65148304 on Build GRCh38
rs796755705 Chr.1: 65148046-65148047 on Build GRCh38
rs1028241267 Chr.1: 65147980-65147980 on Build GRCh38
rs751499026 Chr.1: 65148392-65148392 on Build GRCh38
rs1053294809 Chr.1: 65147967-65147967 on Build GRCh38
rs764043259 Chr.1: 65148391-65148391 on Build GRCh38
rs538412701 Chr.1: 65148101-65148101 on Build GRCh38
rs928884773 Chr.1: 65148357-65148357 on Build GRCh38
rs910861892 Chr.1: 65148388-65148388 on Build GRCh38
rs934834839 Chr.1: 65148124-65148124 on Build GRCh38
rs537989843 Chr.1: 65147973-65147973 on Build GRCh38
rs952260811 Chr.1: 65148236-65148236 on Build GRCh38
rs993855434 Chr.1: 65148383-65148383 on Build GRCh38
rs761047604 Chr.1: 65148366-65148366 on Build GRCh38
rs543548043 Chr.1: 65148194-65148194 on Build GRCh38
rs916391220 Chr.1: 65148216-65148216 on Build GRCh38
rs1009803609 Chr.1: 65148205-65148205 on Build GRCh38
rs879883218 Chr.1: 65148082-65148097 on Build GRCh38
rs970046185 Chr.1: 65148341-65148341 on Build GRCh38
rs892076611 Chr.1: 65148296-65148296 on Build GRCh38
rs773538987 Chr.1: 65148360-65148360 on Build GRCh38
rs1041372210 Chr.1: 65148199-65148199 on Build GRCh38
rs1019386075 Chr.1: 65148372-65148372 on Build GRCh38
rs1002796452 Chr.1: 65148077-65148077 on Build GRCh38
rs528035465 Chr.1: 65148312-65148312 on Build GRCh38
rs985174392 Chr.1: 65147957-65147957 on Build GRCh38
rs1026823423 Chr.1: 65148055-65148055 on Build GRCh38
rs776901702 Chr.1: 65148388-65148388 on Build GRCh38
rs918523244 Chr.1: 65148185-65148185 on Build GRCh38
rs1019482665 Chr.1: 65148030-65148030 on Build GRCh38
rs972646858 Chr.1: 65147969-65147969 on Build GRCh38
rs1026394365 Chr.1: 65148231-65148231 on Build GRCh38
rs567548396 Chr.1: 65147968-65147968 on Build GRCh38
rs868202144 Chr.1: 65148018-65148018 on Build GRCh38
rs7550993 Chr.1: 65148046-65148046 on Build GRCh38
rs1053314402 Chr.1: 65148095-65148095 on Build GRCh38
rs952038955 Chr.1: 65148073-65148073 on Build GRCh38
rs984793744 Chr.1: 65148075-65148075 on Build GRCh38
rs117302037 Chr.1: 65148120-65148120 on Build GRCh38
rs768882760 Chr.1: 65148387-65148388 on Build GRCh38
rs531550563 Chr.1: 65148370-65148370 on Build GRCh38
rs377591406 Chr.1: 65148175-65148175 on Build GRCh38
rs889741929 Chr.1: 65148011-65148011 on Build GRCh38
rs948801288 Chr.1: 65148021-65148021 on Build GRCh38
rs972361619 Chr.1: 65148182-65148182 on Build GRCh38
rs758157637 Chr.1: 65148387-65148387 on Build GRCh38
rs34080963 Chr.1: 65148280-65148280 on Build GRCh38
rs889688012 Chr.1: 65148190-65148190 on Build GRCh38
rs549402541 Chr.1: 65148319-65148319 on Build GRCh38
rs1053031326 Chr.1: 65148283-65148283 on Build GRCh38
rs77445149 Chr.1: 65148368-65148368 on Build GRCh38
rs560457050 Chr.1: 65148225-65148225 on Build GRCh38
rs554593284 Chr.1: 65148157-65148157 on Build GRCh38
rs756758889 Chr.1: 65148398-65148398 on Build GRCh38
rs57193680 Chr.1: 65148351-65148351 on Build GRCh38
rs545648428 Chr.1: 65148224-65148224 on Build GRCh38
rs931173212 Chr.1: 65148014-65148014 on Build GRCh38
rs906931603 Chr.1: 65147963-65147963 on Build GRCh38
rs956602147 Chr.1: 65148180-65148180 on Build GRCh38
rs867207961 Chr.1: 65148305-65148305 on Build GRCh38
rs1044408955 Chr.1: 65148024-65148024 on Build GRCh38
rs7550994 Chr.1: 65148047-65148047 on Build GRCh38
rs549384647 Chr.1: 65147965-65147965 on Build GRCh38
rs896548613 Chr.1: 65147989-65147989 on Build GRCh38
rs983999926 Chr.1: 65148207-65148207 on Build GRCh38
rs542556850 Chr.1: 65148139-65148139 on Build GRCh38
rs1013562939 Chr.1: 65147993-65147993 on Build GRCh38
rs1052834087 Chr.1: 65148130-65148130 on Build GRCh38

View Full Product Details