Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 147655069-147655570 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACP6
Entrez Gene ID
51205
Gene Name
acid phosphatase 6, lysophosphatidic
Gene Aliases
ACPL1, LPAP, PACPL1
UniGene
Hs.562154
Gene Chromosome Location
Chr.1: 147629658-147670543 on Build GRCh38
Transcripts
NM_001323625.1 NM_016361.4 NR_136633.1 NR_136634.1 NR_136635.1 NR_136636.1 XM_011509601.2 XM_017001422.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1688479 c.595G>A p.E199K Chr.1: 147655213-147655213 on Build GRCh38
3473211 c.589G>A p.D197N Chr.1: 147655219-147655219 on Build GRCh38
2121798 c.635G>T p.R212M Chr.1: 147655173-147655173 on Build GRCh38
4870180 c.573C>T p.I191I Chr.1: 147655235-147655235 on Build GRCh38
3473210 c.589G>A p.D197N Chr.1: 147655219-147655219 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146958747 Chr.1: 147655223-147655223 on Build GRCh38
rs372140614 Chr.1: 147655221-147655221 on Build GRCh38
rs1050054383 Chr.1: 147655357-147655357 on Build GRCh38
rs893556989 Chr.1: 147655310-147655310 on Build GRCh38
rs962021785 Chr.1: 147655446-147655446 on Build GRCh38
rs370703050 Chr.1: 147655243-147655243 on Build GRCh38
rs781871574 Chr.1: 147655204-147655205 on Build GRCh38
rs587645994 Chr.1: 147655330-147655330 on Build GRCh38
rs885239 Chr.1: 147655487-147655487 on Build GRCh38
rs775862659 Chr.1: 147655483-147655483 on Build GRCh38
rs782656387 Chr.1: 147655171-147655171 on Build GRCh38
rs199697773 Chr.1: 147655187-147655187 on Build GRCh38
rs140840216 Chr.1: 147655248-147655248 on Build GRCh38
rs587692956 Chr.1: 147655274-147655274 on Build GRCh38
rs781979495 Chr.1: 147655155-147655155 on Build GRCh38
rs782544035 Chr.1: 147655292-147655292 on Build GRCh38
rs995929615 Chr.1: 147655349-147655349 on Build GRCh38
rs990879213 Chr.1: 147655497-147655497 on Build GRCh38
rs368786453 Chr.1: 147655178-147655178 on Build GRCh38
rs587754801 Chr.1: 147655508-147655508 on Build GRCh38
rs782159489 Chr.1: 147655385-147655385 on Build GRCh38
rs782137483 Chr.1: 147655279-147655279 on Build GRCh38
rs769331972 Chr.1: 147655465-147655465 on Build GRCh38
rs916555128 Chr.1: 147655501-147655501 on Build GRCh38
rs902787240 Chr.1: 147655261-147655261 on Build GRCh38
rs149218889 Chr.1: 147655205-147655205 on Build GRCh38
rs962169156 Chr.1: 147655096-147655096 on Build GRCh38
rs781783425 Chr.1: 147655241-147655241 on Build GRCh38
rs782821093 Chr.1: 147655228-147655228 on Build GRCh38
rs1015243964 Chr.1: 147655426-147655426 on Build GRCh38
rs890145436 Chr.1: 147655406-147655406 on Build GRCh38
rs782595470 Chr.1: 147655298-147655298 on Build GRCh38
rs782535969 Chr.1: 147655121-147655121 on Build GRCh38
rs919998905 Chr.1: 147655164-147655164 on Build GRCh38
rs782195228 Chr.1: 147655135-147655135 on Build GRCh38
rs190533863 Chr.1: 147655456-147655456 on Build GRCh38
rs587678861 Chr.1: 147655412-147655412 on Build GRCh38
rs587723021 Chr.1: 147655185-147655185 on Build GRCh38
rs782468417 Chr.1: 147655137-147655137 on Build GRCh38
rs782323936 Chr.1: 147655179-147655179 on Build GRCh38
rs973289776 Chr.1: 147655115-147655115 on Build GRCh38
rs782245916 Chr.1: 147655161-147655161 on Build GRCh38
rs75106646 Chr.1: 147655486-147655486 on Build GRCh38
rs375087381 Chr.1: 147655259-147655259 on Build GRCh38
rs782563216 Chr.1: 147655131-147655131 on Build GRCh38

View Full Product Details