Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 149926982-149927530 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SF3B4
Entrez Gene ID
10262
Gene Name
splicing factor 3b subunit 4
Gene Aliases
AFD1, Hsh49, SAP49, SF3b49
UniGene
Hs.516160
Gene Chromosome Location
Chr.1: 149923317-149928252 on Build GRCh38
Transcripts
NM_005850.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6776285 c.156G>A p.Q52Q Chr.1: 149927173-149927173 on Build GRCh38
3770299 c.110G>T p.G37V Chr.1: 149927219-149927219 on Build GRCh38
1320015 c.125_126insAC p.H43fs*21 Chr.1: 149927203-149927204 on Build GRCh38
4021901 c.93A>T p.E31D Chr.1: 149927236-149927236 on Build GRCh38
5558938 c.59T>A p.L20Q Chr.1: 149927270-149927270 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61601659 Chr.1: 149927073-149927073 on Build GRCh38
rs781157974 Chr.1: 149927444-149927444 on Build GRCh38
rs781851440 Chr.1: 149927301-149927301 on Build GRCh38
rs587597218 Chr.1: 149927108-149927108 on Build GRCh38
rs988668666 Chr.1: 149927377-149927377 on Build GRCh38
rs782253120 Chr.1: 149927148-149927148 on Build GRCh38
rs934468632 Chr.1: 149927354-149927354 on Build GRCh38
rs1049441633 Chr.1: 149927064-149927064 on Build GRCh38
rs782819179 Chr.1: 149927258-149927258 on Build GRCh38
rs112658154 Chr.1: 149927395-149927395 on Build GRCh38
rs782472259 Chr.1: 149927284-149927284 on Build GRCh38
rs782545261 Chr.1: 149927346-149927346 on Build GRCh38
rs782137683 Chr.1: 149927331-149927331 on Build GRCh38
rs947299827 Chr.1: 149927399-149927399 on Build GRCh38
rs373226217 Chr.1: 149927185-149927185 on Build GRCh38
rs782214031 Chr.1: 149927137-149927137 on Build GRCh38
rs993600960 Chr.1: 149927461-149927461 on Build GRCh38
rs781988019 Chr.1: 149927326-149927326 on Build GRCh38
rs60200537 Chr.1: 149927072-149927072 on Build GRCh38
rs781877955 Chr.1: 149927334-149927335 on Build GRCh38
rs782219908 Chr.1: 149927319-149927319 on Build GRCh38
rs782769568 Chr.1: 149927224-149927224 on Build GRCh38
rs782146401 Chr.1: 149927246-149927246 on Build GRCh38
rs782364613 Chr.1: 149927323-149927323 on Build GRCh38
rs886045256 Chr.1: 149927181-149927181 on Build GRCh38
rs934519273 Chr.1: 149927025-149927025 on Build GRCh38
rs782614820 Chr.1: 149927290-149927290 on Build GRCh38
rs1047895669 Chr.1: 149927489-149927489 on Build GRCh38
rs374282221 Chr.1: 149927302-149927302 on Build GRCh38
rs781915879 Chr.1: 149927159-149927159 on Build GRCh38
rs797045956 Chr.1: 149927283-149927284 on Build GRCh38
rs782333528 Chr.1: 149927188-149927188 on Build GRCh38
rs587657536 Chr.1: 149927317-149927317 on Build GRCh38
rs781875043 Chr.1: 149927244-149927244 on Build GRCh38
rs115397237 Chr.1: 149927337-149927337 on Build GRCh38
rs782403964 Chr.1: 149927157-149927157 on Build GRCh38
rs782032346 Chr.1: 149927332-149927332 on Build GRCh38
rs893088397 Chr.1: 149927040-149927040 on Build GRCh38
rs369731978 Chr.1: 149927031-149927031 on Build GRCh38
rs782106639 Chr.1: 149927215-149927215 on Build GRCh38
rs782275913 Chr.1: 149927125-149927125 on Build GRCh38
rs1053015923 Chr.1: 149927032-149927032 on Build GRCh38
rs781838123 Chr.1: 149927340-149927340 on Build GRCh38
rs200302531 Chr.1: 149927120-149927120 on Build GRCh38
rs781806467 Chr.1: 149927272-149927272 on Build GRCh38
rs770682995 Chr.1: 149927352-149927352 on Build GRCh38
rs781955987 Chr.1: 149927207-149927207 on Build GRCh38
rs902009309 Chr.1: 149927167-149927167 on Build GRCh38
rs943743013 Chr.1: 149927410-149927410 on Build GRCh38
rs782553649 Chr.1: 149927129-149927129 on Build GRCh38
rs781996437 Chr.1: 149927147-149927147 on Build GRCh38
rs1006431478 Chr.1: 149927498-149927498 on Build GRCh38
rs16836630 Chr.1: 149927059-149927059 on Build GRCh38
rs1049829729 Chr.1: 149927400-149927400 on Build GRCh38
rs782774900 Chr.1: 149927346-149927346 on Build GRCh38
rs970507513 Chr.1: 149927324-149927324 on Build GRCh38
rs377537483 Chr.1: 149927116-149927116 on Build GRCh38
rs587673569 Chr.1: 149927112-149927112 on Build GRCh38
rs782629859 Chr.1: 149927145-149927145 on Build GRCh38
rs74124093 Chr.1: 149927010-149927010 on Build GRCh38
rs587708376 Chr.1: 149927134-149927134 on Build GRCh38
rs587636381 Chr.1: 149927043-149927043 on Build GRCh38
rs2305387 Chr.1: 149927408-149927408 on Build GRCh38
rs782116003 Chr.1: 149927341-149927341 on Build GRCh38
rs368160350 Chr.1: 149927303-149927303 on Build GRCh38
rs928744296 Chr.1: 149927351-149927351 on Build GRCh38
rs782643604 Chr.1: 149927117-149927117 on Build GRCh38
rs201447834 Chr.1: 149927245-149927245 on Build GRCh38
rs782051278 Chr.1: 149927333-149927333 on Build GRCh38
rs1026503531 Chr.1: 149927280-149927280 on Build GRCh38
rs587641710 Chr.1: 149927505-149927505 on Build GRCh38
rs782652150 Chr.1: 149927304-149927304 on Build GRCh38
rs782353230 Chr.1: 149927086-149927086 on Build GRCh38
rs782284128 Chr.1: 149927305-149927305 on Build GRCh38
rs797045134 Chr.1: 149927241-149927241 on Build GRCh38
rs782584634 Chr.1: 149927130-149927130 on Build GRCh38
rs45548131 Chr.1: 149927497-149927497 on Build GRCh38
rs782336866 Chr.1: 149927418-149927418 on Build GRCh38

View Full Product Details