Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137462449-137462980 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PNPLA7
Entrez Gene ID
375775
Gene Name
patatin like phospholipase domain containing 7
Gene Aliases
C9orf111, NTE-R1, NTEL1
UniGene
Hs.294147
Gene Chromosome Location
Chr.9: 137459449-137550666 on Build GRCh38
Transcripts
NM_001098537.2 NM_152286.4 XM_006717102.1 XM_006717104.3 XM_017014709.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3745918 c.3315C>T p.D1105D Chr.9: 137462787-137462787 on Build GRCh38
7233957 c.3344-2A>T p.? Chr.9: 137462835-137462835 on Build GRCh38
3716274 c.3408G>A p.T1136T Chr.9: 137462694-137462694 on Build GRCh38
7233958 c.3269-2A>T p.? Chr.9: 137462835-137462835 on Build GRCh38
4944208 c.3413A>T p.D1138V Chr.9: 137462764-137462764 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779042139 Chr.9: 137462795-137462795 on Build GRCh38
rs942614091 Chr.9: 137462512-137462512 on Build GRCh38
rs111934135 Chr.9: 137462696-137462696 on Build GRCh38
rs538240638 Chr.9: 137462909-137462909 on Build GRCh38
rs558573662 Chr.9: 137462561-137462561 on Build GRCh38
rs780059289 Chr.9: 137462637-137462637 on Build GRCh38
rs772885608 Chr.9: 137462639-137462639 on Build GRCh38
rs1021888911 Chr.9: 137462705-137462705 on Build GRCh38
rs866850793 Chr.9: 137462640-137462640 on Build GRCh38
rs947534926 Chr.9: 137462944-137462944 on Build GRCh38
rs763694583 Chr.9: 137462858-137462858 on Build GRCh38
rs540714781 Chr.9: 137462690-137462690 on Build GRCh38
rs546269845 Chr.9: 137462822-137462822 on Build GRCh38
rs748212059 Chr.9: 137462816-137462816 on Build GRCh38
rs751565269 Chr.9: 137462776-137462776 on Build GRCh38
rs374416034 Chr.9: 137462833-137462833 on Build GRCh38
rs531984473 Chr.9: 137462766-137462766 on Build GRCh38
rs763175109 Chr.9: 137462672-137462672 on Build GRCh38
rs893170276 Chr.9: 137462954-137462954 on Build GRCh38
rs1021607566 Chr.9: 137462497-137462497 on Build GRCh38
rs924730217 Chr.9: 137462478-137462478 on Build GRCh38
rs746821963 Chr.9: 137462715-137462715 on Build GRCh38
rs933915664 Chr.9: 137462800-137462800 on Build GRCh38
rs919557621 Chr.9: 137462599-137462599 on Build GRCh38
rs890859620 Chr.9: 137462662-137462662 on Build GRCh38
rs1011802362 Chr.9: 137462693-137462693 on Build GRCh38
rs575536310 Chr.9: 137462595-137462595 on Build GRCh38
rs772519440 Chr.9: 137462747-137462747 on Build GRCh38
rs774310424 Chr.9: 137462742-137462742 on Build GRCh38
rs369634710 Chr.9: 137462832-137462832 on Build GRCh38
rs943828210 Chr.9: 137462668-137462668 on Build GRCh38
rs774886835 Chr.9: 137462676-137462676 on Build GRCh38
rs538277530 Chr.9: 137462550-137462550 on Build GRCh38
rs745911932 Chr.9: 137462866-137462867 on Build GRCh38
rs747121425 Chr.9: 137462882-137462882 on Build GRCh38
rs377751983 Chr.9: 137462839-137462839 on Build GRCh38
rs538044782 Chr.9: 137462471-137462471 on Build GRCh38
rs199798551 Chr.9: 137462860-137462860 on Build GRCh38
rs41297243 Chr.9: 137462524-137462524 on Build GRCh38
rs146857442 Chr.9: 137462757-137462757 on Build GRCh38
rs757491308 Chr.9: 137462861-137462862 on Build GRCh38
rs752612353 Chr.9: 137462702-137462702 on Build GRCh38
rs989294219 Chr.9: 137462509-137462509 on Build GRCh38
rs147430973 Chr.9: 137462627-137462627 on Build GRCh38
rs544189968 Chr.9: 137462618-137462618 on Build GRCh38
rs562746536 Chr.9: 137462763-137462763 on Build GRCh38
rs973972877 Chr.9: 137462589-137462589 on Build GRCh38
rs781489343 Chr.9: 137462863-137462864 on Build GRCh38
rs546218119 Chr.9: 137462741-137462741 on Build GRCh38
rs761072855 Chr.9: 137462749-137462749 on Build GRCh38
rs1013543578 Chr.9: 137462885-137462885 on Build GRCh38
rs770119844 Chr.9: 137462721-137462721 on Build GRCh38
rs141473646 Chr.9: 137462734-137462734 on Build GRCh38
rs568562894 Chr.9: 137462519-137462519 on Build GRCh38
rs907420274 Chr.9: 137462939-137462939 on Build GRCh38
rs960184083 Chr.9: 137462534-137462534 on Build GRCh38
rs746090822 Chr.9: 137462645-137462645 on Build GRCh38
rs778425762 Chr.9: 137462698-137462698 on Build GRCh38
rs145096429 Chr.9: 137462752-137462752 on Build GRCh38
rs773591065 Chr.9: 137462748-137462748 on Build GRCh38
rs908404615 Chr.9: 137462567-137462567 on Build GRCh38
rs762364688 Chr.9: 137462743-137462743 on Build GRCh38
rs781501723 Chr.9: 137462919-137462919 on Build GRCh38
rs943755876 Chr.9: 137462902-137462902 on Build GRCh38
rs552663296 Chr.9: 137462533-137462533 on Build GRCh38
rs764295231 Chr.9: 137462673-137462673 on Build GRCh38
rs1031654653 Chr.9: 137462952-137462952 on Build GRCh38
rs560596524 Chr.9: 137462695-137462695 on Build GRCh38
rs369324475 Chr.9: 137462634-137462634 on Build GRCh38
rs753679904 Chr.9: 137462750-137462751 on Build GRCh38
rs890919399 Chr.9: 137462473-137462473 on Build GRCh38
rs754763599 Chr.9: 137462818-137462819 on Build GRCh38
rs954018020 Chr.9: 137462799-137462799 on Build GRCh38
rs112219485 Chr.9: 137462485-137462485 on Build GRCh38
rs145413682 Chr.9: 137462710-137462710 on Build GRCh38
rs568603035 Chr.9: 137462818-137462818 on Build GRCh38
rs200913584 Chr.9: 137462767-137462767 on Build GRCh38
rs566118472 Chr.9: 137462535-137462535 on Build GRCh38
rs767411094 Chr.9: 137462877-137462877 on Build GRCh38
rs777620121 Chr.9: 137462775-137462775 on Build GRCh38
rs773539669 Chr.9: 137462820-137462820 on Build GRCh38
rs912250825 Chr.9: 137462863-137462863 on Build GRCh38
rs762234547 Chr.9: 137462677-137462677 on Build GRCh38
rs777622263 Chr.9: 137462713-137462713 on Build GRCh38
rs191119375 Chr.9: 137462468-137462468 on Build GRCh38
rs1046716323 Chr.9: 137462545-137462545 on Build GRCh38
rs200124849 Chr.9: 137462669-137462669 on Build GRCh38
rs1011854417 Chr.9: 137462489-137462489 on Build GRCh38
rs542777162 Chr.9: 137462467-137462467 on Build GRCh38
rs781143660 Chr.9: 137462786-137462786 on Build GRCh38
rs113253090 Chr.9: 137462511-137462511 on Build GRCh38
rs894636571 Chr.9: 137462881-137462881 on Build GRCh38
rs750807912 Chr.9: 137462683-137462683 on Build GRCh38
rs939915866 Chr.9: 137462587-137462587 on Build GRCh38
rs765018245 Chr.9: 137462856-137462857 on Build GRCh38
rs1043280061 Chr.9: 137462945-137462945 on Build GRCh38
rs775681574 Chr.9: 137462671-137462671 on Build GRCh38
rs202161097 Chr.9: 137462821-137462821 on Build GRCh38
rs766056386 Chr.9: 137462694-137462694 on Build GRCh38
rs183661910 Chr.9: 137462777-137462777 on Build GRCh38
rs767868155 Chr.9: 137462681-137462681 on Build GRCh38
rs913364866 Chr.9: 137462920-137462920 on Build GRCh38
rs376246625 Chr.9: 137462787-137462787 on Build GRCh38
rs1008805631 Chr.9: 137462474-137462474 on Build GRCh38
rs1052864194 Chr.9: 137462916-137462916 on Build GRCh38
rs566278578 Chr.9: 137462861-137462861 on Build GRCh38
rs751747209 Chr.9: 137462859-137462860 on Build GRCh38
rs892036457 Chr.9: 137462486-137462486 on Build GRCh38
rs766752239 Chr.9: 137462750-137462750 on Build GRCh38
rs762322711 Chr.9: 137462834-137462834 on Build GRCh38
rs372947422 Chr.9: 137462728-137462728 on Build GRCh38

View Full Product Details