Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 7049138-7049647 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PUDP
Entrez Gene ID
8226
Gene Name
pseudouridine 5'-phosphatase
Gene Aliases
DXF68S1E, FAM16AX, GS1, HDHD1, HDHD1A
UniGene
Hs.185910
Gene Chromosome Location
Chr.X: 6768840-7148190 on Build GRCh38
Transcripts
NM_001135565.1 NM_001178136.1 NM_012080.4

SNP Detail

SNP ID Chromosome Location
rs1040294257 Chr.X: 7049454-7049454 on Build GRCh38
rs745754701 Chr.X: 7049570-7049570 on Build GRCh38
rs184721669 Chr.X: 7049389-7049389 on Build GRCh38
rs1041825575 Chr.X: 7049507-7049507 on Build GRCh38
rs942854200 Chr.X: 7049353-7049353 on Build GRCh38
rs1011071056 Chr.X: 7049363-7049363 on Build GRCh38
rs910153532 Chr.X: 7049347-7049347 on Build GRCh38
rs755072308 Chr.X: 7049185-7049185 on Build GRCh38
rs1005989998 Chr.X: 7049617-7049617 on Build GRCh38
rs1020737608 Chr.X: 7049384-7049384 on Build GRCh38
rs934070819 Chr.X: 7049497-7049497 on Build GRCh38
rs751686739 Chr.X: 7049178-7049178 on Build GRCh38
rs929664478 Chr.X: 7049280-7049280 on Build GRCh38
rs773511808 Chr.X: 7049486-7049486 on Build GRCh38
rs772473747 Chr.X: 7049484-7049484 on Build GRCh38
rs1803676 Chr.X: 7049556-7049556 on Build GRCh38
rs1032233549 Chr.X: 7049543-7049543 on Build GRCh38
rs984351774 Chr.X: 7049312-7049312 on Build GRCh38
rs1003185356 Chr.X: 7049460-7049460 on Build GRCh38
rs1000183856 Chr.X: 7049540-7049540 on Build GRCh38
rs767543514 Chr.X: 7049191-7049191 on Build GRCh38
rs890357021 Chr.X: 7049247-7049247 on Build GRCh38
rs756304004 Chr.X: 7049282-7049282 on Build GRCh38
rs1534276 Chr.X: 7049313-7049313 on Build GRCh38
rs150384387 Chr.X: 7049539-7049539 on Build GRCh38
rs901217592 Chr.X: 7049482-7049482 on Build GRCh38
rs140378369 Chr.X: 7049414-7049414 on Build GRCh38
rs970469661 Chr.X: 7049237-7049237 on Build GRCh38
rs189757478 Chr.X: 7049620-7049620 on Build GRCh38
rs752919515 Chr.X: 7049281-7049281 on Build GRCh38
rs748530570 Chr.X: 7049360-7049360 on Build GRCh38
rs755772190 Chr.X: 7049256-7049256 on Build GRCh38
rs777488955 Chr.X: 7049295-7049295 on Build GRCh38
rs137892869 Chr.X: 7049594-7049594 on Build GRCh38
rs779727336 Chr.X: 7049257-7049257 on Build GRCh38

View Full Product Details