Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 130902650-130903154 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FIBCD1
Entrez Gene ID
84929
Gene Name
fibrinogen C domain containing 1
Gene Aliases
-
UniGene
Hs.133205
Gene Chromosome Location
Chr.9: 130902438-130939068 on Build GRCh38
Transcripts
NM_001145106.1 NM_032843.4

SNP Detail

SNP ID Chromosome Location
rs568481756 Chr.9: 130902862-130902862 on Build GRCh38
rs944558420 Chr.9: 130902830-130902830 on Build GRCh38
rs921749279 Chr.9: 130902984-130902984 on Build GRCh38
rs909097385 Chr.9: 130903024-130903024 on Build GRCh38
rs1003349734 Chr.9: 130902763-130902763 on Build GRCh38
rs908143724 Chr.9: 130902942-130902942 on Build GRCh38
rs973655537 Chr.9: 130903114-130903114 on Build GRCh38
rs963753828 Chr.9: 130902693-130902693 on Build GRCh38
rs906044608 Chr.9: 130903091-130903091 on Build GRCh38
rs975181010 Chr.9: 130902697-130902697 on Build GRCh38
rs142764384 Chr.9: 130902971-130902971 on Build GRCh38
rs949098953 Chr.9: 130903079-130903079 on Build GRCh38
rs940700682 Chr.9: 130902721-130902721 on Build GRCh38
rs939183340 Chr.9: 130902993-130902993 on Build GRCh38
rs988144800 Chr.9: 130902710-130902710 on Build GRCh38
rs1052799879 Chr.9: 130902784-130902784 on Build GRCh38
rs1015086303 Chr.9: 130903104-130903104 on Build GRCh38
rs531689132 Chr.9: 130903105-130903105 on Build GRCh38
rs11791437 Chr.9: 130902730-130902730 on Build GRCh38
rs940379185 Chr.9: 130903025-130903025 on Build GRCh38
rs566389314 Chr.9: 130903043-130903043 on Build GRCh38
rs548498754 Chr.9: 130903118-130903118 on Build GRCh38
rs535213023 Chr.9: 130902791-130902791 on Build GRCh38
rs922131360 Chr.9: 130902700-130902700 on Build GRCh38
rs545737956 Chr.9: 130902930-130902930 on Build GRCh38
rs1024477565 Chr.9: 130902907-130902907 on Build GRCh38
rs764054489 Chr.9: 130903082-130903082 on Build GRCh38
rs953323057 Chr.9: 130902992-130902992 on Build GRCh38
rs970291388 Chr.9: 130902746-130902746 on Build GRCh38
rs1022003088 Chr.9: 130902828-130902828 on Build GRCh38
rs948176037 Chr.9: 130902745-130902745 on Build GRCh38
rs1029386961 Chr.9: 130902867-130902867 on Build GRCh38
rs931895565 Chr.9: 130902849-130902849 on Build GRCh38
rs552826197 Chr.9: 130902940-130902940 on Build GRCh38
rs906387120 Chr.9: 130902758-130902758 on Build GRCh38
rs1048704162 Chr.9: 130902692-130902693 on Build GRCh38
rs892295165 Chr.9: 130902998-130902998 on Build GRCh38
rs111869806 Chr.9: 130902751-130902751 on Build GRCh38
rs576816625 Chr.9: 130902919-130902919 on Build GRCh38
rs537504284 Chr.9: 130902875-130902875 on Build GRCh38
rs186236650 Chr.9: 130902687-130902687 on Build GRCh38
rs398087595 Chr.9: 130902697-130902697 on Build GRCh38
rs996504685 Chr.9: 130903058-130903058 on Build GRCh38
rs532958005 Chr.9: 130902997-130902997 on Build GRCh38
rs557451795 Chr.9: 130903101-130903101 on Build GRCh38
rs114528303 Chr.9: 130902708-130902708 on Build GRCh38
rs1020701176 Chr.9: 130903117-130903117 on Build GRCh38
rs566326037 Chr.9: 130902787-130902787 on Build GRCh38
rs570383173 Chr.9: 130902709-130902709 on Build GRCh38
rs963701906 Chr.9: 130902842-130902842 on Build GRCh38
rs112612902 Chr.9: 130903030-130903030 on Build GRCh38
rs920588823 Chr.9: 130902740-130902740 on Build GRCh38
rs928047447 Chr.9: 130902986-130902986 on Build GRCh38
rs754082332 Chr.9: 130903100-130903100 on Build GRCh38
rs1043644761 Chr.9: 130903040-130903040 on Build GRCh38
rs892347808 Chr.9: 130902786-130902786 on Build GRCh38
rs187988491 Chr.9: 130902972-130902972 on Build GRCh38
rs896191988 Chr.9: 130902885-130902885 on Build GRCh38
rs893344816 Chr.9: 130903103-130903103 on Build GRCh38
rs879403606 Chr.9: 130903076-130903076 on Build GRCh38
rs572735981 Chr.9: 130902946-130902946 on Build GRCh38
rs908112582 Chr.9: 130902720-130902720 on Build GRCh38
rs138705248 Chr.9: 130902876-130902876 on Build GRCh38
rs200266546 Chr.9: 130902802-130902802 on Build GRCh38
rs191041984 Chr.9: 130902960-130902960 on Build GRCh38
rs566211174 Chr.9: 130902959-130902959 on Build GRCh38
rs887435279 Chr.9: 130902850-130902850 on Build GRCh38
rs182521928 Chr.9: 130902966-130902966 on Build GRCh38
rs1022055409 Chr.9: 130902692-130902692 on Build GRCh38
rs1029830553 Chr.9: 130903090-130903090 on Build GRCh38
rs1010582383 Chr.9: 130902819-130902819 on Build GRCh38
rs997022078 Chr.9: 130902845-130902845 on Build GRCh38
rs1014466904 Chr.9: 130902904-130902904 on Build GRCh38
rs940319559 Chr.9: 130902866-130902866 on Build GRCh38
rs11312213 Chr.9: 130902693-130902693 on Build GRCh38

View Full Product Details