Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 133362555-133363052 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SURF4
Entrez Gene ID
6836
Gene Name
surfeit 4
Gene Aliases
ERV29
UniGene
Hs.512465
Gene Chromosome Location
Chr.9: 133361449-133377949 on Build GRCh38
Transcripts
NM_001280788.1 NM_001280789.1 NM_001280790.1 NM_001280791.1 NM_001280792.1 NM_001280793.1 NM_033161.3 XM_011518943.1 XM_011518944.2 XM_011518945.2 XM_017015062.1

SNP Detail

SNP ID Chromosome Location
rs112902455 Chr.9: 133362638-133362638 on Build GRCh38
rs916716707 Chr.9: 133362637-133362637 on Build GRCh38
rs149744232 Chr.9: 133362931-133362931 on Build GRCh38
rs181331016 Chr.9: 133362796-133362796 on Build GRCh38
rs587758357 Chr.9: 133362624-133362624 on Build GRCh38
rs587719147 Chr.9: 133362911-133362911 on Build GRCh38
rs185237735 Chr.9: 133362840-133362840 on Build GRCh38
rs892232124 Chr.9: 133362841-133362841 on Build GRCh38
rs186392195 Chr.9: 133362949-133362949 on Build GRCh38
rs201309595 Chr.9: 133362613-133362613 on Build GRCh38
rs968131979 Chr.9: 133362798-133362798 on Build GRCh38
rs939588765 Chr.9: 133362618-133362618 on Build GRCh38
rs968018557 Chr.9: 133362603-133362603 on Build GRCh38
rs78266970 Chr.9: 133362910-133362910 on Build GRCh38
rs963949408 Chr.9: 133362635-133362635 on Build GRCh38
rs587757813 Chr.9: 133362916-133362916 on Build GRCh38
rs890836419 Chr.9: 133362712-133362712 on Build GRCh38
rs907986382 Chr.9: 133362710-133362711 on Build GRCh38
rs929611027 Chr.9: 133362675-133362675 on Build GRCh38
rs371842564 Chr.9: 133362954-133362955 on Build GRCh38
rs986590800 Chr.9: 133362690-133362690 on Build GRCh38
rs41302669 Chr.9: 133362876-133362876 on Build GRCh38
rs116430927 Chr.9: 133362787-133362787 on Build GRCh38
rs899540396 Chr.9: 133362902-133362902 on Build GRCh38
rs1052541860 Chr.9: 133362814-133362814 on Build GRCh38
rs782200451 Chr.9: 133362926-133362926 on Build GRCh38
rs200974230 Chr.9: 133362987-133362989 on Build GRCh38
rs924180097 Chr.9: 133362607-133362607 on Build GRCh38
rs992461877 Chr.9: 133362625-133362625 on Build GRCh38
rs587670687 Chr.9: 133362795-133362795 on Build GRCh38
rs2494 Chr.9: 133362957-133362957 on Build GRCh38
rs978232465 Chr.9: 133362815-133362815 on Build GRCh38
rs1045355508 Chr.9: 133362792-133362792 on Build GRCh38
rs202135735 Chr.9: 133362950-133362951 on Build GRCh38
rs112514889 Chr.9: 133362989-133362989 on Build GRCh38
rs1038056158 Chr.9: 133362644-133362644 on Build GRCh38
rs587702406 Chr.9: 133362598-133362598 on Build GRCh38
rs1007910308 Chr.9: 133362884-133362884 on Build GRCh38
rs113643863 Chr.9: 133362988-133362988 on Build GRCh38
rs960761654 Chr.9: 133362851-133362851 on Build GRCh38
rs1018222395 Chr.9: 133362766-133362766 on Build GRCh38
rs759994156 Chr.9: 133362711-133362711 on Build GRCh38
rs35905067 Chr.9: 133362840-133362841 on Build GRCh38
rs1008545422 Chr.9: 133362754-133362754 on Build GRCh38
rs952613026 Chr.9: 133362643-133362643 on Build GRCh38
rs114749077 Chr.9: 133362978-133362978 on Build GRCh38
rs898253969 Chr.9: 133362674-133362674 on Build GRCh38
rs992209498 Chr.9: 133362965-133362965 on Build GRCh38
rs948250631 Chr.9: 133362641-133362641 on Build GRCh38
rs782699943 Chr.9: 133362605-133362605 on Build GRCh38
rs948198803 Chr.9: 133362919-133362919 on Build GRCh38
rs899640859 Chr.9: 133362783-133362783 on Build GRCh38
rs587633441 Chr.9: 133362632-133362632 on Build GRCh38
rs1019244712 Chr.9: 133362893-133362893 on Build GRCh38
rs943732755 Chr.9: 133362970-133362970 on Build GRCh38
rs587763416 Chr.9: 133362829-133362829 on Build GRCh38
rs899755409 Chr.9: 133363020-133363020 on Build GRCh38
rs587754501 Chr.9: 133362898-133362912 on Build GRCh38
rs981161796 Chr.9: 133363016-133363016 on Build GRCh38
rs1019212190 Chr.9: 133362622-133362622 on Build GRCh38
rs929725344 Chr.9: 133362934-133362934 on Build GRCh38
rs915342962 Chr.9: 133362591-133362591 on Build GRCh38
rs71281255 Chr.9: 133362841-133362842 on Build GRCh38

View Full Product Details