Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 153935271-153935815 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RENBP
Entrez Gene ID
5973
Gene Name
renin binding protein
Gene Aliases
RBP, RNBP
UniGene
Hs.158331
Gene Chromosome Location
Chr.X: 153935263-153944780 on Build GRCh38
Transcripts
NM_002910.5 XM_017029698.1

SNP Detail

SNP ID Chromosome Location
rs782146542 Chr.X: 153935611-153935611 on Build GRCh38
rs782559240 Chr.X: 153935512-153935512 on Build GRCh38
rs782681906 Chr.X: 153935651-153935651 on Build GRCh38
rs782208847 Chr.X: 153935604-153935604 on Build GRCh38
rs371634111 Chr.X: 153935408-153935408 on Build GRCh38
rs781819467 Chr.X: 153935465-153935465 on Build GRCh38
rs782791865 Chr.X: 153935452-153935452 on Build GRCh38
rs374422791 Chr.X: 153935419-153935419 on Build GRCh38
rs367723486 Chr.X: 153935434-153935434 on Build GRCh38
rs782638616 Chr.X: 153935518-153935518 on Build GRCh38
rs782382654 Chr.X: 153935393-153935393 on Build GRCh38
rs782031567 Chr.X: 153935437-153935437 on Build GRCh38
rs183725640 Chr.X: 153935586-153935586 on Build GRCh38
rs781913876 Chr.X: 153935420-153935420 on Build GRCh38
rs41311382 Chr.X: 153935704-153935704 on Build GRCh38
rs781829210 Chr.X: 153935419-153935419 on Build GRCh38
rs199842412 Chr.X: 153935480-153935480 on Build GRCh38
rs529444121 Chr.X: 153935572-153935572 on Build GRCh38
rs782799079 Chr.X: 153935299-153935299 on Build GRCh38
rs782168856 Chr.X: 153935541-153935541 on Build GRCh38
rs782470753 Chr.X: 153935529-153935529 on Build GRCh38
rs146752040 Chr.X: 153935571-153935571 on Build GRCh38
rs781890302 Chr.X: 153935484-153935484 on Build GRCh38
rs781879128 Chr.X: 153935527-153935527 on Build GRCh38
rs782618876 Chr.X: 153935333-153935333 on Build GRCh38
rs782619559 Chr.X: 153935540-153935540 on Build GRCh38
rs781854646 Chr.X: 153935307-153935307 on Build GRCh38
rs913977166 Chr.X: 153935598-153935598 on Build GRCh38
rs782194193 Chr.X: 153935762-153935762 on Build GRCh38
rs782051268 Chr.X: 153935472-153935472 on Build GRCh38
rs1055784594 Chr.X: 153935771-153935771 on Build GRCh38
rs782553907 Chr.X: 153935732-153935732 on Build GRCh38
rs782423931 Chr.X: 153935640-153935640 on Build GRCh38
rs782668246 Chr.X: 153935375-153935375 on Build GRCh38
rs782216959 Chr.X: 153935412-153935412 on Build GRCh38
rs782296539 Chr.X: 153935388-153935388 on Build GRCh38
rs782308917 Chr.X: 153935606-153935606 on Build GRCh38
rs781792536 Chr.X: 153935357-153935357 on Build GRCh38
rs894178331 Chr.X: 153935772-153935772 on Build GRCh38
rs782051988 Chr.X: 153935318-153935319 on Build GRCh38
rs945418192 Chr.X: 153935615-153935615 on Build GRCh38
rs782512087 Chr.X: 153935330-153935330 on Build GRCh38
rs782448933 Chr.X: 153935558-153935558 on Build GRCh38
rs782063125 Chr.X: 153935426-153935426 on Build GRCh38
rs925427110 Chr.X: 153935683-153935683 on Build GRCh38
rs782106958 Chr.X: 153935441-153935441 on Build GRCh38

View Full Product Details