Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154791691-154792191 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MPP1
Entrez Gene ID
4354
Gene Name
membrane palmitoylated protein 1
Gene Aliases
AAG12, DXS552E, EMP55, MRG1, PEMP
UniGene
Hs.496984
Gene Chromosome Location
Chr.X: 154778684-154805527 on Build GRCh38
Transcripts
NM_001166460.1 NM_001166461.1 NM_001166462.1 NM_002436.3 XM_011531167.1 XM_011531169.1

SNP Detail

SNP ID Chromosome Location
rs782605910 Chr.X: 154791821-154791821 on Build GRCh38
rs374791319 Chr.X: 154791805-154791805 on Build GRCh38
rs782754739 Chr.X: 154791740-154791740 on Build GRCh38
rs956911643 Chr.X: 154791949-154791949 on Build GRCh38
rs7474090 Chr.X: 154791938-154791938 on Build GRCh38
rs200723584 Chr.X: 154791863-154791863 on Build GRCh38
rs994515038 Chr.X: 154791836-154791836 on Build GRCh38
rs113285457 Chr.X: 154791918-154791918 on Build GRCh38
rs367591122 Chr.X: 154791860-154791860 on Build GRCh38
rs782151706 Chr.X: 154791776-154791776 on Build GRCh38
rs781783367 Chr.X: 154791758-154791758 on Build GRCh38
rs781966177 Chr.X: 154792131-154792131 on Build GRCh38
rs1022553048 Chr.X: 154791968-154791968 on Build GRCh38
rs140876301 Chr.X: 154791919-154791919 on Build GRCh38
rs782074032 Chr.X: 154791734-154791734 on Build GRCh38
rs782340240 Chr.X: 154792121-154792121 on Build GRCh38
rs782273481 Chr.X: 154791873-154791873 on Build GRCh38
rs782524498 Chr.X: 154791810-154791810 on Build GRCh38
rs782686305 Chr.X: 154791864-154791864 on Build GRCh38
rs781958435 Chr.X: 154791864-154791864 on Build GRCh38
rs960765689 Chr.X: 154792077-154792077 on Build GRCh38
rs992228306 Chr.X: 154792114-154792114 on Build GRCh38
rs980737292 Chr.X: 154792032-154792032 on Build GRCh38
rs1010205055 Chr.X: 154791954-154791954 on Build GRCh38
rs940551111 Chr.X: 154792124-154792124 on Build GRCh38
rs368500350 Chr.X: 154792132-154792132 on Build GRCh38
rs782259947 Chr.X: 154792120-154792120 on Build GRCh38
rs968414458 Chr.X: 154792019-154792019 on Build GRCh38
rs1033676469 Chr.X: 154792046-154792046 on Build GRCh38
rs375041440 Chr.X: 154792103-154792103 on Build GRCh38
rs782432122 Chr.X: 154791894-154791894 on Build GRCh38
rs1126762 Chr.X: 154791839-154791839 on Build GRCh38
rs190068634 Chr.X: 154791730-154791730 on Build GRCh38

View Full Product Details