Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.Y: 2975049-2975562 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFY
Entrez Gene ID
7544
Gene Name
zinc finger protein, Y-linked
Gene Aliases
ZNF911
UniGene
Hs.522845
Gene Chromosome Location
Chr.Y: 2934402-2982508 on Build GRCh38
Transcripts
NM_001145275.1 NM_001145276.1 NM_003411.3 XM_005262570.2 XM_011531473.2 XM_011531474.2 XM_011531475.2 XM_017030075.1 XM_017030076.1 XM_017030077.1

SNP Detail

SNP ID Chromosome Location
rs770565158 Chr.Y: 2975145-2975145 on Build GRCh38
rs931471187 Chr.Y: 2975156-2975156 on Build GRCh38
rs754997967 Chr.Y: 2975394-2975394 on Build GRCh38
rs1006899126 Chr.Y: 2975426-2975426 on Build GRCh38
rs776034863 Chr.Y: 2975167-2975167 on Build GRCh38
rs759155098 Chr.Y: 2975171-2975171 on Build GRCh38
rs769406147 Chr.Y: 2975240-2975240 on Build GRCh38
rs781323393 Chr.Y: 2975437-2975437 on Build GRCh38
rs762917941 Chr.Y: 2975519-2975519 on Build GRCh38
rs111915341 Chr.Y: 2975497-2975497 on Build GRCh38
rs372086650 Chr.Y: 2975164-2975164 on Build GRCh38
rs751493608 Chr.Y: 2975353-2975353 on Build GRCh38
rs897439284 Chr.Y: 2975110-2975110 on Build GRCh38
rs889853105 Chr.Y: 2975409-2975409 on Build GRCh38

View Full Product Details