Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 50804502-50805037 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
A1CF
Entrez Gene ID
29974
Gene Name
APOBEC1 complementation factor
Gene Aliases
ACF, ACF64, ACF65, APOBEC1CF, ASP
UniGene
Hs.282795
Gene Chromosome Location
Chr.10: 50799409-50885681 on Build GRCh38
Transcripts
NM_001198818.1 NM_001198819.1 NM_001198820.1 NM_014576.3 NM_138932.2 NM_138933.2 XM_005269718.2 XM_005269720.3 XM_011539729.2 XM_011539730.2 XM_017016160.1

SNP Detail

SNP ID Chromosome Location
rs1019292048 Chr.10: 50804583-50804583 on Build GRCh38
rs1045961016 Chr.10: 50804828-50804828 on Build GRCh38
rs1033626249 Chr.10: 50804572-50804572 on Build GRCh38
rs771797387 Chr.10: 50804927-50804927 on Build GRCh38
rs937474336 Chr.10: 50804918-50804918 on Build GRCh38
rs994453191 Chr.10: 50804528-50804528 on Build GRCh38
rs4619096 Chr.10: 50804661-50804661 on Build GRCh38
rs541821379 Chr.10: 50804683-50804683 on Build GRCh38
rs957483162 Chr.10: 50804731-50804731 on Build GRCh38
rs572153349 Chr.10: 50804865-50804865 on Build GRCh38
rs946053598 Chr.10: 50804821-50804821 on Build GRCh38
rs778506303 Chr.10: 50804621-50804621 on Build GRCh38
rs966494548 Chr.10: 50804711-50804711 on Build GRCh38
rs994566748 Chr.10: 50804987-50804987 on Build GRCh38
rs943556337 Chr.10: 50804816-50804816 on Build GRCh38
rs1027313769 Chr.10: 50804530-50804530 on Build GRCh38
rs912061765 Chr.10: 50804815-50804815 on Build GRCh38
rs1002110228 Chr.10: 50804531-50804531 on Build GRCh38
rs913261119 Chr.10: 50804768-50804768 on Build GRCh38
rs541265731 Chr.10: 50805007-50805007 on Build GRCh38
rs80080606 Chr.10: 50804764-50804764 on Build GRCh38
rs974575146 Chr.10: 50804660-50804660 on Build GRCh38
rs1004831478 Chr.10: 50804533-50804533 on Build GRCh38
rs35839127 Chr.10: 50804892-50804893 on Build GRCh38
rs1055921379 Chr.10: 50804958-50804958 on Build GRCh38
rs74874346 Chr.10: 50805008-50805008 on Build GRCh38
rs957664175 Chr.10: 50804654-50804654 on Build GRCh38
rs772726270 Chr.10: 50804984-50804984 on Build GRCh38
rs965882387 Chr.10: 50804636-50804636 on Build GRCh38

View Full Product Details