Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.Y: 9809223-9809718 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTTY22
Entrez Gene ID
252954
Gene Name
testis-specific transcript, Y-linked 22 (non-protein coding)
Gene Aliases
NCRNA00147
UniGene
Hs.511961
Gene Chromosome Location
Chr.Y: 9801153-9813245 on Build GRCh38
Transcripts
NR_001539.1

SNP Detail

SNP ID Chromosome Location
rs370922831 Chr.Y: 9809624-9809624 on Build GRCh38
rs72618768 Chr.Y: 9809371-9809371 on Build GRCh38

View Full Product Details