Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.Y: 20779417-20779921 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPS4Y2
Entrez Gene ID
140032
Gene Name
ribosomal protein S4, Y-linked 2
Gene Aliases
RPS4Y2P
UniGene
Hs.367761
Gene Chromosome Location
Chr.Y: 20756068-20781032 on Build GRCh38
Transcripts
NM_001039567.2

SNP Detail

SNP ID Chromosome Location
rs868795488 Chr.Y: 20779872-20779872 on Build GRCh38
rs759165354 Chr.Y: 20779547-20779547 on Build GRCh38
rs764861035 Chr.Y: 20779578-20779578 on Build GRCh38
rs762805360 Chr.Y: 20779690-20779690 on Build GRCh38
rs750177682 Chr.Y: 20779849-20779849 on Build GRCh38
rs761083480 Chr.Y: 20779479-20779479 on Build GRCh38
rs770552391 Chr.Y: 20779487-20779487 on Build GRCh38
rs957088501 Chr.Y: 20779763-20779763 on Build GRCh38
rs781733855 Chr.Y: 20779604-20779604 on Build GRCh38
rs773459948 Chr.Y: 20779475-20779475 on Build GRCh38
rs772561374 Chr.Y: 20779473-20779473 on Build GRCh38
rs776390587 Chr.Y: 20779543-20779543 on Build GRCh38
rs1031390349 Chr.Y: 20779619-20779619 on Build GRCh38

View Full Product Details