Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 22103031-22103553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VWA3A
Entrez Gene ID
146177
Gene Name
von Willebrand factor A domain containing 3A
Gene Aliases
-
UniGene
Hs.10697
Gene Chromosome Location
Chr.16: 22091930-22156966 on Build GRCh38
Transcripts
NM_173615.4 XM_011545742.2 XM_017022947.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3278319 c.452G>A p.R151K Chr.16: 22103498-22103498 on Build GRCh38
5470463 c.483+6G>A p.? Chr.16: 22103535-22103535 on Build GRCh38
3420846 c.480G>T p.K160N Chr.16: 22103526-22103526 on Build GRCh38
7423063 c.482A>T p.K161M Chr.16: 22103528-22103528 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs879745670 Chr.16: 22103370-22103371 on Build GRCh38
rs181235092 Chr.16: 22103283-22103283 on Build GRCh38
rs551662711 Chr.16: 22103236-22103236 on Build GRCh38
rs1011943813 Chr.16: 22103338-22103338 on Build GRCh38
rs1031554666 Chr.16: 22103173-22103173 on Build GRCh38
rs552091055 Chr.16: 22103364-22103364 on Build GRCh38
rs1001029889 Chr.16: 22103371-22103371 on Build GRCh38
rs139030402 Chr.16: 22103435-22103435 on Build GRCh38
rs554193063 Chr.16: 22103313-22103313 on Build GRCh38
rs1038169181 Chr.16: 22103304-22103304 on Build GRCh38
rs145635355 Chr.16: 22103276-22103276 on Build GRCh38
rs541359646 Chr.16: 22103509-22103509 on Build GRCh38
rs896135470 Chr.16: 22103326-22103326 on Build GRCh38
rs766484904 Chr.16: 22103521-22103521 on Build GRCh38
rs1033111155 Chr.16: 22103381-22103381 on Build GRCh38
rs183467108 Chr.16: 22103399-22103399 on Build GRCh38
rs1007608358 Chr.16: 22103286-22103286 on Build GRCh38
rs763860166 Chr.16: 22103436-22103436 on Build GRCh38
rs989003222 Chr.16: 22103392-22103392 on Build GRCh38
rs906589828 Chr.16: 22103066-22103066 on Build GRCh38
rs941421903 Chr.16: 22103439-22103439 on Build GRCh38
rs941190882 Chr.16: 22103284-22103284 on Build GRCh38
rs147339120 Chr.16: 22103345-22103345 on Build GRCh38
rs188143530 Chr.16: 22103479-22103479 on Build GRCh38
rs892629728 Chr.16: 22103177-22103177 on Build GRCh38
rs948056413 Chr.16: 22103419-22103419 on Build GRCh38
rs576517532 Chr.16: 22103406-22103406 on Build GRCh38
rs1023407672 Chr.16: 22103090-22103090 on Build GRCh38
rs978991837 Chr.16: 22103134-22103134 on Build GRCh38
rs1018714041 Chr.16: 22103294-22103294 on Build GRCh38
rs928080323 Chr.16: 22103480-22103480 on Build GRCh38
rs540083842 Chr.16: 22103314-22103314 on Build GRCh38
rs568932287 Chr.16: 22103156-22103156 on Build GRCh38
rs939468017 Chr.16: 22103489-22103489 on Build GRCh38
rs35723175 Chr.16: 22103280-22103281 on Build GRCh38
rs773728851 Chr.16: 22103424-22103424 on Build GRCh38
rs762085723 Chr.16: 22103324-22103324 on Build GRCh38
rs550828364 Chr.16: 22103261-22103261 on Build GRCh38
rs985933676 Chr.16: 22103282-22103282 on Build GRCh38
rs956256327 Chr.16: 22103179-22103179 on Build GRCh38
rs572397934 Chr.16: 22103496-22103496 on Build GRCh38
rs966145259 Chr.16: 22103303-22103303 on Build GRCh38
rs117491702 Chr.16: 22103194-22103194 on Build GRCh38
rs915802542 Chr.16: 22103402-22103402 on Build GRCh38
rs1003719287 Chr.16: 22103112-22103112 on Build GRCh38
rs1029896635 Chr.16: 22103397-22103397 on Build GRCh38

View Full Product Details