Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 27313526-27314064 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL4R
Entrez Gene ID
3566
Gene Name
interleukin 4 receptor
Gene Aliases
CD124, IL-4RA, IL4RA
UniGene
Hs.513457
Gene Chromosome Location
Chr.16: 27313668-27364778 on Build GRCh38
Transcripts
NM_000418.3 NM_001257406.1 NM_001257407.1 NM_001257997.1 XM_011545825.1 XM_011545826.2 XM_011545829.1 XM_011545831.1 XM_011545832.1 XM_017023211.1

SNP Detail

SNP ID Chromosome Location
rs1038690291 Chr.16: 27314024-27314024 on Build GRCh38
rs1047469198 Chr.16: 27313809-27313809 on Build GRCh38
rs372984959 Chr.16: 27313560-27313560 on Build GRCh38
rs1008505853 Chr.16: 27313973-27313973 on Build GRCh38
rs1008811607 Chr.16: 27313612-27313612 on Build GRCh38
rs890083097 Chr.16: 27313594-27313594 on Build GRCh38
rs8060798 Chr.16: 27313559-27313559 on Build GRCh38
rs55876407 Chr.16: 27313616-27313616 on Build GRCh38
rs79270619 Chr.16: 27313980-27313980 on Build GRCh38
rs890155104 Chr.16: 27313812-27313812 on Build GRCh38
rs55919742 Chr.16: 27313707-27313707 on Build GRCh38
rs199882945 Chr.16: 27313553-27313554 on Build GRCh38
rs35583025 Chr.16: 27313943-27313943 on Build GRCh38
rs55701409 Chr.16: 27313607-27313607 on Build GRCh38
rs528716128 Chr.16: 27313558-27313558 on Build GRCh38
rs55854826 Chr.16: 27313814-27313814 on Build GRCh38
rs997171964 Chr.16: 27314037-27314037 on Build GRCh38
rs750394942 Chr.16: 27313784-27313785 on Build GRCh38
rs1025942165 Chr.16: 27313591-27313591 on Build GRCh38
rs909322678 Chr.16: 27313881-27313881 on Build GRCh38
rs1038547102 Chr.16: 27313775-27313775 on Build GRCh38
rs927511108 Chr.16: 27313735-27313735 on Build GRCh38
rs992510503 Chr.16: 27313933-27313933 on Build GRCh38
rs56298792 Chr.16: 27313723-27313723 on Build GRCh38
rs993622831 Chr.16: 27313561-27313561 on Build GRCh38
rs8060938 Chr.16: 27313560-27313560 on Build GRCh38
rs376195936 Chr.16: 27313670-27313670 on Build GRCh38
rs12927172 Chr.16: 27313700-27313700 on Build GRCh38
rs112844743 Chr.16: 27313701-27313701 on Build GRCh38
rs754822955 Chr.16: 27313782-27313784 on Build GRCh38
rs1042223633 Chr.16: 27313978-27313978 on Build GRCh38
rs909279512 Chr.16: 27313613-27313613 on Build GRCh38
rs878863628 Chr.16: 27313849-27313849 on Build GRCh38
rs545839516 Chr.16: 27313780-27313781 on Build GRCh38
rs964939798 Chr.16: 27313810-27313810 on Build GRCh38
rs535327953 Chr.16: 27313884-27313884 on Build GRCh38
rs956064040 Chr.16: 27314014-27314014 on Build GRCh38
rs565226334 Chr.16: 27313781-27313782 on Build GRCh38
rs927243348 Chr.16: 27313910-27313910 on Build GRCh38
rs941583436 Chr.16: 27313743-27313743 on Build GRCh38
rs35276089 Chr.16: 27313558-27313559 on Build GRCh38
rs56054580 Chr.16: 27313751-27313751 on Build GRCh38
rs386790133 Chr.16: 27313559-27313560 on Build GRCh38
rs1031029266 Chr.16: 27314045-27314045 on Build GRCh38
rs35311840 Chr.16: 27313979-27313979 on Build GRCh38
rs900155335 Chr.16: 27314030-27314030 on Build GRCh38
rs978600872 Chr.16: 27313647-27313647 on Build GRCh38
rs566296906 Chr.16: 27313781-27313781 on Build GRCh38
rs1056306972 Chr.16: 27313686-27313686 on Build GRCh38
rs980378650 Chr.16: 27313726-27313726 on Build GRCh38
rs1044408714 Chr.16: 27313624-27313624 on Build GRCh38
rs950971941 Chr.16: 27313844-27313852 on Build GRCh38
rs879703995 Chr.16: 27313773-27313773 on Build GRCh38
rs964932681 Chr.16: 27313621-27313621 on Build GRCh38
rs865841439 Chr.16: 27313626-27313626 on Build GRCh38
rs765857740 Chr.16: 27313659-27313659 on Build GRCh38
rs929071376 Chr.16: 27313796-27313796 on Build GRCh38
rs1038952741 Chr.16: 27313975-27313975 on Build GRCh38
rs550744399 Chr.16: 27313676-27313676 on Build GRCh38
rs750606512 Chr.16: 27314020-27314020 on Build GRCh38
rs575497917 Chr.16: 27313969-27313969 on Build GRCh38
rs997695743 Chr.16: 27313835-27313835 on Build GRCh38
rs937922904 Chr.16: 27313921-27313921 on Build GRCh38
rs963454197 Chr.16: 27313891-27313891 on Build GRCh38
rs879476013 Chr.16: 27313559-27313560 on Build GRCh38
rs55885488 Chr.16: 27313685-27313685 on Build GRCh38
rs980099380 Chr.16: 27313903-27313903 on Build GRCh38
rs1032853198 Chr.16: 27314008-27314008 on Build GRCh38
rs983625214 Chr.16: 27313877-27313877 on Build GRCh38
rs12927543 Chr.16: 27313702-27313702 on Build GRCh38
rs1030518599 Chr.16: 27313843-27313843 on Build GRCh38
rs530910297 Chr.16: 27313565-27313565 on Build GRCh38
rs796489515 Chr.16: 27313700-27313702 on Build GRCh38
rs1000492520 Chr.16: 27314003-27314003 on Build GRCh38

View Full Product Details