Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12188656-12189217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF136
Entrez Gene ID
7695
Gene Name
zinc finger protein 136
Gene Aliases
pHZ-20
UniGene
Hs.479874
Gene Chromosome Location
Chr.19: 12162672-12189880 on Build GRCh38
Transcripts
NM_003437.3 XM_011528269.2 XM_017027243.1

SNP Detail

SNP ID Chromosome Location
rs773914931 Chr.19: 12188844-12188844 on Build GRCh38
rs368640791 Chr.19: 12188741-12188741 on Build GRCh38
rs546485772 Chr.19: 12188885-12188885 on Build GRCh38
rs772264325 Chr.19: 12189031-12189032 on Build GRCh38
rs563913225 Chr.19: 12188830-12188830 on Build GRCh38
rs531108413 Chr.19: 12188862-12188862 on Build GRCh38
rs779530261 Chr.19: 12188774-12188774 on Build GRCh38
rs775442163 Chr.19: 12189102-12189102 on Build GRCh38
rs747628495 Chr.19: 12188680-12188680 on Build GRCh38
rs899561149 Chr.19: 12188889-12188889 on Build GRCh38
rs777087355 Chr.19: 12188951-12188951 on Build GRCh38
rs1047949774 Chr.19: 12189075-12189075 on Build GRCh38
rs575584195 Chr.19: 12188809-12188809 on Build GRCh38
rs1033387108 Chr.19: 12189117-12189117 on Build GRCh38
rs746011908 Chr.19: 12188828-12188828 on Build GRCh38
rs527316486 Chr.19: 12188741-12188741 on Build GRCh38
rs563891619 Chr.19: 12189047-12189047 on Build GRCh38
rs914140555 Chr.19: 12189151-12189151 on Build GRCh38
rs917788005 Chr.19: 12188774-12188775 on Build GRCh38
rs967013993 Chr.19: 12188789-12188789 on Build GRCh38
rs980888235 Chr.19: 12189072-12189072 on Build GRCh38
rs897800037 Chr.19: 12189022-12189022 on Build GRCh38
rs892142279 Chr.19: 12188678-12188678 on Build GRCh38
rs1014899217 Chr.19: 12188730-12188730 on Build GRCh38
rs1013987073 Chr.19: 12189033-12189033 on Build GRCh38
rs981345968 Chr.19: 12188818-12188818 on Build GRCh38
rs997069381 Chr.19: 12188719-12188719 on Build GRCh38
rs958220227 Chr.19: 12189116-12189116 on Build GRCh38
rs555822590 Chr.19: 12188770-12188770 on Build GRCh38
rs45563835 Chr.19: 12189087-12189087 on Build GRCh38
rs189664381 Chr.19: 12189186-12189186 on Build GRCh38
rs201633686 Chr.19: 12188740-12188740 on Build GRCh38
rs1004165524 Chr.19: 12188728-12188728 on Build GRCh38
rs185193915 Chr.19: 12189131-12189131 on Build GRCh38
rs1004093427 Chr.19: 12189002-12189002 on Build GRCh38
rs747515267 Chr.19: 12188846-12188846 on Build GRCh38
rs935987611 Chr.19: 12188876-12188876 on Build GRCh38
rs1026841384 Chr.19: 12188721-12188721 on Build GRCh38
rs899634095 Chr.19: 12188684-12188684 on Build GRCh38
rs925508674 Chr.19: 12188835-12188835 on Build GRCh38
rs936893315 Chr.19: 12188837-12188837 on Build GRCh38
rs537828144 Chr.19: 12188871-12188871 on Build GRCh38
rs1041213007 Chr.19: 12188681-12188681 on Build GRCh38
rs879295381 Chr.19: 12188793-12188794 on Build GRCh38
rs542427921 Chr.19: 12188823-12188823 on Build GRCh38
rs368046689 Chr.19: 12188794-12188794 on Build GRCh38
rs1005848338 Chr.19: 12189098-12189098 on Build GRCh38
rs759706737 Chr.19: 12188996-12188996 on Build GRCh38
rs142656100 Chr.19: 12188731-12188741 on Build GRCh38
rs199892108 Chr.19: 12188740-12188741 on Build GRCh38
rs1015537143 Chr.19: 12189014-12189014 on Build GRCh38
rs929483933 Chr.19: 12189071-12189073 on Build GRCh38
rs772472499 Chr.19: 12188691-12188691 on Build GRCh38
rs952602049 Chr.19: 12188722-12188722 on Build GRCh38
rs969871444 Chr.19: 12189067-12189067 on Build GRCh38
rs991987949 Chr.19: 12188773-12188773 on Build GRCh38
rs575016064 Chr.19: 12188689-12188689 on Build GRCh38
rs932456087 Chr.19: 12188945-12188945 on Build GRCh38
rs1025467496 Chr.19: 12188755-12188755 on Build GRCh38
rs913189076 Chr.19: 12188906-12188906 on Build GRCh38
rs910596120 Chr.19: 12188915-12188915 on Build GRCh38
rs1040742691 Chr.19: 12188886-12188886 on Build GRCh38
rs1006315278 Chr.19: 12188720-12188720 on Build GRCh38
rs962040091 Chr.19: 12188737-12188737 on Build GRCh38
rs969525344 Chr.19: 12188806-12188806 on Build GRCh38

View Full Product Details